Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55161851-55162346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF3
Entrez Gene ID
352909
Gene Name
dynein axonemal assembly factor 3
Gene Aliases
C19orf51, CILD2, DAB1, PCD, PF22
UniGene
Hs.351582
Gene Chromosome Location
Chr.19: 55158661-55166722 on Build GRCh38
Transcripts
NM_001256714.1 NM_001256715.1 NM_001256716.1 NM_178837.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6209980 c.522C>T p.A174A Chr.19: 55162232-55162232 on Build GRCh38
6209982 c.219C>T p.A73A Chr.19: 55162232-55162232 on Build GRCh38
4593515 c.179T>G p.L60R Chr.19: 55162272-55162272 on Build GRCh38
4634462 c.196G>A p.A66T Chr.19: 55162255-55162255 on Build GRCh38
4591797 c.488T>G p.V163G Chr.19: 55162266-55162266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048936417 Chr.19: 55162241-55162241 on Build GRCh38
rs919914917 Chr.19: 55162217-55162217 on Build GRCh38
rs928846699 Chr.19: 55162188-55162188 on Build GRCh38
rs562110613 Chr.19: 55162199-55162199 on Build GRCh38
rs567554040 Chr.19: 55162311-55162311 on Build GRCh38
rs10407660 Chr.19: 55162307-55162307 on Build GRCh38
rs375786609 Chr.19: 55162315-55162315 on Build GRCh38
rs1018825953 Chr.19: 55162079-55162079 on Build GRCh38
rs372945469 Chr.19: 55162147-55162147 on Build GRCh38
rs372753112 Chr.19: 55162094-55162094 on Build GRCh38
rs528126745 Chr.19: 55162226-55162226 on Build GRCh38
rs3848618 Chr.19: 55162286-55162286 on Build GRCh38
rs1045752118 Chr.19: 55161909-55161909 on Build GRCh38
rs779712863 Chr.19: 55162134-55162134 on Build GRCh38
rs901703224 Chr.19: 55162280-55162280 on Build GRCh38
rs539059776 Chr.19: 55162078-55162078 on Build GRCh38
rs567514867 Chr.19: 55161924-55161924 on Build GRCh38
rs924519062 Chr.19: 55162121-55162121 on Build GRCh38
rs773052536 Chr.19: 55162210-55162210 on Build GRCh38
rs201986299 Chr.19: 55162294-55162294 on Build GRCh38
rs946234784 Chr.19: 55161886-55161886 on Build GRCh38
rs192924578 Chr.19: 55162259-55162259 on Build GRCh38
rs574725370 Chr.19: 55162143-55162143 on Build GRCh38
rs1013125499 Chr.19: 55162219-55162219 on Build GRCh38
rs751887577 Chr.19: 55162119-55162119 on Build GRCh38
rs931283937 Chr.19: 55162186-55162186 on Build GRCh38
rs920046859 Chr.19: 55162177-55162177 on Build GRCh38
rs759904558 Chr.19: 55162235-55162235 on Build GRCh38
rs201905107 Chr.19: 55162095-55162095 on Build GRCh38
rs1021364861 Chr.19: 55162087-55162087 on Build GRCh38
rs931354583 Chr.19: 55162317-55162317 on Build GRCh38
rs765396737 Chr.19: 55162255-55162255 on Build GRCh38
rs551100219 Chr.19: 55162250-55162250 on Build GRCh38
rs898519728 Chr.19: 55161976-55161976 on Build GRCh38
rs561398611 Chr.19: 55162142-55162142 on Build GRCh38
rs375739321 Chr.19: 55161941-55161941 on Build GRCh38
rs747667670 Chr.19: 55162194-55162194 on Build GRCh38
rs369116839 Chr.19: 55162144-55162144 on Build GRCh38
rs994736556 Chr.19: 55161987-55161987 on Build GRCh38
rs3848617 Chr.19: 55161877-55161877 on Build GRCh38
rs770403610 Chr.19: 55162263-55162263 on Build GRCh38
rs968469725 Chr.19: 55162232-55162232 on Build GRCh38
rs76267081 Chr.19: 55161961-55161961 on Build GRCh38
rs957023679 Chr.19: 55162310-55162310 on Build GRCh38
rs749955680 Chr.19: 55161962-55161962 on Build GRCh38
rs550592244 Chr.19: 55161882-55161882 on Build GRCh38
rs573343395 Chr.19: 55162031-55162031 on Build GRCh38
rs771929224 Chr.19: 55162209-55162209 on Build GRCh38
rs553540882 Chr.19: 55162026-55162026 on Build GRCh38
rs35599151 Chr.19: 55161885-55161886 on Build GRCh38
rs1011667749 Chr.19: 55162312-55162312 on Build GRCh38
rs558869777 Chr.19: 55162083-55162083 on Build GRCh38
rs757597159 Chr.19: 55162128-55162128 on Build GRCh38
rs559042351 Chr.19: 55162215-55162215 on Build GRCh38
rs770679152 Chr.19: 55162222-55162222 on Build GRCh38
rs886236180 Chr.19: 55162041-55162041 on Build GRCh38
rs1004727762 Chr.19: 55162075-55162075 on Build GRCh38
rs549295806 Chr.19: 55162295-55162295 on Build GRCh38
rs532034805 Chr.19: 55161881-55161881 on Build GRCh38
rs958918715 Chr.19: 55162030-55162030 on Build GRCh38
rs1056296743 Chr.19: 55162211-55162211 on Build GRCh38
rs796864871 Chr.19: 55162124-55162124 on Build GRCh38
rs1040183266 Chr.19: 55162165-55162165 on Build GRCh38
rs981566868 Chr.19: 55162187-55162187 on Build GRCh38
rs868815876 Chr.19: 55162264-55162264 on Build GRCh38
rs1040210720 Chr.19: 55162265-55162265 on Build GRCh38

View Full Product Details