Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53218094-53218605 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1orf123
Entrez Gene ID
54987
Gene Name
chromosome 1 open reading frame 123
Gene Aliases
-
UniGene
Hs.525391
Gene Chromosome Location
Chr.1: 53214099-53220639 on Build GRCh38
Transcripts
NM_001304759.1 NM_001304760.1 NM_017887.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3419321 c.261+2T>C p.? Chr.1: 53218170-53218170 on Build GRCh38
7358216 c.187A>G p.M63V Chr.1: 53218456-53218456 on Build GRCh38
7076406 c.230-1G>C p.? Chr.1: 53218204-53218204 on Build GRCh38
5846973 c.229+1G>A p.? Chr.1: 53218413-53218413 on Build GRCh38
3490738 c.186C>T p.S62S Chr.1: 53218457-53218457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745990853 Chr.1: 53218179-53218179 on Build GRCh38
rs770439959 Chr.1: 53218411-53218411 on Build GRCh38
rs200900856 Chr.1: 53218528-53218528 on Build GRCh38
rs72673130 Chr.1: 53218236-53218236 on Build GRCh38
rs189510666 Chr.1: 53218473-53218473 on Build GRCh38
rs756188383 Chr.1: 53218387-53218387 on Build GRCh38
rs779495665 Chr.1: 53218483-53218483 on Build GRCh38
rs544562776 Chr.1: 53218192-53218192 on Build GRCh38
rs768999922 Chr.1: 53218190-53218190 on Build GRCh38
rs762965134 Chr.1: 53218438-53218438 on Build GRCh38
rs751726732 Chr.1: 53218171-53218171 on Build GRCh38
rs555568109 Chr.1: 53218552-53218552 on Build GRCh38
rs199913224 Chr.1: 53218403-53218403 on Build GRCh38
rs760221907 Chr.1: 53218215-53218215 on Build GRCh38
rs373880930 Chr.1: 53218538-53218538 on Build GRCh38
rs201416768 Chr.1: 53218500-53218500 on Build GRCh38
rs185347619 Chr.1: 53218339-53218339 on Build GRCh38
rs777985329 Chr.1: 53218162-53218162 on Build GRCh38
rs11546995 Chr.1: 53218444-53218444 on Build GRCh38
rs1010450490 Chr.1: 53218265-53218265 on Build GRCh38
rs757561191 Chr.1: 53218173-53218173 on Build GRCh38
rs763801420 Chr.1: 53218541-53218542 on Build GRCh38
rs145021398 Chr.1: 53218418-53218418 on Build GRCh38
rs145124211 Chr.1: 53218462-53218462 on Build GRCh38
rs376808171 Chr.1: 53218122-53218122 on Build GRCh38
rs767565531 Chr.1: 53218461-53218461 on Build GRCh38
rs774833856 Chr.1: 53218197-53218197 on Build GRCh38
rs113038301 Chr.1: 53218374-53218374 on Build GRCh38
rs750456320 Chr.1: 53218381-53218381 on Build GRCh38
rs780171532 Chr.1: 53218397-53218397 on Build GRCh38
rs749762985 Chr.1: 53218206-53218207 on Build GRCh38
rs776437572 Chr.1: 53218414-53218414 on Build GRCh38
rs146156858 Chr.1: 53218305-53218305 on Build GRCh38
rs753436579 Chr.1: 53218136-53218136 on Build GRCh38
rs770249204 Chr.1: 53218186-53218186 on Build GRCh38
rs761694363 Chr.1: 53218460-53218460 on Build GRCh38
rs560506317 Chr.1: 53218285-53218285 on Build GRCh38
rs200743226 Chr.1: 53218145-53218145 on Build GRCh38
rs375523271 Chr.1: 53218410-53218410 on Build GRCh38
rs763050513 Chr.1: 53218250-53218250 on Build GRCh38
rs866608378 Chr.1: 53218298-53218298 on Build GRCh38
rs752607452 Chr.1: 53218304-53218304 on Build GRCh38
rs745315369 Chr.1: 53218415-53218415 on Build GRCh38
rs576038288 Chr.1: 53218398-53218398 on Build GRCh38
rs775961420 Chr.1: 53218235-53218235 on Build GRCh38
rs758283704 Chr.1: 53218159-53218159 on Build GRCh38
rs902586910 Chr.1: 53218284-53218284 on Build GRCh38
rs920814813 Chr.1: 53218424-53218424 on Build GRCh38
rs765296408 Chr.1: 53218141-53218141 on Build GRCh38
rs1053883708 Chr.1: 53218527-53218527 on Build GRCh38
rs775007939 Chr.1: 53218372-53218372 on Build GRCh38
rs781113815 Chr.1: 53218175-53218175 on Build GRCh38
rs781510282 Chr.1: 53218375-53218375 on Build GRCh38
rs755553635 Chr.1: 53218399-53218399 on Build GRCh38
rs780548676 Chr.1: 53218188-53218188 on Build GRCh38
rs562231137 Chr.1: 53218471-53218471 on Build GRCh38
rs765486105 Chr.1: 53218293-53218293 on Build GRCh38
rs113531134 Chr.1: 53218482-53218482 on Build GRCh38
rs771488417 Chr.1: 53218213-53218214 on Build GRCh38
rs760437711 Chr.1: 53218123-53218123 on Build GRCh38
rs79453977 Chr.1: 53218327-53218327 on Build GRCh38
rs141056335 Chr.1: 53218456-53218456 on Build GRCh38
rs145613337 Chr.1: 53218463-53218463 on Build GRCh38
rs200312423 Chr.1: 53218229-53218229 on Build GRCh38
rs1012254961 Chr.1: 53218534-53218534 on Build GRCh38
rs12140585 Chr.1: 53218249-53218249 on Build GRCh38
rs748574766 Chr.1: 53218407-53218407 on Build GRCh38
rs906441119 Chr.1: 53218556-53218556 on Build GRCh38
rs765789702 Chr.1: 53218125-53218125 on Build GRCh38
rs760163030 Chr.1: 53218473-53218473 on Build GRCh38
rs769621068 Chr.1: 53218539-53218539 on Build GRCh38
rs759513664 Chr.1: 53218137-53218137 on Build GRCh38
rs75176256 Chr.1: 53218340-53218340 on Build GRCh38
rs776731664 Chr.1: 53218180-53218180 on Build GRCh38
rs755399907 Chr.1: 53218474-53218474 on Build GRCh38
rs1039275714 Chr.1: 53218217-53218217 on Build GRCh38
rs775112921 Chr.1: 53218540-53218540 on Build GRCh38
rs191880219 Chr.1: 53218546-53218546 on Build GRCh38
rs999648919 Chr.1: 53218291-53218291 on Build GRCh38
rs149527063 Chr.1: 53218470-53218470 on Build GRCh38
rs112803850 Chr.1: 53218413-53218413 on Build GRCh38
rs142980878 Chr.1: 53218434-53218434 on Build GRCh38

View Full Product Details