Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 54653030-54653523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MROH7
Entrez Gene ID
374977
Gene Name
maestro heat like repeat family member 7
Gene Aliases
C1orf175, HEATR8
UniGene
Hs.412482
Gene Chromosome Location
Chr.1: 54641740-54710267 on Build GRCh38
Transcripts
NM_001039464.3 NM_001291332.1 NR_026782.2 NR_111931.1 NR_037639.2 NR_037640.2 NR_037641.2

Gene Symbol
MROH7-TTC4
Entrez Gene ID
100527960
Gene Name
MROH7-TTC4 readthrough (NMD candidate)
Gene Aliases
HEATR8-TTC4
UniGene
Gene Chromosome Location
Chr.1: 54641740-54742657 on Build GRCh38
Transcripts
NM_001039464.3 NM_001291332.1 NR_026782.2 NR_111931.1 NR_037639.2 NR_037640.2 NR_037641.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6715211 c.283C>T p.Q95* Chr.1: 54653209-54653209 on Build GRCh38
4187234 c.248C>G p.P83R Chr.1: 54653174-54653174 on Build GRCh38
3490886 c.368C>T p.P123L Chr.1: 54653294-54653294 on Build GRCh38
5631633 c.338C>G p.S113* Chr.1: 54653264-54653264 on Build GRCh38
4515332 c.103_104CC>TT p.P35F Chr.1: 54653029-54653030 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150191046 Chr.1: 54653117-54653117 on Build GRCh38
rs577982847 Chr.1: 54653095-54653095 on Build GRCh38
rs374996813 Chr.1: 54653080-54653080 on Build GRCh38
rs184477025 Chr.1: 54653294-54653294 on Build GRCh38
rs748424041 Chr.1: 54653178-54653178 on Build GRCh38
rs750119628 Chr.1: 54653339-54653339 on Build GRCh38
rs778567344 Chr.1: 54653381-54653381 on Build GRCh38
rs752529386 Chr.1: 54653386-54653386 on Build GRCh38
rs771891230 Chr.1: 54653332-54653332 on Build GRCh38
rs765791852 Chr.1: 54653499-54653499 on Build GRCh38
rs1038809482 Chr.1: 54653062-54653062 on Build GRCh38
rs941746170 Chr.1: 54653271-54653271 on Build GRCh38
rs970158617 Chr.1: 54653411-54653411 on Build GRCh38
rs982279895 Chr.1: 54653473-54653474 on Build GRCh38
rs758597398 Chr.1: 54653389-54653389 on Build GRCh38
rs761575153 Chr.1: 54653241-54653241 on Build GRCh38
rs758037622 Chr.1: 54653125-54653125 on Build GRCh38
rs776964535 Chr.1: 54653298-54653298 on Build GRCh38
rs765668531 Chr.1: 54653227-54653227 on Build GRCh38
rs751249059 Chr.1: 54653142-54653142 on Build GRCh38
rs777924560 Chr.1: 54653398-54653398 on Build GRCh38
rs192479874 Chr.1: 54653159-54653159 on Build GRCh38
rs754223717 Chr.1: 54653122-54653122 on Build GRCh38
rs530118000 Chr.1: 54653333-54653333 on Build GRCh38
rs746351821 Chr.1: 54653467-54653467 on Build GRCh38
rs1053493689 Chr.1: 54653317-54653317 on Build GRCh38
rs769997976 Chr.1: 54653323-54653323 on Build GRCh38
rs776059628 Chr.1: 54653329-54653329 on Build GRCh38
rs757219968 Chr.1: 54653443-54653443 on Build GRCh38
rs981667870 Chr.1: 54653212-54653212 on Build GRCh38
rs950999890 Chr.1: 54653477-54653477 on Build GRCh38
rs547740508 Chr.1: 54653057-54653057 on Build GRCh38
rs768796455 Chr.1: 54653088-54653088 on Build GRCh38
rs753658186 Chr.1: 54653376-54653376 on Build GRCh38
rs775165440 Chr.1: 54653079-54653079 on Build GRCh38
rs375679935 Chr.1: 54653193-54653193 on Build GRCh38
rs760749304 Chr.1: 54653335-54653335 on Build GRCh38
rs568753329 Chr.1: 54653163-54653163 on Build GRCh38
rs369773156 Chr.1: 54653198-54653198 on Build GRCh38
rs1051630276 Chr.1: 54653295-54653295 on Build GRCh38
rs1023048326 Chr.1: 54653410-54653410 on Build GRCh38
rs1011622435 Chr.1: 54653380-54653380 on Build GRCh38
rs769649902 Chr.1: 54653078-54653078 on Build GRCh38
rs745834215 Chr.1: 54653065-54653065 on Build GRCh38
rs749083568 Chr.1: 54653172-54653172 on Build GRCh38
rs760386537 Chr.1: 54653496-54653496 on Build GRCh38
rs758045075 Chr.1: 54653049-54653049 on Build GRCh38
rs1023099190 Chr.1: 54653205-54653205 on Build GRCh38
rs779805295 Chr.1: 54653261-54653261 on Build GRCh38
rs374706424 Chr.1: 54653379-54653379 on Build GRCh38
rs765916521 Chr.1: 54653366-54653366 on Build GRCh38
rs756094958 Chr.1: 54653259-54653259 on Build GRCh38
rs368503437 Chr.1: 54653385-54653385 on Build GRCh38
rs781348564 Chr.1: 54653461-54653461 on Build GRCh38
rs777333204 Chr.1: 54653055-54653055 on Build GRCh38
rs377717151 Chr.1: 54653211-54653211 on Build GRCh38
rs751192948 Chr.1: 54653231-54653231 on Build GRCh38
rs375822785 Chr.1: 54653387-54653387 on Build GRCh38
rs767613226 Chr.1: 54653106-54653106 on Build GRCh38
rs764535178 Chr.1: 54653208-54653215 on Build GRCh38
rs761001348 Chr.1: 54653146-54653146 on Build GRCh38
rs763704539 Chr.1: 54653139-54653139 on Build GRCh38
rs531104510 Chr.1: 54653285-54653285 on Build GRCh38
rs760060392 Chr.1: 54653363-54653363 on Build GRCh38
rs747264454 Chr.1: 54653403-54653403 on Build GRCh38
rs370342214 Chr.1: 54653242-54653242 on Build GRCh38
rs756183210 Chr.1: 54653164-54653164 on Build GRCh38
rs370379031 Chr.1: 54653338-54653338 on Build GRCh38
rs908886897 Chr.1: 54653256-54653256 on Build GRCh38
rs551587041 Chr.1: 54653483-54653483 on Build GRCh38
rs771382138 Chr.1: 54653204-54653204 on Build GRCh38
rs771067754 Chr.1: 54653297-54653297 on Build GRCh38
rs898394974 Chr.1: 54653420-54653420 on Build GRCh38
rs566978822 Chr.1: 54653470-54653470 on Build GRCh38
rs754661318 Chr.1: 54653282-54653282 on Build GRCh38
rs376881648 Chr.1: 54653108-54653108 on Build GRCh38
rs11206407 Chr.1: 54653471-54653471 on Build GRCh38
rs373089784 Chr.1: 54653284-54653284 on Build GRCh38
rs749990498 Chr.1: 54653255-54653255 on Build GRCh38
rs951015500 Chr.1: 54653219-54653219 on Build GRCh38
rs138958132 Chr.1: 54653188-54653188 on Build GRCh38
rs201689263 Chr.1: 54653309-54653309 on Build GRCh38
rs759771708 Chr.1: 54653217-54653217 on Build GRCh38
rs545216139 Chr.1: 54653283-54653283 on Build GRCh38
rs779864108 Chr.1: 54653165-54653165 on Build GRCh38
rs777159695 Chr.1: 54653333-54653333 on Build GRCh38
rs1055091560 Chr.1: 54653260-54653260 on Build GRCh38
rs75579421 Chr.1: 54653199-54653199 on Build GRCh38
rs749616801 Chr.1: 54653481-54653481 on Build GRCh38
rs781230144 Chr.1: 54653064-54653064 on Build GRCh38
rs761828972 Chr.1: 54653098-54653098 on Build GRCh38
rs756790243 Chr.1: 54653147-54653147 on Build GRCh38
rs762228723 Chr.1: 54653133-54653133 on Build GRCh38

View Full Product Details