Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 46480298-46480789 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM247
Entrez Gene ID
388946
Gene Name
transmembrane protein 247
Gene Aliases
-
UniGene
Hs.733348
Gene Chromosome Location
Chr.2: 46479565-46484425 on Build GRCh38
Transcripts
NM_001145051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3695518 c.401T>C p.V134A Chr.2: 46480688-46480688 on Build GRCh38
3695519 c.467C>T p.A156V Chr.2: 46480754-46480754 on Build GRCh38
5790364 c.256G>A p.G86R Chr.2: 46480543-46480543 on Build GRCh38
3391745 c.241G>C p.G81R Chr.2: 46480528-46480528 on Build GRCh38
5889823 c.242G>A p.G81D Chr.2: 46480529-46480529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000983770 Chr.2: 46480460-46480460 on Build GRCh38
rs1034053027 Chr.2: 46480574-46480574 on Build GRCh38
rs72875605 Chr.2: 46480559-46480559 on Build GRCh38
rs12997610 Chr.2: 46480608-46480608 on Build GRCh38
rs1020050838 Chr.2: 46480542-46480542 on Build GRCh38
rs536082369 Chr.2: 46480323-46480323 on Build GRCh38
rs750901510 Chr.2: 46480749-46480750 on Build GRCh38
rs201742486 Chr.2: 46480745-46480745 on Build GRCh38
rs372990692 Chr.2: 46480578-46480578 on Build GRCh38
rs909945854 Chr.2: 46480499-46480499 on Build GRCh38
rs778735967 Chr.2: 46480385-46480385 on Build GRCh38
rs566145397 Chr.2: 46480626-46480626 on Build GRCh38
rs758072503 Chr.2: 46480740-46480740 on Build GRCh38
rs549670167 Chr.2: 46480616-46480616 on Build GRCh38
rs770800896 Chr.2: 46480497-46480497 on Build GRCh38
rs771659307 Chr.2: 46480672-46480673 on Build GRCh38
rs941408129 Chr.2: 46480509-46480509 on Build GRCh38
rs751904338 Chr.2: 46480553-46480553 on Build GRCh38
rs756013142 Chr.2: 46480751-46480751 on Build GRCh38
rs1005528831 Chr.2: 46480349-46480349 on Build GRCh38
rs139055089 Chr.2: 46480456-46480456 on Build GRCh38
rs769823616 Chr.2: 46480680-46480680 on Build GRCh38
rs748587672 Chr.2: 46480374-46480374 on Build GRCh38
rs752362754 Chr.2: 46480599-46480599 on Build GRCh38
rs768940112 Chr.2: 46480699-46480699 on Build GRCh38
rs774750926 Chr.2: 46480756-46480756 on Build GRCh38
rs951639490 Chr.2: 46480755-46480755 on Build GRCh38
rs762178494 Chr.2: 46480745-46480748 on Build GRCh38
rs906036393 Chr.2: 46480670-46480708 on Build GRCh38
rs74318890 Chr.2: 46480669-46480669 on Build GRCh38
rs775332040 Chr.2: 46480673-46480674 on Build GRCh38
rs770888776 Chr.2: 46480640-46480640 on Build GRCh38
rs530468852 Chr.2: 46480584-46480584 on Build GRCh38
rs983937840 Chr.2: 46480589-46480589 on Build GRCh38
rs781149530 Chr.2: 46480659-46480659 on Build GRCh38
rs746955772 Chr.2: 46480496-46480496 on Build GRCh38
rs549582359 Chr.2: 46480328-46480328 on Build GRCh38
rs1007387595 Chr.2: 46480689-46480689 on Build GRCh38
rs577968366 Chr.2: 46480388-46480388 on Build GRCh38
rs769027858 Chr.2: 46480754-46480754 on Build GRCh38
rs917392074 Chr.2: 46480316-46480316 on Build GRCh38
rs965793198 Chr.2: 46480713-46480713 on Build GRCh38
rs879814156 Chr.2: 46480669-46480670 on Build GRCh38
rs921279567 Chr.2: 46480635-46480635 on Build GRCh38
rs757833783 Chr.2: 46480453-46480453 on Build GRCh38
rs775635461 Chr.2: 46480686-46480686 on Build GRCh38
rs761243026 Chr.2: 46480725-46480725 on Build GRCh38
rs941461614 Chr.2: 46480624-46480624 on Build GRCh38
rs550572145 Chr.2: 46480602-46480602 on Build GRCh38
rs368909438 Chr.2: 46480539-46480539 on Build GRCh38
rs972757288 Chr.2: 46480715-46480715 on Build GRCh38
rs968461693 Chr.2: 46480560-46480560 on Build GRCh38
rs780009765 Chr.2: 46480752-46480752 on Build GRCh38
rs748355604 Chr.2: 46480758-46480758 on Build GRCh38
rs757093481 Chr.2: 46480746-46480746 on Build GRCh38
rs765666900 Chr.2: 46480749-46480749 on Build GRCh38
rs764620057 Chr.2: 46480552-46480552 on Build GRCh38
rs1019662199 Chr.2: 46480708-46480708 on Build GRCh38
rs61995902 Chr.2: 46480712-46480712 on Build GRCh38
rs764662107 Chr.2: 46480734-46480734 on Build GRCh38
rs752197440 Chr.2: 46480735-46480735 on Build GRCh38
rs116983452 Chr.2: 46480535-46480535 on Build GRCh38
rs764530259 Chr.2: 46480411-46480411 on Build GRCh38
rs72875604 Chr.2: 46480381-46480381 on Build GRCh38
rs772620209 Chr.2: 46480759-46480759 on Build GRCh38
rs542450747 Chr.2: 46480532-46480532 on Build GRCh38
rs550741414 Chr.2: 46480700-46480700 on Build GRCh38
rs537077243 Chr.2: 46480706-46480706 on Build GRCh38
rs12612916 Chr.2: 46480747-46480747 on Build GRCh38
rs968366552 Chr.2: 46480428-46480428 on Build GRCh38
rs774282506 Chr.2: 46480563-46480564 on Build GRCh38
rs556923418 Chr.2: 46480741-46480741 on Build GRCh38
rs70940616 Chr.2: 46480744-46480745 on Build GRCh38
rs889926929 Chr.2: 46480672-46480672 on Build GRCh38
rs1057190181 Chr.2: 46480399-46480399 on Build GRCh38
rs564004743 Chr.2: 46480613-46480613 on Build GRCh38
rs1009942383 Chr.2: 46480420-46480420 on Build GRCh38
rs978425491 Chr.2: 46480446-46480446 on Build GRCh38
rs1052423 Chr.2: 46480667-46480667 on Build GRCh38
rs764621923 Chr.2: 46480739-46480740 on Build GRCh38
rs917476729 Chr.2: 46480594-46480594 on Build GRCh38
rs868569085 Chr.2: 46480657-46480657 on Build GRCh38
rs547792479 Chr.2: 46480615-46480615 on Build GRCh38
rs912507042 Chr.2: 46480333-46480333 on Build GRCh38
rs879115276 Chr.2: 46480682-46480682 on Build GRCh38
rs1034474473 Chr.2: 46480732-46480732 on Build GRCh38
rs72875603 Chr.2: 46480340-46480340 on Build GRCh38
rs141185381 Chr.2: 46480329-46480329 on Build GRCh38
rs775624069 Chr.2: 46480538-46480538 on Build GRCh38
rs779714419 Chr.2: 46480454-46480454 on Build GRCh38
rs753138071 Chr.2: 46480398-46480398 on Build GRCh38
rs564981383 Chr.2: 46480573-46480573 on Build GRCh38
rs762423075 Chr.2: 46480555-46480555 on Build GRCh38
rs753417832 Chr.2: 46480562-46480562 on Build GRCh38
rs61995899 Chr.2: 46480570-46480570 on Build GRCh38
rs61995900 Chr.2: 46480637-46480637 on Build GRCh38
rs776628047 Chr.2: 46480373-46480373 on Build GRCh38
rs996150102 Chr.2: 46480534-46480534 on Build GRCh38
rs60236492 Chr.2: 46480531-46480531 on Build GRCh38
rs895476191 Chr.2: 46480618-46480618 on Build GRCh38
rs114982496 Chr.2: 46480760-46480760 on Build GRCh38
rs1039738121 Chr.2: 46480529-46480529 on Build GRCh38
rs758108629 Chr.2: 46480424-46480424 on Build GRCh38
rs927060156 Chr.2: 46480593-46480593 on Build GRCh38
rs79819367 Chr.2: 46480673-46480673 on Build GRCh38
rs759003079 Chr.2: 46480662-46480663 on Build GRCh38
rs1014274547 Chr.2: 46480441-46480441 on Build GRCh38
rs780081579 Chr.2: 46480756-46480756 on Build GRCh38
rs770789607 Chr.2: 46480502-46480502 on Build GRCh38
rs973068979 Chr.2: 46480547-46480547 on Build GRCh38
rs752170015 Chr.2: 46480426-46480426 on Build GRCh38
rs927130760 Chr.2: 46480762-46480762 on Build GRCh38
rs368102006 Chr.2: 46480691-46480691 on Build GRCh38
rs954081467 Chr.2: 46480325-46480325 on Build GRCh38
rs376176475 Chr.2: 46480487-46480487 on Build GRCh38
rs61995901 Chr.2: 46480688-46480688 on Build GRCh38
rs975902751 Chr.2: 46480634-46480634 on Build GRCh38
rs778385438 Chr.2: 46480470-46480470 on Build GRCh38
rs371653273 Chr.2: 46480457-46480457 on Build GRCh38
rs771823668 Chr.2: 46480471-46480471 on Build GRCh38
rs1009993373 Chr.2: 46480543-46480543 on Build GRCh38

View Full Product Details