Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 53670605-53671115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB3
Entrez Gene ID
51130
Gene Name
ankyrin repeat and SOCS box containing 3
Gene Aliases
ASB-3
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53669980-53787009 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001164165.1

Gene Symbol
GPR75-ASB3
Entrez Gene ID
100302652
Gene Name
GPR75-ASB3 readthrough
Gene Aliases
-
UniGene
Hs.40763
Gene Chromosome Location
Chr.2: 53670293-53860033 on Build GRCh38
Transcripts
NM_001201965.1 NM_016115.4 NM_145863.2 NM_001164165.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6040657 c.1432C>T p.R478C Chr.2: 53670628-53670628 on Build GRCh38
6467303 c.1543G>A p.E515K Chr.2: 53670631-53670631 on Build GRCh38
4846360 c.1420C>G p.L474V Chr.2: 53670640-53670640 on Build GRCh38
721558 c.1420C>T p.L474L Chr.2: 53670640-53670640 on Build GRCh38
7155342 c.1382C>T p.S461F Chr.2: 53670678-53670678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761361891 Chr.2: 53670640-53670640 on Build GRCh38
rs1053936421 Chr.2: 53670868-53670868 on Build GRCh38
rs575890629 Chr.2: 53671037-53671037 on Build GRCh38
rs551838521 Chr.2: 53670789-53670789 on Build GRCh38
rs775146250 Chr.2: 53670852-53670852 on Build GRCh38
rs781631475 Chr.2: 53670705-53670705 on Build GRCh38
rs938245480 Chr.2: 53671046-53671046 on Build GRCh38
rs2287512 Chr.2: 53670895-53670895 on Build GRCh38
rs535516769 Chr.2: 53670837-53670837 on Build GRCh38
rs532245771 Chr.2: 53670763-53670763 on Build GRCh38
rs777975662 Chr.2: 53670695-53670695 on Build GRCh38
rs138524946 Chr.2: 53670954-53670954 on Build GRCh38
rs753750238 Chr.2: 53670661-53670661 on Build GRCh38
rs749890563 Chr.2: 53670651-53670651 on Build GRCh38
rs892863758 Chr.2: 53670634-53670634 on Build GRCh38
rs746327359 Chr.2: 53670706-53670706 on Build GRCh38
rs116175127 Chr.2: 53670905-53670905 on Build GRCh38
rs747096058 Chr.2: 53670720-53670721 on Build GRCh38
rs76786252 Chr.2: 53670766-53670768 on Build GRCh38
rs767347896 Chr.2: 53670648-53670648 on Build GRCh38
rs755080104 Chr.2: 53670723-53670726 on Build GRCh38
rs552233141 Chr.2: 53670964-53670964 on Build GRCh38
rs962612298 Chr.2: 53670863-53670863 on Build GRCh38
rs553825995 Chr.2: 53670631-53670631 on Build GRCh38
rs915405235 Chr.2: 53671074-53671074 on Build GRCh38
rs982388926 Chr.2: 53670857-53670857 on Build GRCh38
rs386646205 Chr.2: 53670985-53670986 on Build GRCh38
rs972061122 Chr.2: 53670798-53670798 on Build GRCh38
rs749670735 Chr.2: 53670731-53670731 on Build GRCh38
rs926914773 Chr.2: 53671039-53671039 on Build GRCh38
rs912230416 Chr.2: 53670942-53670942 on Build GRCh38
rs541940239 Chr.2: 53671007-53671007 on Build GRCh38
rs1011266082 Chr.2: 53670641-53670641 on Build GRCh38
rs549107776 Chr.2: 53670847-53670847 on Build GRCh38
rs1045178091 Chr.2: 53671042-53671042 on Build GRCh38
rs759333093 Chr.2: 53670811-53670811 on Build GRCh38
rs533693056 Chr.2: 53670762-53670762 on Build GRCh38
rs538662278 Chr.2: 53670759-53670759 on Build GRCh38
rs953239815 Chr.2: 53670760-53670760 on Build GRCh38
rs759668373 Chr.2: 53670649-53670649 on Build GRCh38
rs748353461 Chr.2: 53671091-53671091 on Build GRCh38
rs1030621087 Chr.2: 53670756-53670756 on Build GRCh38
rs567913502 Chr.2: 53670864-53670864 on Build GRCh38
rs754984344 Chr.2: 53670662-53670662 on Build GRCh38
rs754193764 Chr.2: 53670966-53670966 on Build GRCh38
rs963458420 Chr.2: 53670983-53670983 on Build GRCh38
rs898309020 Chr.2: 53671045-53671045 on Build GRCh38
rs561792870 Chr.2: 53671017-53671017 on Build GRCh38
rs942469539 Chr.2: 53670712-53670712 on Build GRCh38
rs764920811 Chr.2: 53670667-53670667 on Build GRCh38
rs1023881495 Chr.2: 53670856-53670856 on Build GRCh38
rs989678194 Chr.2: 53671054-53671054 on Build GRCh38
rs557208924 Chr.2: 53670927-53670927 on Build GRCh38
rs780588603 Chr.2: 53670720-53670720 on Build GRCh38
rs1025915469 Chr.2: 53671084-53671084 on Build GRCh38
rs997437265 Chr.2: 53670746-53670746 on Build GRCh38
rs371957173 Chr.2: 53671023-53671023 on Build GRCh38
rs566862079 Chr.2: 53670774-53670774 on Build GRCh38
rs544780860 Chr.2: 53671040-53671040 on Build GRCh38
rs191814299 Chr.2: 53670993-53670993 on Build GRCh38
rs34574323 Chr.2: 53670986-53670986 on Build GRCh38
rs963404567 Chr.2: 53670786-53670786 on Build GRCh38
rs1056440543 Chr.2: 53671018-53671018 on Build GRCh38
rs755412751 Chr.2: 53671006-53671006 on Build GRCh38
rs776407941 Chr.2: 53670761-53670761 on Build GRCh38
rs771796257 Chr.2: 53670740-53670740 on Build GRCh38
rs11899933 Chr.2: 53670985-53670985 on Build GRCh38
rs532574843 Chr.2: 53670767-53670767 on Build GRCh38
rs763637408 Chr.2: 53670926-53670926 on Build GRCh38
rs757348032 Chr.2: 53670700-53670700 on Build GRCh38
rs752594117 Chr.2: 53670674-53670674 on Build GRCh38
rs766370904 Chr.2: 53670660-53670660 on Build GRCh38
rs967318850 Chr.2: 53670735-53670735 on Build GRCh38
rs926834803 Chr.2: 53670860-53670860 on Build GRCh38
rs764813623 Chr.2: 53670851-53670851 on Build GRCh38
rs760278760 Chr.2: 53670656-53670656 on Build GRCh38
rs747033341 Chr.2: 53670697-53670697 on Build GRCh38
rs765438192 Chr.2: 53670782-53670782 on Build GRCh38
rs775960620 Chr.2: 53670635-53670635 on Build GRCh38
rs3179795 Chr.2: 53670676-53670676 on Build GRCh38
rs935588014 Chr.2: 53671013-53671013 on Build GRCh38
rs750743140 Chr.2: 53670700-53670701 on Build GRCh38
rs902848017 Chr.2: 53670915-53670915 on Build GRCh38
rs770379066 Chr.2: 53670710-53670710 on Build GRCh38
rs756637443 Chr.2: 53670957-53670957 on Build GRCh38
rs564659688 Chr.2: 53670729-53670729 on Build GRCh38
rs947108426 Chr.2: 53670893-53670893 on Build GRCh38
rs892916300 Chr.2: 53670874-53670874 on Build GRCh38
rs775716137 Chr.2: 53670711-53670711 on Build GRCh38
rs758613686 Chr.2: 53670680-53670680 on Build GRCh38
rs1007363878 Chr.2: 53670956-53670956 on Build GRCh38
rs758926249 Chr.2: 53670717-53670725 on Build GRCh38

View Full Product Details