Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55232707-55233209 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS27A
Entrez Gene ID
6233
Gene Name
ribosomal protein S27a
Gene Aliases
CEP80, HEL112, S27A, UBA80, UBC, UBCEP1, UBCEP80
UniGene
Hs.311640
Gene Chromosome Location
Chr.2: 55231903-55235853 on Build GRCh38
Transcripts
NM_001135592.2 NM_001177413.1 NM_002954.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3582315 c.29G>A p.G10E Chr.2: 55232853-55232853 on Build GRCh38
461010 c.12C>T p.F4F Chr.2: 55232836-55232836 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs754706035 Chr.2: 55232819-55232819 on Build GRCh38
rs1048438761 Chr.2: 55233071-55233071 on Build GRCh38
rs760271756 Chr.2: 55232923-55232923 on Build GRCh38
rs567089177 Chr.2: 55232876-55232877 on Build GRCh38
rs1017769617 Chr.2: 55233136-55233136 on Build GRCh38
rs770305845 Chr.2: 55232909-55232909 on Build GRCh38
rs955804183 Chr.2: 55233047-55233047 on Build GRCh38
rs374536442 Chr.2: 55232917-55232917 on Build GRCh38
rs753294900 Chr.2: 55233073-55233073 on Build GRCh38
rs1008874313 Chr.2: 55232756-55232756 on Build GRCh38
rs916888341 Chr.2: 55232857-55232857 on Build GRCh38
rs188817563 Chr.2: 55232970-55232970 on Build GRCh38
rs1024012782 Chr.2: 55233009-55233009 on Build GRCh38
rs767151322 Chr.2: 55232864-55232864 on Build GRCh38
rs772158174 Chr.2: 55232837-55232837 on Build GRCh38
rs773734998 Chr.2: 55232780-55232780 on Build GRCh38
rs776993141 Chr.2: 55232846-55232846 on Build GRCh38
rs925700138 Chr.2: 55232740-55232740 on Build GRCh38
rs996944477 Chr.2: 55232774-55232774 on Build GRCh38
rs978420065 Chr.2: 55233022-55233022 on Build GRCh38
rs767801252 Chr.2: 55232913-55232914 on Build GRCh38
rs370364740 Chr.2: 55232830-55232830 on Build GRCh38
rs796336298 Chr.2: 55232917-55232917 on Build GRCh38
rs948504639 Chr.2: 55232731-55232731 on Build GRCh38
rs528669947 Chr.2: 55233133-55233133 on Build GRCh38
rs1010056068 Chr.2: 55232768-55232768 on Build GRCh38
rs752925911 Chr.2: 55232883-55232883 on Build GRCh38
rs578030686 Chr.2: 55233130-55233130 on Build GRCh38
rs771537910 Chr.2: 55232845-55232845 on Build GRCh38
rs543843126 Chr.2: 55232787-55232787 on Build GRCh38
rs528688523 Chr.2: 55232886-55232886 on Build GRCh38
rs759998127 Chr.2: 55232847-55232847 on Build GRCh38
rs776603515 Chr.2: 55232730-55232730 on Build GRCh38
rs1027246388 Chr.2: 55233119-55233119 on Build GRCh38
rs986272993 Chr.2: 55233056-55233056 on Build GRCh38
rs370372426 Chr.2: 55233168-55233168 on Build GRCh38
rs750427691 Chr.2: 55232959-55232959 on Build GRCh38
rs371010709 Chr.2: 55232915-55232915 on Build GRCh38
rs150005663 Chr.2: 55233148-55233148 on Build GRCh38
rs968720876 Chr.2: 55233008-55233008 on Build GRCh38
rs376328333 Chr.2: 55233155-55233155 on Build GRCh38
rs1000149447 Chr.2: 55232796-55232796 on Build GRCh38
rs945378706 Chr.2: 55233076-55233076 on Build GRCh38
rs910133363 Chr.2: 55233164-55233164 on Build GRCh38
rs978387444 Chr.2: 55232885-55232885 on Build GRCh38
rs765303845 Chr.2: 55232771-55232771 on Build GRCh38
rs573055584 Chr.2: 55232971-55232971 on Build GRCh38
rs936874213 Chr.2: 55232743-55232743 on Build GRCh38
rs760088876 Chr.2: 55232870-55232870 on Build GRCh38
rs779145696 Chr.2: 55232823-55232823 on Build GRCh38
rs894405680 Chr.2: 55232977-55232977 on Build GRCh38
rs951773640 Chr.2: 55233052-55233052 on Build GRCh38
rs201568039 Chr.2: 55232907-55232907 on Build GRCh38
rs777768687 Chr.2: 55232839-55232839 on Build GRCh38
rs1043828014 Chr.2: 55232960-55232960 on Build GRCh38
rs531196007 Chr.2: 55233165-55233165 on Build GRCh38
rs905805240 Chr.2: 55233099-55233099 on Build GRCh38
rs1050630869 Chr.2: 55232746-55232747 on Build GRCh38
rs993140671 Chr.2: 55233112-55233112 on Build GRCh38
rs1055659481 Chr.2: 55233094-55233094 on Build GRCh38
rs751238040 Chr.2: 55232793-55232793 on Build GRCh38
rs552346316 Chr.2: 55232817-55232817 on Build GRCh38
rs539471097 Chr.2: 55233171-55233171 on Build GRCh38
rs766285620 Chr.2: 55233059-55233059 on Build GRCh38
rs35302335 Chr.2: 55232754-55232754 on Build GRCh38
rs760303882 Chr.2: 55232848-55232848 on Build GRCh38
rs777965161 Chr.2: 55232892-55232892 on Build GRCh38
rs559453539 Chr.2: 55233115-55233115 on Build GRCh38
rs1044631943 Chr.2: 55233177-55233177 on Build GRCh38
rs369376992 Chr.2: 55232802-55232802 on Build GRCh38
rs1042747350 Chr.2: 55232737-55232737 on Build GRCh38
rs17046786 Chr.2: 55232951-55232951 on Build GRCh38
rs932872537 Chr.2: 55233070-55233070 on Build GRCh38
rs752021725 Chr.2: 55232889-55232890 on Build GRCh38
rs1041066145 Chr.2: 55232759-55232759 on Build GRCh38
rs905672488 Chr.2: 55233186-55233186 on Build GRCh38
rs2028139 Chr.2: 55232881-55232881 on Build GRCh38
rs1034276146 Chr.2: 55232806-55232806 on Build GRCh38
rs568777276 Chr.2: 55232912-55232912 on Build GRCh38
rs563342368 Chr.2: 55232790-55232790 on Build GRCh38
rs150257026 Chr.2: 55232842-55232842 on Build GRCh38
rs183844234 Chr.2: 55232949-55232949 on Build GRCh38
rs988435664 Chr.2: 55233126-55233126 on Build GRCh38
rs373287175 Chr.2: 55233012-55233012 on Build GRCh38
rs575180825 Chr.2: 55232865-55232865 on Build GRCh38
rs1034161464 Chr.2: 55232975-55232975 on Build GRCh38
rs932896961 Chr.2: 55233154-55233154 on Build GRCh38
rs552269596 Chr.2: 55232866-55232866 on Build GRCh38
rs903958217 Chr.2: 55233085-55233088 on Build GRCh38
rs774178398 Chr.2: 55232854-55232854 on Build GRCh38
rs903906081 Chr.2: 55232961-55232961 on Build GRCh38
rs1040406569 Chr.2: 55232937-55232937 on Build GRCh38
rs779699784 Chr.2: 55233067-55233067 on Build GRCh38
rs766387520 Chr.2: 55232875-55232875 on Build GRCh38
rs754832103 Chr.2: 55232878-55232878 on Build GRCh38
rs971721217 Chr.2: 55233016-55233016 on Build GRCh38
rs570436521 Chr.2: 55233169-55233169 on Build GRCh38
rs1052536998 Chr.2: 55232746-55232746 on Build GRCh38
rs540210118 Chr.2: 55233132-55233132 on Build GRCh38
rs888413463 Chr.2: 55233072-55233072 on Build GRCh38
rs779719702 Chr.2: 55232813-55232813 on Build GRCh38
rs576847580 Chr.2: 55233098-55233098 on Build GRCh38
rs368746799 Chr.2: 55233110-55233110 on Build GRCh38
rs1014085064 Chr.2: 55232992-55232992 on Build GRCh38
rs528894828 Chr.2: 55233157-55233157 on Build GRCh38
rs761509538 Chr.2: 55232861-55232861 on Build GRCh38
rs766713370 Chr.2: 55232816-55232816 on Build GRCh38
rs747174865 Chr.2: 55232900-55232900 on Build GRCh38
rs548811915 Chr.2: 55233139-55233139 on Build GRCh38
rs921542914 Chr.2: 55233068-55233068 on Build GRCh38
rs761452955 Chr.2: 55232801-55232801 on Build GRCh38
rs759891368 Chr.2: 55232892-55232894 on Build GRCh38
rs772109083 Chr.2: 55232775-55232775 on Build GRCh38
rs930819743 Chr.2: 55232890-55232890 on Build GRCh38
rs771210460 Chr.2: 55232783-55232783 on Build GRCh38
rs376123816 Chr.2: 55232926-55232926 on Build GRCh38
rs563751402 Chr.2: 55232924-55232924 on Build GRCh38
rs1008901004 Chr.2: 55232752-55232752 on Build GRCh38
rs950138865 Chr.2: 55233175-55233175 on Build GRCh38
rs536640822 Chr.2: 55232918-55232918 on Build GRCh38
rs900079592 Chr.2: 55232950-55232950 on Build GRCh38
rs757264895 Chr.2: 55232901-55232901 on Build GRCh38
rs760520311 Chr.2: 55233057-55233057 on Build GRCh38
rs895910037 Chr.2: 55233172-55233172 on Build GRCh38
rs768484217 Chr.2: 55233149-55233149 on Build GRCh38
rs761117878 Chr.2: 55232781-55232781 on Build GRCh38
rs1020142153 Chr.2: 55232769-55232769 on Build GRCh38
rs1015433501 Chr.2: 55232979-55232979 on Build GRCh38
rs575446699 Chr.2: 55233066-55233066 on Build GRCh38
rs756023288 Chr.2: 55232811-55232811 on Build GRCh38
rs1004270138 Chr.2: 55232976-55232976 on Build GRCh38
rs192022169 Chr.2: 55233015-55233015 on Build GRCh38
rs895890867 Chr.2: 55233085-55233085 on Build GRCh38
rs558078164 Chr.2: 55232990-55232990 on Build GRCh38
rs1000042922 Chr.2: 55233105-55233105 on Build GRCh38
rs3211295 Chr.2: 55232808-55232808 on Build GRCh38
rs955669287 Chr.2: 55233111-55233111 on Build GRCh38
rs542043698 Chr.2: 55232988-55232988 on Build GRCh38
rs925596127 Chr.2: 55233030-55233030 on Build GRCh38
rs985828170 Chr.2: 55233129-55233129 on Build GRCh38
rs563309265 Chr.2: 55233084-55233084 on Build GRCh38
rs192298313 Chr.2: 55232791-55232791 on Build GRCh38
rs944244245 Chr.2: 55233064-55233064 on Build GRCh38
rs184094055 Chr.2: 55233074-55233074 on Build GRCh38
rs758545453 Chr.2: 55232887-55232887 on Build GRCh38
rs1014530971 Chr.2: 55233087-55233087 on Build GRCh38
rs753686670 Chr.2: 55232877-55232877 on Build GRCh38
rs762707912 Chr.2: 55232876-55232876 on Build GRCh38

View Full Product Details