Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 105816478-105816978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NCK2
Entrez Gene ID
8440
Gene Name
NCK adaptor protein 2
Gene Aliases
GRB4, NCKbeta
UniGene
Hs.529244
Gene Chromosome Location
Chr.2: 105744649-105894274 on Build GRCh38
Transcripts
NM_001004722.3 NM_003581.4 XM_006712797.3 XM_011511991.2 XM_017005103.1 XM_017005104.1 XM_017005105.1

SNP Detail

SNP ID Chromosome Location
rs555186519 Chr.2: 105816700-105816700 on Build GRCh38
rs377354223 Chr.2: 105816763-105816763 on Build GRCh38
rs752379954 Chr.2: 105816558-105816558 on Build GRCh38
rs35133141 Chr.2: 105816835-105816836 on Build GRCh38
rs188305622 Chr.2: 105816576-105816576 on Build GRCh38
rs993545013 Chr.2: 105816712-105816712 on Build GRCh38
rs946454523 Chr.2: 105816562-105816562 on Build GRCh38
rs776378213 Chr.2: 105816832-105816835 on Build GRCh38
rs905373832 Chr.2: 105816588-105816588 on Build GRCh38
rs980214937 Chr.2: 105816801-105816801 on Build GRCh38
rs373049830 Chr.2: 105816728-105816728 on Build GRCh38
rs566390929 Chr.2: 105816623-105816623 on Build GRCh38
rs115436337 Chr.2: 105816898-105816898 on Build GRCh38
rs563484511 Chr.2: 105816741-105816741 on Build GRCh38
rs574145699 Chr.2: 105816621-105816621 on Build GRCh38
rs577993437 Chr.2: 105816775-105816775 on Build GRCh38
rs946420417 Chr.2: 105816847-105816847 on Build GRCh38
rs1023948031 Chr.2: 105816768-105816768 on Build GRCh38
rs904139916 Chr.2: 105816683-105816683 on Build GRCh38
rs942584482 Chr.2: 105816513-105816513 on Build GRCh38
rs184059405 Chr.2: 105816748-105816748 on Build GRCh38
rs569184095 Chr.2: 105816563-105816563 on Build GRCh38
rs533836286 Chr.2: 105816648-105816648 on Build GRCh38
rs907734518 Chr.2: 105816904-105816904 on Build GRCh38
rs1029226856 Chr.2: 105816649-105816649 on Build GRCh38
rs529459627 Chr.2: 105816545-105816545 on Build GRCh38
rs755205313 Chr.2: 105816899-105816899 on Build GRCh38
rs563508447 Chr.2: 105816735-105816735 on Build GRCh38
rs113224718 Chr.2: 105816754-105816754 on Build GRCh38
rs922580215 Chr.2: 105816779-105816779 on Build GRCh38
rs930491854 Chr.2: 105816517-105816517 on Build GRCh38
rs751433196 Chr.2: 105816744-105816744 on Build GRCh38
rs913580575 Chr.2: 105816843-105816843 on Build GRCh38
rs975367208 Chr.2: 105816540-105816540 on Build GRCh38
rs192266729 Chr.2: 105816580-105816580 on Build GRCh38
rs556238587 Chr.2: 105816907-105816907 on Build GRCh38
rs987787943 Chr.2: 105816827-105816827 on Build GRCh38
rs1056742211 Chr.2: 105816697-105816697 on Build GRCh38
rs114807783 Chr.2: 105816541-105816541 on Build GRCh38
rs140218688 Chr.2: 105816713-105816713 on Build GRCh38
rs144076108 Chr.2: 105816781-105816781 on Build GRCh38
rs975716229 Chr.2: 105816770-105816770 on Build GRCh38
rs994158643 Chr.2: 105816926-105816926 on Build GRCh38
rs896773398 Chr.2: 105816920-105816920 on Build GRCh38
rs144783153 Chr.2: 105816551-105816551 on Build GRCh38
rs901857440 Chr.2: 105816579-105816579 on Build GRCh38
rs749325039 Chr.2: 105816879-105816879 on Build GRCh38
rs1002433729 Chr.2: 105816689-105816689 on Build GRCh38
rs927225196 Chr.2: 105816839-105816839 on Build GRCh38
rs759059155 Chr.2: 105816537-105816537 on Build GRCh38

View Full Product Details