Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 106122881-106123387 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UXS1
Entrez Gene ID
80146
Gene Name
UDP-glucuronate decarboxylase 1
Gene Aliases
SDR6E1, UGD
UniGene
Hs.469561
Gene Chromosome Location
Chr.2: 106093303-106194339 on Build GRCh38
Transcripts
NM_001253875.1 NM_001253876.1 NM_025076.4 NR_045607.1 XM_006712775.3 XM_011511902.2 XM_011511903.2 XM_011511904.2 XM_017005008.1 XM_017005009.1 XM_017005010.1 XM_017005011.1 XM_017005012.1 XM_017005013.1 XM_017005014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3743253 c.226G>T p.D76Y Chr.2: 106123020-106123020 on Build GRCh38
6897244 c.247G>C p.E83Q Chr.2: 106122999-106122999 on Build GRCh38
1270140 c.709G>A p.V237I Chr.2: 106123005-106123005 on Build GRCh38
3743251 c.694G>T p.D232Y Chr.2: 106123020-106123020 on Build GRCh38
714884 c.629A>C p.E210A Chr.2: 106123085-106123085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768019587 Chr.2: 106122997-106122997 on Build GRCh38
rs377359349 Chr.2: 106123150-106123150 on Build GRCh38
rs1007495187 Chr.2: 106122983-106122983 on Build GRCh38
rs766807110 Chr.2: 106122934-106122934 on Build GRCh38
rs892409213 Chr.2: 106123327-106123327 on Build GRCh38
rs753390976 Chr.2: 106123101-106123101 on Build GRCh38
rs770203522 Chr.2: 106123042-106123042 on Build GRCh38
rs948776316 Chr.2: 106123300-106123301 on Build GRCh38
rs559926382 Chr.2: 106123194-106123194 on Build GRCh38
rs747552442 Chr.2: 106123109-106123109 on Build GRCh38
rs748877295 Chr.2: 106122989-106122989 on Build GRCh38
rs574981459 Chr.2: 106123209-106123209 on Build GRCh38
rs369007917 Chr.2: 106122956-106122956 on Build GRCh38
rs35608734 Chr.2: 106123255-106123256 on Build GRCh38
rs750090315 Chr.2: 106122979-106122979 on Build GRCh38
rs746288779 Chr.2: 106123124-106123124 on Build GRCh38
rs764044038 Chr.2: 106123070-106123070 on Build GRCh38
rs954853026 Chr.2: 106123008-106123008 on Build GRCh38
rs964089463 Chr.2: 106123051-106123051 on Build GRCh38
rs6733841 Chr.2: 106122990-106122990 on Build GRCh38
rs372716743 Chr.2: 106122977-106122977 on Build GRCh38
rs1040325268 Chr.2: 106123000-106123000 on Build GRCh38
rs566608518 Chr.2: 106122956-106122956 on Build GRCh38
rs972723427 Chr.2: 106123198-106123198 on Build GRCh38
rs924460656 Chr.2: 106123200-106123200 on Build GRCh38
rs770254892 Chr.2: 106123128-106123128 on Build GRCh38
rs750674359 Chr.2: 106122945-106122945 on Build GRCh38
rs370644054 Chr.2: 106123040-106123040 on Build GRCh38
rs374105079 Chr.2: 106123300-106123300 on Build GRCh38
rs777012023 Chr.2: 106122935-106122935 on Build GRCh38
rs899144247 Chr.2: 106123331-106123331 on Build GRCh38
rs541546935 Chr.2: 106123180-106123180 on Build GRCh38
rs142723663 Chr.2: 106122911-106122911 on Build GRCh38
rs901849835 Chr.2: 106123310-106123310 on Build GRCh38
rs374696035 Chr.2: 106122936-106122936 on Build GRCh38
rs943424636 Chr.2: 106123287-106123287 on Build GRCh38
rs771434021 Chr.2: 106123036-106123036 on Build GRCh38
rs377181773 Chr.2: 106123020-106123020 on Build GRCh38
rs984999578 Chr.2: 106123223-106123223 on Build GRCh38
rs563892043 Chr.2: 106123319-106123319 on Build GRCh38
rs1022098744 Chr.2: 106123361-106123361 on Build GRCh38
rs150569868 Chr.2: 106123096-106123096 on Build GRCh38
rs558700579 Chr.2: 106123108-106123108 on Build GRCh38
rs548491148 Chr.2: 106123307-106123312 on Build GRCh38
rs200265406 Chr.2: 106123041-106123041 on Build GRCh38
rs148734335 Chr.2: 106122944-106122944 on Build GRCh38
rs930863370 Chr.2: 106122924-106122924 on Build GRCh38
rs755697432 Chr.2: 106122986-106122986 on Build GRCh38
rs778932682 Chr.2: 106123123-106123124 on Build GRCh38
rs969104925 Chr.2: 106123133-106123133 on Build GRCh38
rs538324277 Chr.2: 106123340-106123340 on Build GRCh38
rs552283439 Chr.2: 106123181-106123181 on Build GRCh38
rs1011260582 Chr.2: 106123352-106123352 on Build GRCh38
rs184270758 Chr.2: 106122918-106122918 on Build GRCh38
rs938480058 Chr.2: 106123202-106123202 on Build GRCh38
rs1022445768 Chr.2: 106123129-106123129 on Build GRCh38
rs767306152 Chr.2: 106123095-106123095 on Build GRCh38
rs934698068 Chr.2: 106123312-106123312 on Build GRCh38
rs771533130 Chr.2: 106123297-106123298 on Build GRCh38
rs11325302 Chr.2: 106123297-106123297 on Build GRCh38
rs781762155 Chr.2: 106123111-106123111 on Build GRCh38
rs76374170 Chr.2: 106123142-106123142 on Build GRCh38
rs930792632 Chr.2: 106123218-106123218 on Build GRCh38
rs1002974029 Chr.2: 106122906-106122906 on Build GRCh38
rs755881784 Chr.2: 106123098-106123098 on Build GRCh38
rs764569249 Chr.2: 106123300-106123304 on Build GRCh38
rs761681689 Chr.2: 106123089-106123089 on Build GRCh38
rs373615767 Chr.2: 106123078-106123078 on Build GRCh38
rs567925473 Chr.2: 106122952-106122952 on Build GRCh38
rs559569198 Chr.2: 106123300-106123301 on Build GRCh38
rs769671808 Chr.2: 106123298-106123298 on Build GRCh38
rs894559676 Chr.2: 106123283-106123283 on Build GRCh38
rs539683738 Chr.2: 106123103-106123103 on Build GRCh38
rs142328346 Chr.2: 106122978-106122978 on Build GRCh38
rs761138864 Chr.2: 106122925-106122925 on Build GRCh38
rs1040438932 Chr.2: 106123294-106123294 on Build GRCh38
rs146097866 Chr.2: 106122914-106122914 on Build GRCh38
rs910727482 Chr.2: 106123250-106123250 on Build GRCh38
rs200196579 Chr.2: 106123297-106123297 on Build GRCh38
rs2377455 Chr.2: 106123300-106123300 on Build GRCh38
rs763135254 Chr.2: 106123063-106123063 on Build GRCh38
rs919553331 Chr.2: 106123204-106123204 on Build GRCh38
rs747422760 Chr.2: 106123021-106123021 on Build GRCh38
rs963602643 Chr.2: 106122948-106122948 on Build GRCh38
rs751423534 Chr.2: 106122965-106122965 on Build GRCh38
rs775556547 Chr.2: 106123054-106123054 on Build GRCh38
rs762931906 Chr.2: 106122953-106122953 on Build GRCh38
rs1052771574 Chr.2: 106123318-106123318 on Build GRCh38
rs1026506558 Chr.2: 106123355-106123355 on Build GRCh38
rs771628622 Chr.2: 106123110-106123110 on Build GRCh38
rs754318715 Chr.2: 106123206-106123206 on Build GRCh38

View Full Product Details