Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 110650414-110650913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BUB1
Entrez Gene ID
699
Gene Name
BUB1 mitotic checkpoint serine/threonine kinase
Gene Aliases
BUB1A, BUB1L, hBUB1
UniGene
Hs.469649
Gene Chromosome Location
Chr.2: 110637698-110678107 on Build GRCh38
Transcripts
NM_001278616.1 NM_001278617.1 NM_004336.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4084168 c.2070G>T p.E690D Chr.2: 110650679-110650679 on Build GRCh38
4084167 c.2120T>G p.I707S Chr.2: 110650629-110650629 on Build GRCh38
6613646 c.2030G>A p.R677H Chr.2: 110650719-110650719 on Build GRCh38
2982994 c.2000C>T p.S667L Chr.2: 110650749-110650749 on Build GRCh38
2982992 c.2093G>A p.C698Y Chr.2: 110650656-110650656 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751876699 Chr.2: 110650596-110650596 on Build GRCh38
rs753199863 Chr.2: 110650620-110650620 on Build GRCh38
rs764154158 Chr.2: 110650656-110650656 on Build GRCh38
rs577463953 Chr.2: 110650591-110650591 on Build GRCh38
rs201651544 Chr.2: 110650748-110650748 on Build GRCh38
rs768572140 Chr.2: 110650660-110650660 on Build GRCh38
rs1801395 Chr.2: 110650637-110650637 on Build GRCh38
rs866894405 Chr.2: 110650775-110650779 on Build GRCh38
rs776719073 Chr.2: 110650567-110650567 on Build GRCh38
rs13412425 Chr.2: 110650887-110650887 on Build GRCh38
rs868578212 Chr.2: 110650497-110650497 on Build GRCh38
rs773048422 Chr.2: 110650700-110650701 on Build GRCh38
rs749852626 Chr.2: 110650773-110650773 on Build GRCh38
rs975621280 Chr.2: 110650749-110650749 on Build GRCh38
rs781106840 Chr.2: 110650819-110650819 on Build GRCh38
rs902866952 Chr.2: 110650871-110650871 on Build GRCh38
rs773128235 Chr.2: 110650696-110650696 on Build GRCh38
rs746740455 Chr.2: 110650655-110650655 on Build GRCh38
rs35764391 Chr.2: 110650759-110650760 on Build GRCh38
rs763057015 Chr.2: 110650593-110650593 on Build GRCh38
rs746652851 Chr.2: 110650812-110650812 on Build GRCh38
rs893308096 Chr.2: 110650512-110650512 on Build GRCh38
rs370231061 Chr.2: 110650618-110650618 on Build GRCh38
rs997357080 Chr.2: 110650668-110650668 on Build GRCh38
rs972249054 Chr.2: 110650453-110650453 on Build GRCh38
rs765227031 Chr.2: 110650681-110650682 on Build GRCh38
rs751489579 Chr.2: 110650666-110650666 on Build GRCh38
rs774561598 Chr.2: 110650723-110650723 on Build GRCh38
rs753111609 Chr.2: 110650758-110650758 on Build GRCh38
rs745421465 Chr.2: 110650636-110650636 on Build GRCh38
rs778553647 Chr.2: 110650546-110650546 on Build GRCh38
rs781683255 Chr.2: 110650526-110650526 on Build GRCh38
rs773144988 Chr.2: 110650630-110650630 on Build GRCh38
rs933742158 Chr.2: 110650768-110650768 on Build GRCh38
rs755350549 Chr.2: 110650597-110650597 on Build GRCh38
rs779778345 Chr.2: 110650652-110650652 on Build GRCh38
rs376481913 Chr.2: 110650635-110650635 on Build GRCh38
rs768662095 Chr.2: 110650566-110650566 on Build GRCh38
rs757947166 Chr.2: 110650642-110650642 on Build GRCh38
rs780263145 Chr.2: 110650501-110650501 on Build GRCh38
rs767946529 Chr.2: 110650606-110650606 on Build GRCh38
rs773393098 Chr.2: 110650592-110650592 on Build GRCh38
rs112442046 Chr.2: 110650738-110650738 on Build GRCh38
rs748081729 Chr.2: 110650667-110650667 on Build GRCh38
rs769813047 Chr.2: 110650680-110650680 on Build GRCh38
rs778262912 Chr.2: 110650630-110650630 on Build GRCh38
rs867863036 Chr.2: 110650720-110650720 on Build GRCh38
rs763120668 Chr.2: 110650708-110650708 on Build GRCh38
rs373297870 Chr.2: 110650542-110650542 on Build GRCh38
rs756703353 Chr.2: 110650627-110650627 on Build GRCh38
rs909749796 Chr.2: 110650690-110650690 on Build GRCh38
rs1045594666 Chr.2: 110650484-110650484 on Build GRCh38
rs766659533 Chr.2: 110650594-110650594 on Build GRCh38
rs748738807 Chr.2: 110650539-110650539 on Build GRCh38
rs761914583 Chr.2: 110650571-110650571 on Build GRCh38
rs949547482 Chr.2: 110650467-110650467 on Build GRCh38
rs368843962 Chr.2: 110650790-110650790 on Build GRCh38
rs760577088 Chr.2: 110650564-110650564 on Build GRCh38
rs892450555 Chr.2: 110650552-110650552 on Build GRCh38
rs766458085 Chr.2: 110650719-110650719 on Build GRCh38
rs905256346 Chr.2: 110650494-110650494 on Build GRCh38
rs764693050 Chr.2: 110650769-110650769 on Build GRCh38
rs771803937 Chr.2: 110650551-110650551 on Build GRCh38
rs751962820 Chr.2: 110650509-110650509 on Build GRCh38
rs12468060 Chr.2: 110650847-110650847 on Build GRCh38
rs754733068 Chr.2: 110650817-110650817 on Build GRCh38
rs1023062528 Chr.2: 110650555-110650555 on Build GRCh38
rs199900707 Chr.2: 110650764-110650764 on Build GRCh38
rs918093361 Chr.2: 110650458-110650458 on Build GRCh38
rs557116866 Chr.2: 110650562-110650562 on Build GRCh38
rs759821736 Chr.2: 110650744-110650744 on Build GRCh38
rs779781975 Chr.2: 110650799-110650799 on Build GRCh38
rs543760205 Chr.2: 110650549-110650549 on Build GRCh38
rs755442449 Chr.2: 110650522-110650522 on Build GRCh38

View Full Product Details