Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 111787835-111788341 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC1
Entrez Gene ID
64682
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
APC1, MCPR, TSG24
UniGene
Hs.436527
Gene Chromosome Location
Chr.2: 111766147-111884196 on Build GRCh38
Transcripts
NM_022662.3 XM_011511633.2 XM_011511634.2 XM_017004710.1 XM_017004711.1 XM_017004712.1 XM_017004713.1 XM_017004714.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6599512 c.4732A>C p.N1578H Chr.2: 111788301-111788301 on Build GRCh38
1721473 c.4739C>T p.S1580F Chr.2: 111788294-111788294 on Build GRCh38
4454369 c.4731A>C p.S1577S Chr.2: 111788302-111788302 on Build GRCh38
1005409 c.4736C>A p.S1579Y Chr.2: 111788297-111788297 on Build GRCh38
1005407 c.4747G>A p.A1583T Chr.2: 111788286-111788286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755360922 Chr.2: 111788314-111788314 on Build GRCh38
rs766667840 Chr.2: 111788306-111788306 on Build GRCh38
rs961094143 Chr.2: 111788073-111788073 on Build GRCh38
rs112272775 Chr.2: 111788282-111788282 on Build GRCh38
rs760675319 Chr.2: 111788209-111788209 on Build GRCh38
rs183437423 Chr.2: 111788108-111788108 on Build GRCh38
rs968145226 Chr.2: 111787929-111787929 on Build GRCh38
rs1010054631 Chr.2: 111787898-111787898 on Build GRCh38
rs142179264 Chr.2: 111788301-111788301 on Build GRCh38
rs750574891 Chr.2: 111788299-111788299 on Build GRCh38
rs765246504 Chr.2: 111788298-111788298 on Build GRCh38
rs1040659031 Chr.2: 111788188-111788188 on Build GRCh38
rs770597908 Chr.2: 111788184-111788184 on Build GRCh38
rs767216197 Chr.2: 111788201-111788201 on Build GRCh38
rs574279226 Chr.2: 111787924-111787924 on Build GRCh38
rs960609572 Chr.2: 111788005-111788005 on Build GRCh38
rs771814003 Chr.2: 111788270-111788270 on Build GRCh38
rs1033513174 Chr.2: 111787997-111787997 on Build GRCh38
rs753936913 Chr.2: 111788224-111788224 on Build GRCh38
rs1002001743 Chr.2: 111787947-111787947 on Build GRCh38
rs1050515729 Chr.2: 111788144-111788144 on Build GRCh38
rs764194687 Chr.2: 111788210-111788210 on Build GRCh38
rs1022404580 Chr.2: 111787928-111787928 on Build GRCh38
rs531695171 Chr.2: 111788242-111788242 on Build GRCh38
rs1357215 Chr.2: 111788121-111788121 on Build GRCh38
rs540466790 Chr.2: 111787895-111787895 on Build GRCh38
rs747444495 Chr.2: 111788262-111788262 on Build GRCh38
rs543277272 Chr.2: 111788092-111788092 on Build GRCh38
rs919648023 Chr.2: 111788126-111788126 on Build GRCh38
rs375661772 Chr.2: 111788239-111788239 on Build GRCh38
rs932410887 Chr.2: 111788141-111788141 on Build GRCh38
rs751901403 Chr.2: 111788312-111788312 on Build GRCh38
rs992121846 Chr.2: 111788010-111788010 on Build GRCh38
rs367637586 Chr.2: 111788263-111788263 on Build GRCh38
rs368949293 Chr.2: 111788287-111788287 on Build GRCh38
rs779157640 Chr.2: 111788235-111788235 on Build GRCh38
rs371127987 Chr.2: 111788234-111788234 on Build GRCh38
rs908189184 Chr.2: 111788057-111788057 on Build GRCh38
rs768092062 Chr.2: 111787890-111787893 on Build GRCh38
rs554327758 Chr.2: 111787912-111787912 on Build GRCh38
rs759332420 Chr.2: 111788197-111788197 on Build GRCh38
rs368093626 Chr.2: 111788289-111788289 on Build GRCh38
rs752618808 Chr.2: 111788203-111788203 on Build GRCh38
rs773895065 Chr.2: 111788186-111788186 on Build GRCh38
rs910759797 Chr.2: 111788160-111788160 on Build GRCh38
rs759713917 Chr.2: 111788234-111788235 on Build GRCh38
rs370250588 Chr.2: 111788267-111788267 on Build GRCh38
rs545998162 Chr.2: 111787859-111787859 on Build GRCh38
rs775420197 Chr.2: 111788274-111788274 on Build GRCh38
rs748744660 Chr.2: 111788264-111788264 on Build GRCh38
rs764084708 Chr.2: 111788252-111788253 on Build GRCh38
rs760585060 Chr.2: 111788286-111788286 on Build GRCh38

View Full Product Details