Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112055587-112056086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM87B
Entrez Gene ID
84910
Gene Name
transmembrane protein 87B
Gene Aliases
-
UniGene
Hs.656298
Gene Chromosome Location
Chr.2: 112055223-112119318 on Build GRCh38
Transcripts
NM_032824.2 XM_005263827.2 XM_006712807.2 XM_017005121.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1398815 c.125C>T p.A42V Chr.2: 112055716-112055716 on Build GRCh38
1613295 c.140G>C p.G47A Chr.2: 112055731-112055731 on Build GRCh38
4966520 c.31C>T p.L11F Chr.2: 112055622-112055622 on Build GRCh38
4132998 c.10G>C p.A4P Chr.2: 112055601-112055601 on Build GRCh38
6298409 c.25G>T p.A9S Chr.2: 112055616-112055616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777111962 Chr.2: 112055692-112055692 on Build GRCh38
rs772140248 Chr.2: 112055919-112055919 on Build GRCh38
rs748419381 Chr.2: 112055918-112055918 on Build GRCh38
rs1002700394 Chr.2: 112056049-112056049 on Build GRCh38
rs750745186 Chr.2: 112055883-112055883 on Build GRCh38
rs781557737 Chr.2: 112055865-112055865 on Build GRCh38
rs753623692 Chr.2: 112055753-112055753 on Build GRCh38
rs945817316 Chr.2: 112055617-112055617 on Build GRCh38
rs1051236154 Chr.2: 112055906-112055906 on Build GRCh38
rs763787246 Chr.2: 112055750-112055750 on Build GRCh38
rs536582197 Chr.2: 112055872-112055872 on Build GRCh38
rs768970776 Chr.2: 112055801-112055801 on Build GRCh38
rs769091458 Chr.2: 112055690-112055690 on Build GRCh38
rs1026928944 Chr.2: 112055658-112055658 on Build GRCh38
rs753673192 Chr.2: 112055794-112055802 on Build GRCh38
rs914248294 Chr.2: 112055848-112055848 on Build GRCh38
rs1025736568 Chr.2: 112056031-112056031 on Build GRCh38
rs556161925 Chr.2: 112055890-112055890 on Build GRCh38
rs934156133 Chr.2: 112055898-112055898 on Build GRCh38
rs565853159 Chr.2: 112055791-112055791 on Build GRCh38
rs62001036 Chr.2: 112055605-112055605 on Build GRCh38
rs765715924 Chr.2: 112055653-112055653 on Build GRCh38
rs953817542 Chr.2: 112055885-112055885 on Build GRCh38
rs1054570772 Chr.2: 112055611-112055611 on Build GRCh38
rs563738524 Chr.2: 112055684-112055684 on Build GRCh38
rs962807452 Chr.2: 112055713-112055713 on Build GRCh38
rs780369494 Chr.2: 112055770-112055770 on Build GRCh38
rs768777533 Chr.2: 112055916-112055916 on Build GRCh38
rs898502992 Chr.2: 112055978-112055978 on Build GRCh38
rs764861617 Chr.2: 112055798-112055798 on Build GRCh38
rs995372277 Chr.2: 112055877-112055877 on Build GRCh38
rs907325422 Chr.2: 112056035-112056035 on Build GRCh38
rs945794875 Chr.2: 112055866-112055866 on Build GRCh38
rs913016702 Chr.2: 112055957-112055957 on Build GRCh38
rs958646890 Chr.2: 112055836-112055836 on Build GRCh38
rs567347686 Chr.2: 112055844-112055844 on Build GRCh38
rs777578633 Chr.2: 112055667-112055667 on Build GRCh38
rs530469890 Chr.2: 112055649-112055649 on Build GRCh38
rs764554562 Chr.2: 112055640-112055640 on Build GRCh38
rs199855000 Chr.2: 112055679-112055679 on Build GRCh38
rs890022394 Chr.2: 112055922-112055922 on Build GRCh38
rs558545702 Chr.2: 112055962-112055962 on Build GRCh38
rs749428253 Chr.2: 112055912-112055912 on Build GRCh38
rs759069943 Chr.2: 112055645-112055645 on Build GRCh38
rs1019928152 Chr.2: 112055907-112055913 on Build GRCh38
rs973890294 Chr.2: 112055655-112055655 on Build GRCh38
rs368480880 Chr.2: 112055733-112055733 on Build GRCh38
rs767186599 Chr.2: 112055659-112055659 on Build GRCh38
rs758463203 Chr.2: 112055772-112055772 on Build GRCh38
rs538653317 Chr.2: 112055938-112055938 on Build GRCh38
rs747265797 Chr.2: 112055687-112055687 on Build GRCh38
rs988871620 Chr.2: 112055931-112055931 on Build GRCh38
rs865827826 Chr.2: 112056045-112056045 on Build GRCh38
rs745973100 Chr.2: 112055625-112055625 on Build GRCh38
rs760538028 Chr.2: 112055630-112055631 on Build GRCh38
rs779298988 Chr.2: 112055989-112055989 on Build GRCh38
rs938268335 Chr.2: 112055797-112055797 on Build GRCh38
rs757152832 Chr.2: 112055675-112055675 on Build GRCh38
rs943675961 Chr.2: 112055693-112055693 on Build GRCh38
rs113913839 Chr.2: 112056009-112056009 on Build GRCh38
rs145486099 Chr.2: 112055613-112055613 on Build GRCh38
rs762310705 Chr.2: 112055697-112055697 on Build GRCh38
rs977590162 Chr.2: 112055867-112055867 on Build GRCh38
rs1043866329 Chr.2: 112055742-112055742 on Build GRCh38
rs772333086 Chr.2: 112055631-112055631 on Build GRCh38
rs780517880 Chr.2: 112055911-112055911 on Build GRCh38
rs772054464 Chr.2: 112055683-112055683 on Build GRCh38
rs773174826 Chr.2: 112056007-112056007 on Build GRCh38
rs547194086 Chr.2: 112055833-112055833 on Build GRCh38
rs62000991 Chr.2: 112055717-112055717 on Build GRCh38
rs757097836 Chr.2: 112055762-112055762 on Build GRCh38
rs1025447202 Chr.2: 112055757-112055757 on Build GRCh38
rs576259042 Chr.2: 112055934-112055934 on Build GRCh38
rs572171784 Chr.2: 112056013-112056013 on Build GRCh38
rs756633113 Chr.2: 112055678-112055678 on Build GRCh38
rs942967692 Chr.2: 112055930-112055930 on Build GRCh38
rs978442919 Chr.2: 112055977-112055977 on Build GRCh38
rs945579051 Chr.2: 112055741-112055741 on Build GRCh38
rs35263835 Chr.2: 112055770-112055771 on Build GRCh38
rs532353568 Chr.2: 112055700-112055700 on Build GRCh38
rs759035949 Chr.2: 112055654-112055654 on Build GRCh38
rs896498676 Chr.2: 112055850-112055850 on Build GRCh38
rs965098237 Chr.2: 112055909-112055909 on Build GRCh38
rs764051676 Chr.2: 112055645-112055646 on Build GRCh38
rs748510904 Chr.2: 112055808-112055808 on Build GRCh38
rs904153517 Chr.2: 112055767-112055767 on Build GRCh38
rs1033677217 Chr.2: 112055619-112055619 on Build GRCh38
rs752310440 Chr.2: 112055722-112055722 on Build GRCh38
rs9308665 Chr.2: 112055814-112055814 on Build GRCh38
rs1017377309 Chr.2: 112055709-112055709 on Build GRCh38
rs750332831 Chr.2: 112055771-112055771 on Build GRCh38
rs752309698 Chr.2: 112055664-112055664 on Build GRCh38
rs781630150 Chr.2: 112055803-112055803 on Build GRCh38
rs775632127 Chr.2: 112055635-112055635 on Build GRCh38
rs766944323 Chr.2: 112055719-112055719 on Build GRCh38
rs753729050 Chr.2: 112055669-112055669 on Build GRCh38
rs756656513 Chr.2: 112055906-112055910 on Build GRCh38
rs752398184 Chr.2: 112055802-112055802 on Build GRCh38
rs541122910 Chr.2: 112056020-112056020 on Build GRCh38
rs778796284 Chr.2: 112055917-112055918 on Build GRCh38
rs774464705 Chr.2: 112055624-112055624 on Build GRCh38
rs981317138 Chr.2: 112055793-112055793 on Build GRCh38
rs768969224 Chr.2: 112055852-112055852 on Build GRCh38

View Full Product Details