Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 115161764-115162290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP10
Entrez Gene ID
57628
Gene Name
dipeptidyl peptidase like 10
Gene Aliases
DPL2, DPPY, DPRP-3, DPRP3
UniGene
Hs.580539
Gene Chromosome Location
Chr.2: 114442358-115845752 on Build GRCh38
Transcripts
NM_001004360.3 NM_001178034.1 NM_001178036.1 NM_001178037.1 NM_001321905.1 NM_001321906.1 NM_001321907.1 NM_001321908.1 NM_001321909.1 NM_001321910.1 NM_001321911.1 NM_001321912.1 NM_001321913.1 NM_001321914.1 NM_020868.4 XM_017004565.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4618082 c.17G>A p.S6N Chr.2: 115162250-115162250 on Build GRCh38
5798603 c.12G>T p.A4A Chr.2: 115161968-115161968 on Build GRCh38
7365391 c.35C>T p.P12L Chr.2: 115161991-115161991 on Build GRCh38
6033215 c.24G>A p.P8P Chr.2: 115161980-115161980 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762391336 Chr.2: 115162252-115162253 on Build GRCh38
rs980963144 Chr.2: 115161919-115161919 on Build GRCh38
rs888752081 Chr.2: 115162217-115162217 on Build GRCh38
rs745313676 Chr.2: 115162230-115162230 on Build GRCh38
rs750467930 Chr.2: 115162067-115162067 on Build GRCh38
rs139099509 Chr.2: 115162142-115162142 on Build GRCh38
rs533316032 Chr.2: 115161993-115161993 on Build GRCh38
rs1044573812 Chr.2: 115161903-115161918 on Build GRCh38
rs1035735137 Chr.2: 115162125-115162125 on Build GRCh38
rs866097134 Chr.2: 115161873-115161873 on Build GRCh38
rs142805323 Chr.2: 115162248-115162248 on Build GRCh38
rs1031311725 Chr.2: 115161786-115161786 on Build GRCh38
rs1016437124 Chr.2: 115162059-115162059 on Build GRCh38
rs969816532 Chr.2: 115162105-115162105 on Build GRCh38
rs551229604 Chr.2: 115161945-115161945 on Build GRCh38
rs538173052 Chr.2: 115162054-115162054 on Build GRCh38
rs533542037 Chr.2: 115162238-115162238 on Build GRCh38
rs937476781 Chr.2: 115161974-115161974 on Build GRCh38
rs542012551 Chr.2: 115161829-115161829 on Build GRCh38
rs958961459 Chr.2: 115162137-115162137 on Build GRCh38
rs150023622 Chr.2: 115162143-115162143 on Build GRCh38
rs992065382 Chr.2: 115161988-115161988 on Build GRCh38
rs7573359 Chr.2: 115162101-115162101 on Build GRCh38
rs746830959 Chr.2: 115162256-115162256 on Build GRCh38
rs953183852 Chr.2: 115161925-115161925 on Build GRCh38
rs879563044 Chr.2: 115162100-115162101 on Build GRCh38
rs148919798 Chr.2: 115162162-115162162 on Build GRCh38
rs866155718 Chr.2: 115162255-115162255 on Build GRCh38
rs755230362 Chr.2: 115162200-115162200 on Build GRCh38
rs981205206 Chr.2: 115162107-115162107 on Build GRCh38
rs771785335 Chr.2: 115162232-115162232 on Build GRCh38
rs941364673 Chr.2: 115162145-115162145 on Build GRCh38
rs569004887 Chr.2: 115162099-115162099 on Build GRCh38
rs997201073 Chr.2: 115161898-115161898 on Build GRCh38
rs751983600 Chr.2: 115162192-115162192 on Build GRCh38
rs571454423 Chr.2: 115162213-115162213 on Build GRCh38
rs531091791 Chr.2: 115161879-115161879 on Build GRCh38
rs1011871677 Chr.2: 115161839-115161839 on Build GRCh38
rs924846578 Chr.2: 115162176-115162176 on Build GRCh38
rs768494232 Chr.2: 115162262-115162262 on Build GRCh38
rs1026173189 Chr.2: 115162080-115162080 on Build GRCh38
rs760350563 Chr.2: 115161992-115161992 on Build GRCh38
rs573449601 Chr.2: 115161809-115161809 on Build GRCh38
rs1001094441 Chr.2: 115161835-115161835 on Build GRCh38
rs1011280604 Chr.2: 115162055-115162055 on Build GRCh38
rs1051459937 Chr.2: 115162040-115162040 on Build GRCh38
rs376052525 Chr.2: 115161999-115161999 on Build GRCh38
rs977615525 Chr.2: 115162170-115162170 on Build GRCh38
rs140962684 Chr.2: 115162147-115162147 on Build GRCh38
rs981434201 Chr.2: 115161961-115161961 on Build GRCh38
rs903460849 Chr.2: 115162019-115162019 on Build GRCh38
rs893371025 Chr.2: 115161817-115161817 on Build GRCh38
rs928699488 Chr.2: 115161963-115161963 on Build GRCh38
rs778617201 Chr.2: 115162225-115162225 on Build GRCh38
rs1042277770 Chr.2: 115161818-115161818 on Build GRCh38
rs754760125 Chr.2: 115161949-115161949 on Build GRCh38
rs867428293 Chr.2: 115162252-115162252 on Build GRCh38
rs766460352 Chr.2: 115162183-115162183 on Build GRCh38
rs1054558323 Chr.2: 115162069-115162069 on Build GRCh38
rs147298374 Chr.2: 115162144-115162144 on Build GRCh38
rs761720782 Chr.2: 115162001-115162001 on Build GRCh38
rs775380081 Chr.2: 115161951-115161951 on Build GRCh38
rs535986990 Chr.2: 115162072-115162072 on Build GRCh38
rs562344161 Chr.2: 115161848-115161848 on Build GRCh38
rs939592966 Chr.2: 115161935-115161935 on Build GRCh38
rs961170271 Chr.2: 115162126-115162126 on Build GRCh38
rs753324194 Chr.2: 115162221-115162221 on Build GRCh38
rs1041967857 Chr.2: 115162016-115162016 on Build GRCh38
rs1007796103 Chr.2: 115161940-115161940 on Build GRCh38
rs928743323 Chr.2: 115161795-115161795 on Build GRCh38
rs771603033 Chr.2: 115161950-115161950 on Build GRCh38
rs974074582 Chr.2: 115162171-115162171 on Build GRCh38
rs868636913 Chr.2: 115161834-115161834 on Build GRCh38
rs932959572 Chr.2: 115162022-115162022 on Build GRCh38
rs756659116 Chr.2: 115162224-115162224 on Build GRCh38
rs902393570 Chr.2: 115161828-115161828 on Build GRCh38
rs1032009159 Chr.2: 115162043-115162043 on Build GRCh38
rs144912580 Chr.2: 115162148-115162148 on Build GRCh38
rs1045765254 Chr.2: 115161783-115161783 on Build GRCh38
rs943924317 Chr.2: 115161813-115161813 on Build GRCh38
rs937488495 Chr.2: 115161797-115161797 on Build GRCh38
rs971378306 Chr.2: 115162139-115162139 on Build GRCh38
rs1030067573 Chr.2: 115161900-115161900 on Build GRCh38
rs898936564 Chr.2: 115162233-115162233 on Build GRCh38
rs902822146 Chr.2: 115161897-115161897 on Build GRCh38
rs200257545 Chr.2: 115162098-115162099 on Build GRCh38
rs67216957 Chr.2: 115162100-115162101 on Build GRCh38
rs566958673 Chr.2: 115162045-115162045 on Build GRCh38
rs1022937228 Chr.2: 115162135-115162135 on Build GRCh38
rs1055996860 Chr.2: 115161810-115161810 on Build GRCh38
rs548883749 Chr.2: 115162082-115162082 on Build GRCh38
rs765364507 Chr.2: 115162047-115162047 on Build GRCh38
rs150736667 Chr.2: 115162141-115162141 on Build GRCh38
rs147962378 Chr.2: 115162151-115162151 on Build GRCh38
rs983453836 Chr.2: 115161936-115161936 on Build GRCh38
rs7557011 Chr.2: 115162100-115162100 on Build GRCh38
rs34125495 Chr.2: 115162102-115162102 on Build GRCh38
rs564631625 Chr.2: 115161955-115161955 on Build GRCh38

View Full Product Details