Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 120218546-120219052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM185B
Entrez Gene ID
79134
Gene Name
transmembrane protein 185B
Gene Aliases
FAM11B
UniGene
Hs.376722
Gene Chromosome Location
Chr.2: 120217471-120223408 on Build GRCh38
Transcripts
NM_024121.2

SNP Detail

SNP ID Chromosome Location
rs1023768470 Chr.2: 120218657-120218657 on Build GRCh38
rs1001871285 Chr.2: 120218970-120218970 on Build GRCh38
rs975768620 Chr.2: 120218852-120218852 on Build GRCh38
rs537795262 Chr.2: 120218842-120218842 on Build GRCh38
rs988548559 Chr.2: 120218958-120218958 on Build GRCh38
rs541931833 Chr.2: 120218920-120218920 on Build GRCh38
rs568961441 Chr.2: 120218621-120218621 on Build GRCh38
rs879690937 Chr.2: 120218618-120218618 on Build GRCh38
rs914218545 Chr.2: 120218758-120218758 on Build GRCh38
rs777126129 Chr.2: 120218770-120218770 on Build GRCh38
rs933175492 Chr.2: 120218795-120218795 on Build GRCh38
rs1046885590 Chr.2: 120218743-120218743 on Build GRCh38
rs1017679694 Chr.2: 120219021-120219021 on Build GRCh38
rs1004946780 Chr.2: 120218826-120218826 on Build GRCh38
rs555326326 Chr.2: 120218732-120218732 on Build GRCh38
rs368523224 Chr.2: 120218930-120218930 on Build GRCh38
rs1002831290 Chr.2: 120218775-120218775 on Build GRCh38
rs116159892 Chr.2: 120218794-120218794 on Build GRCh38
rs919850905 Chr.2: 120218883-120218883 on Build GRCh38
rs565159197 Chr.2: 120218601-120218601 on Build GRCh38
rs149506800 Chr.2: 120218869-120218869 on Build GRCh38
rs988904513 Chr.2: 120218717-120218717 on Build GRCh38
rs1025254543 Chr.2: 120218570-120218570 on Build GRCh38
rs573598152 Chr.2: 120218824-120218824 on Build GRCh38
rs1033049603 Chr.2: 120218974-120218974 on Build GRCh38
rs186772314 Chr.2: 120218671-120218671 on Build GRCh38
rs949093821 Chr.2: 120218957-120218957 on Build GRCh38
rs78235591 Chr.2: 120218602-120218602 on Build GRCh38
rs552427695 Chr.2: 120218577-120218577 on Build GRCh38
rs947298150 Chr.2: 120218782-120218782 on Build GRCh38
rs933345251 Chr.2: 120218648-120218648 on Build GRCh38
rs988267987 Chr.2: 120218785-120218785 on Build GRCh38
rs1009702642 Chr.2: 120218977-120218977 on Build GRCh38
rs1012426027 Chr.2: 120218644-120218644 on Build GRCh38
rs942640194 Chr.2: 120218848-120218848 on Build GRCh38
rs970832994 Chr.2: 120218665-120218665 on Build GRCh38
rs980976804 Chr.2: 120218597-120218597 on Build GRCh38
rs74823521 Chr.2: 120218950-120218950 on Build GRCh38
rs577330204 Chr.2: 120218964-120218964 on Build GRCh38
rs900308086 Chr.2: 120218574-120218574 on Build GRCh38
rs987481831 Chr.2: 120219007-120219007 on Build GRCh38
rs970031920 Chr.2: 120218585-120218585 on Build GRCh38
rs776003608 Chr.2: 120218658-120218658 on Build GRCh38
rs190230779 Chr.2: 120218947-120218947 on Build GRCh38
rs970943277 Chr.2: 120218925-120218925 on Build GRCh38
rs904823902 Chr.2: 120218969-120218969 on Build GRCh38
rs574627903 Chr.2: 120218885-120218885 on Build GRCh38
rs548537649 Chr.2: 120218695-120218695 on Build GRCh38
rs144168577 Chr.2: 120218685-120218688 on Build GRCh38
rs942588217 Chr.2: 120218672-120218672 on Build GRCh38
rs199826046 Chr.2: 120218605-120218605 on Build GRCh38
rs928405769 Chr.2: 120218683-120218683 on Build GRCh38
rs1021053786 Chr.2: 120218984-120218984 on Build GRCh38
rs898287727 Chr.2: 120218685-120218685 on Build GRCh38
rs997608695 Chr.2: 120218600-120218600 on Build GRCh38
rs886839962 Chr.2: 120218755-120218755 on Build GRCh38
rs963618411 Chr.2: 120218638-120218638 on Build GRCh38
rs899139644 Chr.2: 120218863-120218863 on Build GRCh38
rs534251961 Chr.2: 120218774-120218774 on Build GRCh38
rs974398450 Chr.2: 120219026-120219026 on Build GRCh38
rs1023879457 Chr.2: 120218892-120218892 on Build GRCh38
rs894045474 Chr.2: 120218976-120218976 on Build GRCh38
rs1024618457 Chr.2: 120218761-120218761 on Build GRCh38
rs138016895 Chr.2: 120218784-120218784 on Build GRCh38
rs746210841 Chr.2: 120218739-120218739 on Build GRCh38
rs955801218 Chr.2: 120218687-120218687 on Build GRCh38
rs762745855 Chr.2: 120218919-120218919 on Build GRCh38
rs1038122076 Chr.2: 120218838-120218838 on Build GRCh38
rs954794401 Chr.2: 120218992-120218992 on Build GRCh38
rs886547954 Chr.2: 120218825-120218825 on Build GRCh38
rs968333403 Chr.2: 120219006-120219006 on Build GRCh38

View Full Product Details