Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 121216552-121217057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFCP2L1
Entrez Gene ID
29842
Gene Name
transcription factor CP2-like 1
Gene Aliases
CRTR1, LBP-9, LBP9
UniGene
Hs.156471
Gene Chromosome Location
Chr.2: 121216587-121285210 on Build GRCh38
Transcripts
NM_014553.2 XM_017003902.1 XM_017003903.1

SNP Detail

SNP ID Chromosome Location
rs896897166 Chr.2: 121216859-121216859 on Build GRCh38
rs1041148651 Chr.2: 121216858-121216858 on Build GRCh38
rs1048208998 Chr.2: 121216885-121216885 on Build GRCh38
rs758257459 Chr.2: 121216699-121216699 on Build GRCh38
rs557923706 Chr.2: 121216686-121216686 on Build GRCh38
rs879151902 Chr.2: 121216989-121216989 on Build GRCh38
rs978557710 Chr.2: 121217026-121217026 on Build GRCh38
rs967230878 Chr.2: 121216737-121216737 on Build GRCh38
rs994354197 Chr.2: 121216874-121216874 on Build GRCh38
rs916346618 Chr.2: 121216980-121216980 on Build GRCh38
rs894659550 Chr.2: 121216599-121216599 on Build GRCh38
rs747402982 Chr.2: 121216812-121216812 on Build GRCh38
rs959330698 Chr.2: 121216670-121216670 on Build GRCh38
rs560842624 Chr.2: 121216959-121216959 on Build GRCh38
rs961284327 Chr.2: 121216829-121216829 on Build GRCh38
rs112229464 Chr.2: 121216651-121216651 on Build GRCh38
rs1031687291 Chr.2: 121216662-121216662 on Build GRCh38
rs948879155 Chr.2: 121216985-121216985 on Build GRCh38
rs1028142363 Chr.2: 121216736-121216736 on Build GRCh38
rs978270114 Chr.2: 121216776-121216776 on Build GRCh38
rs994012848 Chr.2: 121216597-121216597 on Build GRCh38
rs998819725 Chr.2: 121216658-121216658 on Build GRCh38
rs1025424843 Chr.2: 121216993-121216993 on Build GRCh38
rs952620260 Chr.2: 121216644-121216644 on Build GRCh38
rs556317502 Chr.2: 121217030-121217030 on Build GRCh38
rs927673662 Chr.2: 121216886-121216886 on Build GRCh38
rs746485680 Chr.2: 121216774-121216775 on Build GRCh38
rs966800502 Chr.2: 121217009-121217009 on Build GRCh38
rs116440946 Chr.2: 121216796-121216796 on Build GRCh38
rs1001330528 Chr.2: 121216912-121216912 on Build GRCh38
rs1013577441 Chr.2: 121216992-121216992 on Build GRCh38
rs746769567 Chr.2: 121216795-121216795 on Build GRCh38
rs769122137 Chr.2: 121216920-121216920 on Build GRCh38
rs888258390 Chr.2: 121216896-121216896 on Build GRCh38
rs749964607 Chr.2: 121216694-121216694 on Build GRCh38
rs1034249148 Chr.2: 121216661-121216661 on Build GRCh38
rs901253255 Chr.2: 121216669-121216669 on Build GRCh38
rs959985431 Chr.2: 121216973-121216973 on Build GRCh38
rs376129360 Chr.2: 121216928-121216928 on Build GRCh38
rs1043672532 Chr.2: 121216629-121216629 on Build GRCh38
rs918003783 Chr.2: 121216713-121216713 on Build GRCh38
rs943646903 Chr.2: 121216856-121216856 on Build GRCh38
rs780207226 Chr.2: 121216794-121216794 on Build GRCh38
rs911485573 Chr.2: 121216844-121216844 on Build GRCh38
rs1027270457 Chr.2: 121216621-121216621 on Build GRCh38
rs186160224 Chr.2: 121216822-121216822 on Build GRCh38

View Full Product Details