Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 18913327-18913836 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRODH
Entrez Gene ID
5625
Gene Name
proline dehydrogenase 1
Gene Aliases
HSPOX2, PIG6, POX, PRODH1, PRODH2, TP53I6
UniGene
Hs.517352
Gene Chromosome Location
Chr.22: 18912774-18936553 on Build GRCh38
Transcripts
NM_001195226.1 NM_016335.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6756404 c.1615+1G>T p.? Chr.22: 18913437-18913437 on Build GRCh38
1031958 c.1643A>G p.Y548C Chr.22: 18913335-18913335 on Build GRCh38
4004837 c.1562G>A p.R521Q Chr.22: 18913491-18913491 on Build GRCh38
6756403 c.1644C>T p.Y548Y Chr.22: 18913334-18913334 on Build GRCh38
5499432 c.1615+2T>C p.? Chr.22: 18913436-18913436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753114295 Chr.22: 18913391-18913391 on Build GRCh38
rs771965438 Chr.22: 18913457-18913457 on Build GRCh38
rs756133347 Chr.22: 18913537-18913537 on Build GRCh38
rs193087012 Chr.22: 18913552-18913552 on Build GRCh38
rs867893870 Chr.22: 18913475-18913475 on Build GRCh38
rs142972542 Chr.22: 18913493-18913493 on Build GRCh38
rs370957234 Chr.22: 18913394-18913394 on Build GRCh38
rs199660947 Chr.22: 18913361-18913361 on Build GRCh38
rs548818330 Chr.22: 18913807-18913807 on Build GRCh38
rs772033871 Chr.22: 18913415-18913415 on Build GRCh38
rs909393104 Chr.22: 18913643-18913643 on Build GRCh38
rs757806392 Chr.22: 18913515-18913515 on Build GRCh38
rs868561195 Chr.22: 18913615-18913615 on Build GRCh38
rs372942723 Chr.22: 18913672-18913672 on Build GRCh38
rs184880228 Chr.22: 18913719-18913719 on Build GRCh38
rs752177336 Chr.22: 18913408-18913408 on Build GRCh38
rs762508776 Chr.22: 18913497-18913497 on Build GRCh38
rs558011936 Chr.22: 18913570-18913570 on Build GRCh38
rs374270089 Chr.22: 18913443-18913443 on Build GRCh38
rs373157994 Chr.22: 18913442-18913442 on Build GRCh38
rs767975212 Chr.22: 18913520-18913520 on Build GRCh38
rs533071846 Chr.22: 18913760-18913760 on Build GRCh38
rs377148163 Chr.22: 18913750-18913750 on Build GRCh38
rs989175428 Chr.22: 18913508-18913508 on Build GRCh38
rs749561328 Chr.22: 18913547-18913547 on Build GRCh38
rs751849255 Chr.22: 18913504-18913504 on Build GRCh38
rs949712795 Chr.22: 18913710-18913710 on Build GRCh38
rs868344114 Chr.22: 18913527-18913527 on Build GRCh38
rs762463680 Chr.22: 18913393-18913394 on Build GRCh38
rs368092146 Chr.22: 18913388-18913388 on Build GRCh38
rs896852717 Chr.22: 18913414-18913414 on Build GRCh38
rs573141558 Chr.22: 18913732-18913732 on Build GRCh38
rs450046 Chr.22: 18913491-18913491 on Build GRCh38
rs144323192 Chr.22: 18913495-18913495 on Build GRCh38
rs757381009 Chr.22: 18913573-18913573 on Build GRCh38
rs868634585 Chr.22: 18913362-18913362 on Build GRCh38
rs755128435 Chr.22: 18913576-18913576 on Build GRCh38
rs753373264 Chr.22: 18913378-18913378 on Build GRCh38
rs369753900 Chr.22: 18913389-18913389 on Build GRCh38
rs751254651 Chr.22: 18913404-18913407 on Build GRCh38
rs563306272 Chr.22: 18913773-18913773 on Build GRCh38
rs745902339 Chr.22: 18913437-18913437 on Build GRCh38
rs780097778 Chr.22: 18913546-18913546 on Build GRCh38
rs142936184 Chr.22: 18913501-18913501 on Build GRCh38
rs756739806 Chr.22: 18913551-18913552 on Build GRCh38
rs765949326 Chr.22: 18913373-18913373 on Build GRCh38
rs16983347 Chr.22: 18913355-18913355 on Build GRCh38
rs984941246 Chr.22: 18913616-18913616 on Build GRCh38
rs557337663 Chr.22: 18913606-18913606 on Build GRCh38
rs921989572 Chr.22: 18913590-18913590 on Build GRCh38
rs758729476 Chr.22: 18913402-18913402 on Build GRCh38
rs932064595 Chr.22: 18913592-18913592 on Build GRCh38
rs372187772 Chr.22: 18913477-18913477 on Build GRCh38
rs752549161 Chr.22: 18913368-18913369 on Build GRCh38
rs543018473 Chr.22: 18913766-18913766 on Build GRCh38
rs540708898 Chr.22: 18913743-18913743 on Build GRCh38
rs755465802 Chr.22: 18913789-18913790 on Build GRCh38
rs537021425 Chr.22: 18913578-18913578 on Build GRCh38
rs750508839 Chr.22: 18913535-18913535 on Build GRCh38
rs1032806279 Chr.22: 18913762-18913763 on Build GRCh38
rs381848 Chr.22: 18913777-18913777 on Build GRCh38
rs905608062 Chr.22: 18913733-18913733 on Build GRCh38
rs113306778 Chr.22: 18913665-18913665 on Build GRCh38
rs368452830 Chr.22: 18913470-18913470 on Build GRCh38
rs769982914 Chr.22: 18913354-18913354 on Build GRCh38
rs747912714 Chr.22: 18913454-18913454 on Build GRCh38
rs760522404 Chr.22: 18913371-18913371 on Build GRCh38
rs148584334 Chr.22: 18913433-18913433 on Build GRCh38
rs62232347 Chr.22: 18913809-18913809 on Build GRCh38
rs953423447 Chr.22: 18913810-18913811 on Build GRCh38
rs193919334 Chr.22: 18913492-18913492 on Build GRCh38
rs569823639 Chr.22: 18913431-18913431 on Build GRCh38
rs773147149 Chr.22: 18913366-18913366 on Build GRCh38
rs896742209 Chr.22: 18913680-18913680 on Build GRCh38
rs966009289 Chr.22: 18913806-18913806 on Build GRCh38
rs777988236 Chr.22: 18913405-18913405 on Build GRCh38
rs385440 Chr.22: 18913577-18913577 on Build GRCh38
rs763768502 Chr.22: 18913400-18913414 on Build GRCh38

View Full Product Details