Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20967044-20967555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIFM3
Entrez Gene ID
150209
Gene Name
apoptosis inducing factor, mitochondria associated 3
Gene Aliases
AIFL
UniGene
Hs.723246
Gene Chromosome Location
Chr.22: 20965130-20981360 on Build GRCh38
Transcripts
NM_001018060.2 NM_001146288.1 NM_144704.2 NR_027464.1

SNP Detail

SNP ID Chromosome Location
rs926035467 Chr.22: 20967251-20967251 on Build GRCh38
rs536404825 Chr.22: 20967451-20967451 on Build GRCh38
rs1033093789 Chr.22: 20967447-20967447 on Build GRCh38
rs1019549340 Chr.22: 20967108-20967108 on Build GRCh38
rs563791260 Chr.22: 20967072-20967072 on Build GRCh38
rs112472055 Chr.22: 20967195-20967195 on Build GRCh38
rs1009922632 Chr.22: 20967095-20967095 on Build GRCh38
rs111843050 Chr.22: 20967433-20967433 on Build GRCh38
rs978838156 Chr.22: 20967246-20967246 on Build GRCh38
rs998417139 Chr.22: 20967419-20967419 on Build GRCh38
rs949349044 Chr.22: 20967202-20967202 on Build GRCh38
rs369454650 Chr.22: 20967230-20967230 on Build GRCh38
rs552882362 Chr.22: 20967481-20967481 on Build GRCh38
rs909393203 Chr.22: 20967177-20967177 on Build GRCh38
rs967555404 Chr.22: 20967214-20967214 on Build GRCh38
rs550283595 Chr.22: 20967071-20967071 on Build GRCh38
rs1046195584 Chr.22: 20967314-20967314 on Build GRCh38
rs6005063 Chr.22: 20967533-20967533 on Build GRCh38
rs901075036 Chr.22: 20967532-20967532 on Build GRCh38
rs776214523 Chr.22: 20967111-20967131 on Build GRCh38
rs1018527540 Chr.22: 20967199-20967199 on Build GRCh38
rs916489784 Chr.22: 20967288-20967288 on Build GRCh38
rs73164810 Chr.22: 20967502-20967502 on Build GRCh38
rs749110277 Chr.22: 20967504-20967504 on Build GRCh38
rs905289566 Chr.22: 20967075-20967075 on Build GRCh38
rs141871613 Chr.22: 20967203-20967203 on Build GRCh38
rs967503633 Chr.22: 20967435-20967435 on Build GRCh38
rs1018389077 Chr.22: 20967387-20967387 on Build GRCh38
rs145326729 Chr.22: 20967382-20967382 on Build GRCh38
rs937316451 Chr.22: 20967262-20967262 on Build GRCh38
rs918505812 Chr.22: 20967506-20967506 on Build GRCh38
rs984963533 Chr.22: 20967160-20967160 on Build GRCh38
rs1053609190 Chr.22: 20967279-20967279 on Build GRCh38
rs183159029 Chr.22: 20967291-20967291 on Build GRCh38
rs751131503 Chr.22: 20967159-20967159 on Build GRCh38
rs535001251 Chr.22: 20967242-20967242 on Build GRCh38
rs929824333 Chr.22: 20967507-20967507 on Build GRCh38
rs965654761 Chr.22: 20967200-20967200 on Build GRCh38
rs1056237706 Chr.22: 20967263-20967263 on Build GRCh38
rs956638577 Chr.22: 20967083-20967083 on Build GRCh38
rs952379516 Chr.22: 20967178-20967178 on Build GRCh38
rs991492110 Chr.22: 20967484-20967484 on Build GRCh38
rs921010456 Chr.22: 20967147-20967148 on Build GRCh38
rs17551486 Chr.22: 20967344-20967344 on Build GRCh38
rs1027164366 Chr.22: 20967172-20967172 on Build GRCh38
rs942470002 Chr.22: 20967514-20967514 on Build GRCh38
rs745649665 Chr.22: 20967313-20967313 on Build GRCh38
rs368555070 Chr.22: 20967505-20967505 on Build GRCh38
rs1019908962 Chr.22: 20967297-20967297 on Build GRCh38
rs80336761 Chr.22: 20967441-20967441 on Build GRCh38
rs759085051 Chr.22: 20967189-20967189 on Build GRCh38
rs888233390 Chr.22: 20967366-20967366 on Build GRCh38
rs959077158 Chr.22: 20967452-20967452 on Build GRCh38
rs373183163 Chr.22: 20967302-20967302 on Build GRCh38
rs1048104019 Chr.22: 20967358-20967358 on Build GRCh38
rs895057669 Chr.22: 20967081-20967081 on Build GRCh38
rs901408625 Chr.22: 20967405-20967405 on Build GRCh38
rs1048741390 Chr.22: 20967508-20967508 on Build GRCh38
rs12170987 Chr.22: 20967076-20967076 on Build GRCh38
rs896965025 Chr.22: 20967340-20967340 on Build GRCh38
rs913428162 Chr.22: 20967213-20967213 on Build GRCh38
rs147667710 Chr.22: 20967512-20967512 on Build GRCh38
rs796831060 Chr.22: 20967321-20967321 on Build GRCh38
rs1013370237 Chr.22: 20967145-20967145 on Build GRCh38
rs868664790 Chr.22: 20967146-20967146 on Build GRCh38
rs940888025 Chr.22: 20967187-20967187 on Build GRCh38
rs896896090 Chr.22: 20967148-20967148 on Build GRCh38

View Full Product Details