Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 21670902-21671327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPIL2
Entrez Gene ID
23759
Gene Name
peptidylprolyl isomerase like 2
Gene Aliases
CYC4, CYP60, Cyp-60, UBOX7, hCyP-60
UniGene
Hs.438587
Gene Chromosome Location
Chr.22: 21665984-21698276 on Build GRCh38
Transcripts
NM_001317996.1 NM_014337.3 NM_148175.2 NM_148176.2 XM_005261448.3 XM_011530041.2 XM_011530042.2 XM_011530043.2 XM_011530044.2 XM_011530046.2 XM_011530047.2 XM_011530049.2 XM_011530050.2 XM_011530051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3552420 c.130C>G p.L44V Chr.22: 21670998-21670998 on Build GRCh38
4102893 c.169G>A p.D57N Chr.22: 21671037-21671037 on Build GRCh38
7385170 c.189A>C p.L63F Chr.22: 21671057-21671057 on Build GRCh38
7385169 c.189A>C p.L63F Chr.22: 21671057-21671057 on Build GRCh38
4102894 c.169G>A p.D57N Chr.22: 21671037-21671037 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777862182 Chr.22: 21670983-21670983 on Build GRCh38
rs764609156 Chr.22: 21670953-21670953 on Build GRCh38
rs774909103 Chr.22: 21671037-21671037 on Build GRCh38
rs143867306 Chr.22: 21670956-21670956 on Build GRCh38
rs767867119 Chr.22: 21670961-21670961 on Build GRCh38
rs745582053 Chr.22: 21671020-21671020 on Build GRCh38
rs770915233 Chr.22: 21671017-21671017 on Build GRCh38
rs1024255124 Chr.22: 21671135-21671135 on Build GRCh38
rs108815 Chr.22: 21671187-21671187 on Build GRCh38
rs192875862 Chr.22: 21671045-21671045 on Build GRCh38
rs1009725789 Chr.22: 21671012-21671012 on Build GRCh38
rs370871572 Chr.22: 21670996-21670996 on Build GRCh38
rs35747572 Chr.22: 21671269-21671270 on Build GRCh38
rs371051805 Chr.22: 21671099-21671099 on Build GRCh38
rs767976496 Chr.22: 21671046-21671046 on Build GRCh38
rs762174812 Chr.22: 21670960-21670960 on Build GRCh38
rs750719187 Chr.22: 21670962-21670962 on Build GRCh38
rs946796287 Chr.22: 21671285-21671285 on Build GRCh38
rs758539436 Chr.22: 21671289-21671289 on Build GRCh38
rs910277826 Chr.22: 21671262-21671262 on Build GRCh38
rs1647705 Chr.22: 21671173-21671173 on Build GRCh38
rs146708734 Chr.22: 21671277-21671277 on Build GRCh38
rs756270047 Chr.22: 21670968-21670968 on Build GRCh38
rs766131675 Chr.22: 21671136-21671136 on Build GRCh38
rs971294774 Chr.22: 21671160-21671160 on Build GRCh38
rs760477838 Chr.22: 21671133-21671133 on Build GRCh38
rs118046190 Chr.22: 21671299-21671299 on Build GRCh38
rs143244242 Chr.22: 21671265-21671265 on Build GRCh38
rs145883797 Chr.22: 21671021-21671021 on Build GRCh38
rs1025300924 Chr.22: 21671132-21671132 on Build GRCh38
rs755057004 Chr.22: 21670980-21670980 on Build GRCh38
rs767228694 Chr.22: 21670936-21670936 on Build GRCh38
rs750725751 Chr.22: 21671052-21671052 on Build GRCh38
rs370659593 Chr.22: 21670948-21670948 on Build GRCh38
rs984959909 Chr.22: 21671235-21671235 on Build GRCh38
rs772842036 Chr.22: 21670954-21670954 on Build GRCh38
rs149000654 Chr.22: 21671036-21671036 on Build GRCh38
rs774571500 Chr.22: 21670997-21671000 on Build GRCh38
rs765052572 Chr.22: 21671237-21671237 on Build GRCh38
rs112807589 Chr.22: 21671098-21671098 on Build GRCh38
rs761058691 Chr.22: 21671092-21671092 on Build GRCh38
rs780220747 Chr.22: 21670971-21670971 on Build GRCh38
rs746875774 Chr.22: 21671015-21671015 on Build GRCh38
rs202229100 Chr.22: 21671002-21671002 on Build GRCh38
rs762024632 Chr.22: 21671086-21671086 on Build GRCh38
rs374438374 Chr.22: 21671076-21671076 on Build GRCh38
rs377318280 Chr.22: 21670985-21670985 on Build GRCh38
rs762465013 Chr.22: 21671027-21671027 on Build GRCh38
rs919084948 Chr.22: 21671178-21671178 on Build GRCh38
rs889728128 Chr.22: 21671022-21671022 on Build GRCh38
rs752886601 Chr.22: 21671284-21671284 on Build GRCh38
rs993951707 Chr.22: 21671106-21671106 on Build GRCh38
rs902144231 Chr.22: 21671295-21671295 on Build GRCh38
rs374080411 Chr.22: 21670976-21670976 on Build GRCh38

View Full Product Details