Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 22485181-22485717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF280B
Entrez Gene ID
140883
Gene Name
zinc finger protein 280B
Gene Aliases
5'OY11.1, D87009.C22.3, SUHW2, ZNF279, ZNF632
UniGene
Hs.43834
Gene Chromosome Location
Chr.22: 22484421-22509187 on Build GRCh38
Transcripts
NM_080764.3 NR_130642.1 NR_130643.1 XM_011529893.2 XM_011529895.2 XM_011529896.2

SNP Detail

SNP ID Chromosome Location
rs540748568 Chr.22: 22485467-22485467 on Build GRCh38
rs1048151430 Chr.22: 22485265-22485265 on Build GRCh38
rs408785 Chr.22: 22485213-22485213 on Build GRCh38
rs953560063 Chr.22: 22485660-22485660 on Build GRCh38
rs1036758826 Chr.22: 22485604-22485604 on Build GRCh38
rs1036324461 Chr.22: 22485523-22485523 on Build GRCh38
rs1043617974 Chr.22: 22485424-22485424 on Build GRCh38
rs186304504 Chr.22: 22485338-22485338 on Build GRCh38
rs771693691 Chr.22: 22485410-22485410 on Build GRCh38
rs532850699 Chr.22: 22485661-22485661 on Build GRCh38
rs1000469568 Chr.22: 22485377-22485377 on Build GRCh38
rs75324277 Chr.22: 22485667-22485667 on Build GRCh38
rs761744835 Chr.22: 22485299-22485299 on Build GRCh38
rs888227017 Chr.22: 22485278-22485278 on Build GRCh38
rs191279543 Chr.22: 22485347-22485347 on Build GRCh38
rs922144556 Chr.22: 22485474-22485474 on Build GRCh38
rs930902166 Chr.22: 22485476-22485476 on Build GRCh38
rs898118045 Chr.22: 22485570-22485570 on Build GRCh38
rs113092504 Chr.22: 22485375-22485375 on Build GRCh38
rs750676691 Chr.22: 22485383-22485383 on Build GRCh38
rs895485442 Chr.22: 22485610-22485610 on Build GRCh38
rs895454383 Chr.22: 22485318-22485318 on Build GRCh38
rs767761377 Chr.22: 22485356-22485356 on Build GRCh38
rs543509735 Chr.22: 22485402-22485402 on Build GRCh38
rs965995768 Chr.22: 22485431-22485431 on Build GRCh38
rs1015306148 Chr.22: 22485562-22485562 on Build GRCh38
rs1003013558 Chr.22: 22485320-22485320 on Build GRCh38
rs1027806804 Chr.22: 22485608-22485608 on Build GRCh38
rs1039234245 Chr.22: 22485230-22485230 on Build GRCh38
rs576719498 Chr.22: 22485325-22485325 on Build GRCh38
rs1049733631 Chr.22: 22485533-22485533 on Build GRCh38
rs922143188 Chr.22: 22485244-22485244 on Build GRCh38
rs1014197112 Chr.22: 22485359-22485359 on Build GRCh38
rs939765289 Chr.22: 22485567-22485567 on Build GRCh38
rs930768018 Chr.22: 22485249-22485249 on Build GRCh38
rs988845062 Chr.22: 22485418-22485418 on Build GRCh38
rs895867927 Chr.22: 22485547-22485547 on Build GRCh38
rs536536483 Chr.22: 22485406-22485408 on Build GRCh38
rs117942036 Chr.22: 22485430-22485430 on Build GRCh38
rs949664651 Chr.22: 22485649-22485649 on Build GRCh38
rs913825456 Chr.22: 22485341-22485341 on Build GRCh38
rs1014259758 Chr.22: 22485548-22485548 on Build GRCh38
rs975026245 Chr.22: 22485473-22485473 on Build GRCh38
rs1023263275 Chr.22: 22485373-22485374 on Build GRCh38
rs995049177 Chr.22: 22485584-22485584 on Build GRCh38
rs1032840133 Chr.22: 22485385-22485385 on Build GRCh38
rs556585974 Chr.22: 22485300-22485300 on Build GRCh38
rs563483259 Chr.22: 22485541-22485541 on Build GRCh38
rs563868299 Chr.22: 22485626-22485626 on Build GRCh38
rs937438835 Chr.22: 22485458-22485458 on Build GRCh38

View Full Product Details