Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 22548408-22548949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRAME
Entrez Gene ID
23532
Gene Name
preferentially expressed antigen in melanoma
Gene Aliases
CT130, MAPE, OIP-4, OIP4
UniGene
Hs.30743
Gene Chromosome Location
Chr.22: 22547696-22559765 on Build GRCh38
Transcripts
NM_001291715.1 NM_001291716.1 NM_001291717.1 NM_001291719.1 NM_001318126.1 NM_001318127.1 NM_006115.4 NM_206953.2 NM_206954.2 NM_206955.2 NM_206956.2 XM_011530034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
326233 c.1166G>T p.G389V Chr.22: 22548431-22548431 on Build GRCh38
6846962 c.1046G>C p.S349T Chr.22: 22548551-22548551 on Build GRCh38
6874737 c.977C>T p.T326I Chr.22: 22548620-22548620 on Build GRCh38
6754754 c.1022A>G p.H341R Chr.22: 22548575-22548575 on Build GRCh38
5019953 c.1150G>A p.V384I Chr.22: 22548447-22548447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765734975 Chr.22: 22548820-22548820 on Build GRCh38
rs754890608 Chr.22: 22548555-22548555 on Build GRCh38
rs776699996 Chr.22: 22548492-22548492 on Build GRCh38
rs779249986 Chr.22: 22548646-22548646 on Build GRCh38
rs146650047 Chr.22: 22548481-22548481 on Build GRCh38
rs760204498 Chr.22: 22548662-22548662 on Build GRCh38
rs991674796 Chr.22: 22548487-22548487 on Build GRCh38
rs2072049 Chr.22: 22548659-22548659 on Build GRCh38
rs746012471 Chr.22: 22548571-22548571 on Build GRCh38
rs867583467 Chr.22: 22548569-22548569 on Build GRCh38
rs762334583 Chr.22: 22548588-22548588 on Build GRCh38
rs776626400 Chr.22: 22548818-22548818 on Build GRCh38
rs766028676 Chr.22: 22548545-22548545 on Build GRCh38
rs143505065 Chr.22: 22548447-22548447 on Build GRCh38
rs781365000 Chr.22: 22548561-22548561 on Build GRCh38
rs78894197 Chr.22: 22548511-22548511 on Build GRCh38
rs780206008 Chr.22: 22548576-22548576 on Build GRCh38
rs552056269 Chr.22: 22548708-22548708 on Build GRCh38
rs753508888 Chr.22: 22548603-22548606 on Build GRCh38
rs536075028 Chr.22: 22548639-22548639 on Build GRCh38
rs772304313 Chr.22: 22548454-22548454 on Build GRCh38
rs888976983 Chr.22: 22548568-22548568 on Build GRCh38
rs996778883 Chr.22: 22548898-22548898 on Build GRCh38
rs765151275 Chr.22: 22548600-22548600 on Build GRCh38
rs770014359 Chr.22: 22548498-22548498 on Build GRCh38
rs753675074 Chr.22: 22548549-22548549 on Build GRCh38
rs750505216 Chr.22: 22548635-22548635 on Build GRCh38
rs528816538 Chr.22: 22548694-22548694 on Build GRCh38
rs760634822 Chr.22: 22548753-22548753 on Build GRCh38
rs879120195 Chr.22: 22548695-22548695 on Build GRCh38
rs987416006 Chr.22: 22548792-22548792 on Build GRCh38
rs543594835 Chr.22: 22548636-22548636 on Build GRCh38
rs115956040 Chr.22: 22548856-22548856 on Build GRCh38
rs771414610 Chr.22: 22548590-22548590 on Build GRCh38
rs774790656 Chr.22: 22548584-22548584 on Build GRCh38
rs185120829 Chr.22: 22548592-22548592 on Build GRCh38
rs956800769 Chr.22: 22548731-22548731 on Build GRCh38
rs1046780267 Chr.22: 22548843-22548843 on Build GRCh38
rs989687236 Chr.22: 22548750-22548750 on Build GRCh38
rs772616695 Chr.22: 22548656-22548656 on Build GRCh38
rs781076827 Chr.22: 22548629-22548629 on Build GRCh38
rs779054676 Chr.22: 22548459-22548459 on Build GRCh38
rs895360335 Chr.22: 22548916-22548916 on Build GRCh38
rs776378195 Chr.22: 22548671-22548671 on Build GRCh38
rs753588169 Chr.22: 22548597-22548597 on Build GRCh38
rs149436129 Chr.22: 22548599-22548599 on Build GRCh38
rs755792225 Chr.22: 22548522-22548522 on Build GRCh38
rs773078291 Chr.22: 22548785-22548785 on Build GRCh38
rs563802116 Chr.22: 22548638-22548638 on Build GRCh38
rs1024372 Chr.22: 22548773-22548773 on Build GRCh38
rs963930386 Chr.22: 22548665-22548665 on Build GRCh38
rs550449035 Chr.22: 22548819-22548819 on Build GRCh38
rs932953263 Chr.22: 22548791-22548791 on Build GRCh38
rs16989516 Chr.22: 22548752-22548752 on Build GRCh38
rs572462431 Chr.22: 22548881-22548881 on Build GRCh38
rs778850355 Chr.22: 22548615-22548615 on Build GRCh38
rs190419590 Chr.22: 22548869-22548869 on Build GRCh38
rs1029229950 Chr.22: 22548904-22548904 on Build GRCh38
rs755812866 Chr.22: 22548431-22548431 on Build GRCh38
rs752997520 Chr.22: 22548841-22548841 on Build GRCh38
rs779927352 Chr.22: 22548439-22548439 on Build GRCh38
rs113467879 Chr.22: 22548519-22548519 on Build GRCh38
rs1038235045 Chr.22: 22548882-22548882 on Build GRCh38
rs61745687 Chr.22: 22548521-22548521 on Build GRCh38
rs1021793642 Chr.22: 22548729-22548729 on Build GRCh38
rs943237362 Chr.22: 22548824-22548824 on Build GRCh38
rs933195145 Chr.22: 22548550-22548550 on Build GRCh38
rs759002381 Chr.22: 22548638-22548638 on Build GRCh38
rs749620212 Chr.22: 22548577-22548577 on Build GRCh38
rs559750257 Chr.22: 22548668-22548668 on Build GRCh38
rs1024373 Chr.22: 22548872-22548872 on Build GRCh38
rs201948825 Chr.22: 22548445-22548445 on Build GRCh38
rs927893661 Chr.22: 22548796-22548796 on Build GRCh38
rs759293094 Chr.22: 22548688-22548688 on Build GRCh38
rs758362238 Chr.22: 22548625-22548625 on Build GRCh38
rs766066868 Chr.22: 22548529-22548530 on Build GRCh38
rs370135401 Chr.22: 22548474-22548474 on Build GRCh38
rs775928243 Chr.22: 22548499-22548499 on Build GRCh38
rs764578632 Chr.22: 22548510-22548510 on Build GRCh38
rs138626954 Chr.22: 22548507-22548507 on Build GRCh38
rs935148315 Chr.22: 22548567-22548567 on Build GRCh38
rs1016445654 Chr.22: 22548652-22548652 on Build GRCh38
rs906569268 Chr.22: 22548877-22548877 on Build GRCh38
rs755769973 Chr.22: 22548917-22548917 on Build GRCh38
rs74876628 Chr.22: 22548640-22548640 on Build GRCh38
rs573594029 Chr.22: 22548450-22548450 on Build GRCh38
rs771178784 Chr.22: 22548490-22548490 on Build GRCh38
rs371645390 Chr.22: 22548560-22548560 on Build GRCh38
rs769057109 Chr.22: 22548582-22548582 on Build GRCh38
rs748593480 Chr.22: 22548651-22548651 on Build GRCh38
rs766053690 Chr.22: 22548596-22548596 on Build GRCh38
rs1044979137 Chr.22: 22548777-22548777 on Build GRCh38
rs746124086 Chr.22: 22548494-22548494 on Build GRCh38
rs760158493 Chr.22: 22548593-22548593 on Build GRCh38
rs752642311 Chr.22: 22548619-22548619 on Build GRCh38
rs987457339 Chr.22: 22548699-22548699 on Build GRCh38
rs770400840 Chr.22: 22548669-22548669 on Build GRCh38
rs968462679 Chr.22: 22548730-22548730 on Build GRCh38
rs963680803 Chr.22: 22548485-22548485 on Build GRCh38
rs137858452 Chr.22: 22548556-22548556 on Build GRCh38
rs2073725 Chr.22: 22548886-22548886 on Build GRCh38

View Full Product Details