Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23180075-23180574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCR
Entrez Gene ID
613
Gene Name
BCR, RhoGEF and GTPase activating protein
Gene Aliases
ALL, BCR1, CML, D22S11, D22S662, PHL
UniGene
Hs.517461
Gene Chromosome Location
Chr.22: 23180365-23318037 on Build GRCh38
Transcripts
NM_004327.3 NM_021574.2

SNP Detail

SNP ID Chromosome Location
rs918029446 Chr.22: 23180383-23180383 on Build GRCh38
rs992154540 Chr.22: 23180373-23180373 on Build GRCh38
rs1050121223 Chr.22: 23180436-23180436 on Build GRCh38
rs1051408275 Chr.22: 23180247-23180247 on Build GRCh38
rs1051030075 Chr.22: 23180414-23180414 on Build GRCh38
rs947407027 Chr.22: 23180208-23180208 on Build GRCh38
rs898279012 Chr.22: 23180462-23180462 on Build GRCh38
rs557716681 Chr.22: 23180294-23180294 on Build GRCh38
rs924581157 Chr.22: 23180540-23180540 on Build GRCh38
rs987099539 Chr.22: 23180542-23180542 on Build GRCh38
rs942568458 Chr.22: 23180448-23180448 on Build GRCh38
rs890925276 Chr.22: 23180425-23180425 on Build GRCh38
rs890965198 Chr.22: 23180262-23180262 on Build GRCh38
rs931127690 Chr.22: 23180410-23180410 on Build GRCh38
rs549455250 Chr.22: 23180170-23180170 on Build GRCh38
rs951645682 Chr.22: 23180298-23180298 on Build GRCh38
rs968203288 Chr.22: 23180300-23180300 on Build GRCh38
rs919764127 Chr.22: 23180409-23180409 on Build GRCh38
rs1056964396 Chr.22: 23180121-23180121 on Build GRCh38
rs566032957 Chr.22: 23180271-23180271 on Build GRCh38
rs879779282 Chr.22: 23180194-23180194 on Build GRCh38
rs974293781 Chr.22: 23180131-23180131 on Build GRCh38
rs947375893 Chr.22: 23180381-23180381 on Build GRCh38
rs1055889260 Chr.22: 23180176-23180176 on Build GRCh38
rs573764419 Chr.22: 23180517-23180517 on Build GRCh38
rs1049627042 Chr.22: 23180197-23180197 on Build GRCh38
rs982047780 Chr.22: 23180169-23180169 on Build GRCh38
rs577583176 Chr.22: 23180453-23180453 on Build GRCh38
rs1022911135 Chr.22: 23180494-23180494 on Build GRCh38
rs999357266 Chr.22: 23180241-23180241 on Build GRCh38
rs1009768872 Chr.22: 23180292-23180292 on Build GRCh38
rs995197207 Chr.22: 23180279-23180279 on Build GRCh38
rs991699155 Chr.22: 23180519-23180519 on Build GRCh38
rs1003505749 Chr.22: 23180487-23180487 on Build GRCh38
rs995828039 Chr.22: 23180195-23180195 on Build GRCh38
rs980097165 Chr.22: 23180324-23180324 on Build GRCh38
rs995164690 Chr.22: 23180475-23180475 on Build GRCh38
rs1021655583 Chr.22: 23180520-23180520 on Build GRCh38
rs939879638 Chr.22: 23180129-23180129 on Build GRCh38
rs867197904 Chr.22: 23180126-23180126 on Build GRCh38
rs112947658 Chr.22: 23180482-23180482 on Build GRCh38
rs958913509 Chr.22: 23180358-23180358 on Build GRCh38
rs966730642 Chr.22: 23180163-23180164 on Build GRCh38
rs937464002 Chr.22: 23180171-23180171 on Build GRCh38
rs1043603943 Chr.22: 23180212-23180212 on Build GRCh38
rs918861834 Chr.22: 23180151-23180151 on Build GRCh38
rs912505128 Chr.22: 23180544-23180544 on Build GRCh38
rs1039593615 Chr.22: 23180556-23180556 on Build GRCh38
rs968812650 Chr.22: 23180522-23180522 on Build GRCh38
rs955758129 Chr.22: 23180291-23180291 on Build GRCh38
rs942516513 Chr.22: 23180548-23180548 on Build GRCh38
rs369718898 Chr.22: 23180365-23180365 on Build GRCh38
rs1039454702 Chr.22: 23180461-23180461 on Build GRCh38
rs557380208 Chr.22: 23180486-23180486 on Build GRCh38
rs1035962140 Chr.22: 23180492-23180492 on Build GRCh38
rs902467131 Chr.22: 23180228-23180228 on Build GRCh38
rs959655143 Chr.22: 23180370-23180370 on Build GRCh38
rs926863976 Chr.22: 23180325-23180325 on Build GRCh38
rs977424739 Chr.22: 23180406-23180406 on Build GRCh38
rs542797530 Chr.22: 23180534-23180534 on Build GRCh38
rs76654237 Chr.22: 23180275-23180275 on Build GRCh38
rs1036865081 Chr.22: 23180407-23180407 on Build GRCh38
rs1018318776 Chr.22: 23180118-23180118 on Build GRCh38
rs1019396083 Chr.22: 23180288-23180288 on Build GRCh38
rs923856393 Chr.22: 23180407-23180408 on Build GRCh38
rs1036812411 Chr.22: 23180158-23180158 on Build GRCh38
rs940100010 Chr.22: 23180388-23180388 on Build GRCh38

View Full Product Details