Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49918415-49918932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG12
Entrez Gene ID
79087
Gene Name
ALG12, alpha-1,6-mannosyltransferase
Gene Aliases
CDG1G, ECM39, PP14673, hALG12
UniGene
Hs.526711
Gene Chromosome Location
Chr.22: 49860163-49918469 on Build GRCh38
Transcripts
NM_024105.3 XM_011530371.2 XM_017028936.1 XM_017028937.1 NM_001135101.2 NM_001284317.1 NM_001284318.1 NM_024324.4 NR_104295.1 XM_005261737.2 XM_005261738.4 XM_011530372.2 XM_017028938.1

Gene Symbol
CRELD2
Entrez Gene ID
79174
Gene Name
cysteine rich with EGF like domains 2
Gene Aliases
-
UniGene
Hs.211282
Gene Chromosome Location
Chr.22: 49918630-49927540 on Build GRCh38
Transcripts
NM_024105.3 XM_011530371.2 XM_017028936.1 XM_017028937.1 NM_001135101.2 NM_001284317.1 NM_001284318.1 NM_024324.4 NR_104295.1 XM_005261737.2 XM_005261738.4 XM_011530372.2 XM_017028938.1

SNP Detail

SNP ID Chromosome Location
rs367593827 Chr.22: 49918528-49918528 on Build GRCh38
rs899656257 Chr.22: 49918561-49918561 on Build GRCh38
rs913515944 Chr.22: 49918466-49918466 on Build GRCh38
rs968937003 Chr.22: 49918478-49918478 on Build GRCh38
rs553525421 Chr.22: 49918642-49918642 on Build GRCh38
rs910193221 Chr.22: 49918628-49918628 on Build GRCh38
rs139081265 Chr.22: 49918806-49918806 on Build GRCh38
rs1043993729 Chr.22: 49918457-49918457 on Build GRCh38
rs9616380 Chr.22: 49918577-49918577 on Build GRCh38
rs969426083 Chr.22: 49918711-49918711 on Build GRCh38
rs1047055478 Chr.22: 49918544-49918544 on Build GRCh38
rs867250364 Chr.22: 49918867-49918867 on Build GRCh38
rs1014952425 Chr.22: 49918698-49918698 on Build GRCh38
rs534446504 Chr.22: 49918627-49918627 on Build GRCh38
rs1026863707 Chr.22: 49918575-49918575 on Build GRCh38
rs890773426 Chr.22: 49918664-49918664 on Build GRCh38
rs535664407 Chr.22: 49918702-49918702 on Build GRCh38
rs964144857 Chr.22: 49918675-49918675 on Build GRCh38
rs890898137 Chr.22: 49918514-49918514 on Build GRCh38
rs979607940 Chr.22: 49918717-49918717 on Build GRCh38
rs956540530 Chr.22: 49918460-49918460 on Build GRCh38
rs977210559 Chr.22: 49918517-49918517 on Build GRCh38
rs181555967 Chr.22: 49918759-49918759 on Build GRCh38
rs1052125187 Chr.22: 49918657-49918657 on Build GRCh38
rs1000939120 Chr.22: 49918856-49918856 on Build GRCh38
rs948357142 Chr.22: 49918446-49918446 on Build GRCh38
rs537014462 Chr.22: 49918512-49918512 on Build GRCh38
rs956972628 Chr.22: 49918731-49918731 on Build GRCh38
rs552217494 Chr.22: 49918554-49918554 on Build GRCh38
rs532042206 Chr.22: 49918669-49918669 on Build GRCh38
rs763027753 Chr.22: 49918780-49918780 on Build GRCh38
rs1003271767 Chr.22: 49918441-49918441 on Build GRCh38
rs985761190 Chr.22: 49918594-49918594 on Build GRCh38
rs575556167 Chr.22: 49918741-49918741 on Build GRCh38
rs978930112 Chr.22: 49918490-49918490 on Build GRCh38
rs375985631 Chr.22: 49918645-49918645 on Build GRCh38
rs562220912 Chr.22: 49918811-49918811 on Build GRCh38
rs943707029 Chr.22: 49918747-49918747 on Build GRCh38
rs369747419 Chr.22: 49918726-49918726 on Build GRCh38
rs1018135724 Chr.22: 49918520-49918520 on Build GRCh38
rs188771916 Chr.22: 49918756-49918756 on Build GRCh38
rs909891881 Chr.22: 49918813-49918813 on Build GRCh38
rs1004117296 Chr.22: 49918782-49918782 on Build GRCh38
rs1039861090 Chr.22: 49918680-49918680 on Build GRCh38
rs929926997 Chr.22: 49918527-49918527 on Build GRCh38
rs772095130 Chr.22: 49918719-49918719 on Build GRCh38
rs755251564 Chr.22: 49918809-49918811 on Build GRCh38
rs943837749 Chr.22: 49918679-49918679 on Build GRCh38
rs1013709823 Chr.22: 49918709-49918709 on Build GRCh38
rs568599282 Chr.22: 49918681-49918681 on Build GRCh38
rs9616381 Chr.22: 49918734-49918734 on Build GRCh38
rs564175465 Chr.22: 49918663-49918663 on Build GRCh38
rs759148799 Chr.22: 49918835-49918835 on Build GRCh38
rs778994692 Chr.22: 49918812-49918814 on Build GRCh38
rs375498114 Chr.22: 49918785-49918785 on Build GRCh38
rs895694670 Chr.22: 49918791-49918791 on Build GRCh38
rs904998943 Chr.22: 49918581-49918581 on Build GRCh38
rs989080602 Chr.22: 49918461-49918461 on Build GRCh38
rs969485163 Chr.22: 49918834-49918834 on Build GRCh38
rs1048189186 Chr.22: 49918686-49918686 on Build GRCh38
rs1056294099 Chr.22: 49918720-49918720 on Build GRCh38
rs903498327 Chr.22: 49918736-49918736 on Build GRCh38
rs939207127 Chr.22: 49918713-49918713 on Build GRCh38
rs948217593 Chr.22: 49918647-49918647 on Build GRCh38
rs936936001 Chr.22: 49918633-49918633 on Build GRCh38
rs995425322 Chr.22: 49918571-49918571 on Build GRCh38
rs573266948 Chr.22: 49918764-49918764 on Build GRCh38
rs1048236974 Chr.22: 49918769-49918769 on Build GRCh38
rs914116553 Chr.22: 49918763-49918763 on Build GRCh38
rs956856030 Chr.22: 49918876-49918876 on Build GRCh38
rs983216911 Chr.22: 49918697-49918697 on Build GRCh38
rs1018441016 Chr.22: 49918742-49918742 on Build GRCh38
rs1024141452 Chr.22: 49918710-49918710 on Build GRCh38
rs1034705542 Chr.22: 49918451-49918451 on Build GRCh38
rs925533364 Chr.22: 49918653-49918653 on Build GRCh38
rs995309292 Chr.22: 49918685-49918685 on Build GRCh38
rs941244481 Chr.22: 49918562-49918562 on Build GRCh38
rs1039339850 Chr.22: 49918752-49918752 on Build GRCh38
rs911373663 Chr.22: 49918493-49918493 on Build GRCh38
rs887042496 Chr.22: 49918690-49918690 on Build GRCh38
rs1052066054 Chr.22: 49918468-49918468 on Build GRCh38
rs903560699 Chr.22: 49918462-49918462 on Build GRCh38
rs888395061 Chr.22: 49918549-49918549 on Build GRCh38
rs886926530 Chr.22: 49918773-49918773 on Build GRCh38
rs535581871 Chr.22: 49918900-49918901 on Build GRCh38
rs1004066301 Chr.22: 49918696-49918696 on Build GRCh38
rs867521586 Chr.22: 49918655-49918655 on Build GRCh38
rs983427895 Chr.22: 49918624-49918624 on Build GRCh38
rs979555558 Chr.22: 49918651-49918651 on Build GRCh38
rs775782693 Chr.22: 49918724-49918724 on Build GRCh38
rs1008376293 Chr.22: 49918518-49918518 on Build GRCh38
rs1057419860 Chr.22: 49918786-49918786 on Build GRCh38
rs113143399 Chr.22: 49918895-49918895 on Build GRCh38
rs934986962 Chr.22: 49918465-49918465 on Build GRCh38
rs1015314370 Chr.22: 49918625-49918625 on Build GRCh38

View Full Product Details