Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50193440-50193945 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRABD
Entrez Gene ID
80305
Gene Name
TraB domain containing
Gene Aliases
LP6054, PP2447
UniGene
Hs.517731
Gene Chromosome Location
Chr.22: 50185889-50199599 on Build GRCh38
Transcripts
NM_001320484.1 NM_001320485.1 NM_001320487.1 NM_001320488.1 NM_025204.3 NR_135275.1 XM_006724422.3 XM_006724423.3 XM_011530712.2 XM_011530713.2 XM_011530715.2 XM_011530716.2 XM_011530717.2 XM_011530718.2 XM_017028943.1 XM_017028944.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4735686 c.74C>T p.P25L Chr.22: 50193616-50193616 on Build GRCh38
1417024 c.57G>A p.P19P Chr.22: 50193599-50193599 on Build GRCh38
3555647 c.56C>T p.P19L Chr.22: 50193598-50193598 on Build GRCh38
1268453 c.62A>T p.E21V Chr.22: 50193604-50193604 on Build GRCh38
5567628 c.59C>G p.S20* Chr.22: 50193601-50193601 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763435979 Chr.22: 50193538-50193538 on Build GRCh38
rs759022716 Chr.22: 50193688-50193688 on Build GRCh38
rs911036324 Chr.22: 50193477-50193477 on Build GRCh38
rs999210938 Chr.22: 50193924-50193924 on Build GRCh38
rs986755516 Chr.22: 50193485-50193485 on Build GRCh38
rs1039131293 Chr.22: 50193490-50193490 on Build GRCh38
rs952941866 Chr.22: 50193875-50193875 on Build GRCh38
rs777999093 Chr.22: 50193567-50193567 on Build GRCh38
rs377072888 Chr.22: 50193659-50193659 on Build GRCh38
rs199578697 Chr.22: 50193687-50193687 on Build GRCh38
rs756193270 Chr.22: 50193566-50193566 on Build GRCh38
rs5845924 Chr.22: 50193739-50193739 on Build GRCh38
rs767996028 Chr.22: 50193640-50193640 on Build GRCh38
rs948251587 Chr.22: 50193840-50193840 on Build GRCh38
rs776067855 Chr.22: 50193641-50193641 on Build GRCh38
rs780951012 Chr.22: 50193671-50193671 on Build GRCh38
rs770357868 Chr.22: 50193694-50193694 on Build GRCh38
rs200520083 Chr.22: 50193681-50193681 on Build GRCh38
rs754032115 Chr.22: 50193572-50193572 on Build GRCh38
rs907858955 Chr.22: 50193756-50193756 on Build GRCh38
rs774852339 Chr.22: 50193695-50193695 on Build GRCh38
rs990930187 Chr.22: 50193883-50193883 on Build GRCh38
rs1056670416 Chr.22: 50193588-50193588 on Build GRCh38
rs903638023 Chr.22: 50193923-50193923 on Build GRCh38
rs779899634 Chr.22: 50193581-50193581 on Build GRCh38
rs144604113 Chr.22: 50193626-50193626 on Build GRCh38
rs77316590 Chr.22: 50193516-50193516 on Build GRCh38
rs764464531 Chr.22: 50193658-50193658 on Build GRCh38
rs751466608 Chr.22: 50193669-50193669 on Build GRCh38
rs751920310 Chr.22: 50193541-50193541 on Build GRCh38
rs571958944 Chr.22: 50193496-50193496 on Build GRCh38
rs368487392 Chr.22: 50193584-50193584 on Build GRCh38
rs939310293 Chr.22: 50193764-50193764 on Build GRCh38
rs767726299 Chr.22: 50193551-50193551 on Build GRCh38
rs539079045 Chr.22: 50193503-50193503 on Build GRCh38
rs767187922 Chr.22: 50193557-50193557 on Build GRCh38
rs895355855 Chr.22: 50193792-50193792 on Build GRCh38
rs554616255 Chr.22: 50193511-50193511 on Build GRCh38
rs916698720 Chr.22: 50193890-50193890 on Build GRCh38
rs1044283739 Chr.22: 50193903-50193903 on Build GRCh38
rs572479491 Chr.22: 50193517-50193517 on Build GRCh38
rs752979431 Chr.22: 50193562-50193562 on Build GRCh38
rs1012802376 Chr.22: 50193612-50193612 on Build GRCh38
rs766291477 Chr.22: 50193665-50193665 on Build GRCh38
rs6010154 Chr.22: 50193616-50193616 on Build GRCh38
rs747984416 Chr.22: 50193680-50193680 on Build GRCh38
rs930713843 Chr.22: 50193718-50193718 on Build GRCh38
rs769492339 Chr.22: 50193605-50193605 on Build GRCh38
rs771425482 Chr.22: 50193620-50193620 on Build GRCh38
rs527264439 Chr.22: 50193774-50193774 on Build GRCh38
rs61473705 Chr.22: 50193599-50193599 on Build GRCh38
rs373734493 Chr.22: 50193558-50193558 on Build GRCh38
rs148680758 Chr.22: 50193478-50193478 on Build GRCh38
rs961346935 Chr.22: 50193804-50193804 on Build GRCh38
rs766844056 Chr.22: 50193539-50193539 on Build GRCh38
rs774738415 Chr.22: 50193622-50193622 on Build GRCh38
rs1052841540 Chr.22: 50193655-50193655 on Build GRCh38
rs1008059030 Chr.22: 50193773-50193773 on Build GRCh38
rs757312892 Chr.22: 50193663-50193663 on Build GRCh38
rs773093046 Chr.22: 50193856-50193856 on Build GRCh38
rs142161138 Chr.22: 50193924-50193925 on Build GRCh38
rs62239319 Chr.22: 50193688-50193688 on Build GRCh38
rs549063990 Chr.22: 50193852-50193852 on Build GRCh38
rs760967692 Chr.22: 50193642-50193642 on Build GRCh38
rs920019887 Chr.22: 50193861-50193861 on Build GRCh38
rs758337950 Chr.22: 50193580-50193580 on Build GRCh38
rs949551589 Chr.22: 50193902-50193902 on Build GRCh38
rs939571908 Chr.22: 50193563-50193563 on Build GRCh38
rs754736050 Chr.22: 50193670-50193670 on Build GRCh38
rs202183574 Chr.22: 50193534-50193534 on Build GRCh38
rs554720663 Chr.22: 50193546-50193546 on Build GRCh38
rs1056745317 Chr.22: 50193775-50193775 on Build GRCh38
rs71318408 Chr.22: 50193531-50193531 on Build GRCh38
rs372847795 Chr.22: 50193597-50193597 on Build GRCh38
rs536073403 Chr.22: 50193895-50193895 on Build GRCh38
rs748788228 Chr.22: 50193617-50193617 on Build GRCh38
rs983480754 Chr.22: 50193747-50193747 on Build GRCh38

View Full Product Details