Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50218597-50219090 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBGCP6
Entrez Gene ID
85378
Gene Name
tubulin gamma complex associated protein 6
Gene Aliases
GCP-6, GCP6, MCCRP, MCCRP1, MCPHCR
UniGene
Hs.336431
Gene Chromosome Location
Chr.22: 50217689-50245024 on Build GRCh38
Transcripts
NM_020461.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3065008 c.4729G>A p.A1577T Chr.22: 50218795-50218795 on Build GRCh38
3964426 c.4834C>T p.L1612F Chr.22: 50218608-50218608 on Build GRCh38
4167344 c.4774G>A p.A1592T Chr.22: 50218750-50218750 on Build GRCh38
3065011 c.4689C>T p.S1563S Chr.22: 50218835-50218835 on Build GRCh38
7301297 c.4782C>T p.N1594N Chr.22: 50218742-50218742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940051903 Chr.22: 50218736-50218736 on Build GRCh38
rs577410965 Chr.22: 50219016-50219016 on Build GRCh38
rs773434604 Chr.22: 50218709-50218709 on Build GRCh38
rs201997192 Chr.22: 50218747-50218747 on Build GRCh38
rs750507390 Chr.22: 50218893-50218893 on Build GRCh38
rs775438615 Chr.22: 50218653-50218653 on Build GRCh38
rs200220671 Chr.22: 50218778-50218778 on Build GRCh38
rs780119708 Chr.22: 50218767-50218767 on Build GRCh38
rs750957948 Chr.22: 50218831-50218831 on Build GRCh38
rs766398798 Chr.22: 50219052-50219052 on Build GRCh38
rs758448066 Chr.22: 50218902-50218902 on Build GRCh38
rs761530929 Chr.22: 50218741-50218741 on Build GRCh38
rs747943188 Chr.22: 50218833-50218833 on Build GRCh38
rs753569974 Chr.22: 50218677-50218677 on Build GRCh38
rs763433603 Chr.22: 50218685-50218686 on Build GRCh38
rs752154749 Chr.22: 50218757-50218757 on Build GRCh38
rs760576687 Chr.22: 50218666-50218666 on Build GRCh38
rs548779798 Chr.22: 50218737-50218737 on Build GRCh38
rs554751109 Chr.22: 50218855-50218855 on Build GRCh38
rs961281716 Chr.22: 50219055-50219055 on Build GRCh38
rs771535295 Chr.22: 50218942-50218942 on Build GRCh38
rs141350146 Chr.22: 50218742-50218742 on Build GRCh38
rs138207892 Chr.22: 50218777-50218777 on Build GRCh38
rs912013141 Chr.22: 50218966-50218986 on Build GRCh38
rs371917612 Chr.22: 50218859-50218859 on Build GRCh38
rs758454623 Chr.22: 50219046-50219046 on Build GRCh38
rs755251059 Chr.22: 50218759-50218759 on Build GRCh38
rs777550104 Chr.22: 50218919-50218919 on Build GRCh38
rs374226178 Chr.22: 50218860-50218860 on Build GRCh38
rs767358055 Chr.22: 50218651-50218651 on Build GRCh38
rs749917931 Chr.22: 50218744-50218744 on Build GRCh38
rs756544270 Chr.22: 50218860-50218860 on Build GRCh38
rs762426410 Chr.22: 50218795-50218795 on Build GRCh38
rs760094171 Chr.22: 50218800-50218800 on Build GRCh38
rs997217584 Chr.22: 50218953-50218953 on Build GRCh38
rs141728151 Chr.22: 50218629-50218629 on Build GRCh38
rs775636763 Chr.22: 50218729-50218729 on Build GRCh38
rs777119989 Chr.22: 50219027-50219027 on Build GRCh38
rs376321384 Chr.22: 50218877-50218877 on Build GRCh38
rs748883647 Chr.22: 50218925-50218925 on Build GRCh38
rs768743463 Chr.22: 50218627-50218627 on Build GRCh38
rs764578957 Chr.22: 50218878-50218878 on Build GRCh38
rs762331176 Chr.22: 50219030-50219030 on Build GRCh38
rs780401321 Chr.22: 50218620-50218620 on Build GRCh38
rs55985560 Chr.22: 50218689-50218689 on Build GRCh38
rs762732524 Chr.22: 50218646-50218647 on Build GRCh38
rs756059959 Chr.22: 50219053-50219053 on Build GRCh38
rs753188505 Chr.22: 50218876-50218876 on Build GRCh38
rs775389383 Chr.22: 50218956-50218956 on Build GRCh38
rs770605705 Chr.22: 50218937-50218937 on Build GRCh38
rs779058012 Chr.22: 50218883-50218883 on Build GRCh38
rs965719982 Chr.22: 50218763-50218763 on Build GRCh38
rs144740402 Chr.22: 50218881-50218881 on Build GRCh38
rs748427454 Chr.22: 50218705-50218705 on Build GRCh38
rs897167905 Chr.22: 50218667-50218667 on Build GRCh38
rs781058339 Chr.22: 50218832-50218832 on Build GRCh38
rs776019851 Chr.22: 50219020-50219020 on Build GRCh38
rs770082911 Chr.22: 50218652-50218653 on Build GRCh38
rs761566467 Chr.22: 50218784-50218784 on Build GRCh38
rs141156818 Chr.22: 50218958-50218958 on Build GRCh38
rs987096023 Chr.22: 50218901-50218901 on Build GRCh38
rs764757813 Chr.22: 50218682-50218682 on Build GRCh38
rs755861466 Chr.22: 50218918-50218918 on Build GRCh38
rs372267747 Chr.22: 50218675-50218675 on Build GRCh38
rs1055288575 Chr.22: 50218628-50218628 on Build GRCh38
rs987549976 Chr.22: 50218914-50218923 on Build GRCh38
rs1037029805 Chr.22: 50218663-50218663 on Build GRCh38
rs369748358 Chr.22: 50219059-50219059 on Build GRCh38
rs149368919 Chr.22: 50218824-50218824 on Build GRCh38
rs763640829 Chr.22: 50218649-50218650 on Build GRCh38
rs769725405 Chr.22: 50218707-50218707 on Build GRCh38
rs769431336 Chr.22: 50218788-50218788 on Build GRCh38
rs950159058 Chr.22: 50219029-50219029 on Build GRCh38
rs772123001 Chr.22: 50218720-50218720 on Build GRCh38
rs892926026 Chr.22: 50218816-50218816 on Build GRCh38
rs565372303 Chr.22: 50218913-50218913 on Build GRCh38
rs778608879 Chr.22: 50219062-50219062 on Build GRCh38
rs1011413688 Chr.22: 50218854-50218854 on Build GRCh38
rs548926600 Chr.22: 50218977-50218977 on Build GRCh38
rs370144692 Chr.22: 50218636-50218636 on Build GRCh38
rs1052731526 Chr.22: 50219009-50219010 on Build GRCh38
rs745532555 Chr.22: 50219063-50219063 on Build GRCh38
rs377132506 Chr.22: 50218681-50218681 on Build GRCh38
rs953358449 Chr.22: 50218961-50218961 on Build GRCh38
rs11912397 Chr.22: 50218683-50218683 on Build GRCh38
rs961868989 Chr.22: 50219014-50219014 on Build GRCh38
rs201702737 Chr.22: 50219043-50219043 on Build GRCh38
rs764493267 Chr.22: 50219024-50219024 on Build GRCh38
rs759027531 Chr.22: 50218861-50218861 on Build GRCh38
rs572104432 Chr.22: 50218811-50218811 on Build GRCh38
rs749374620 Chr.22: 50218765-50218765 on Build GRCh38
rs368868828 Chr.22: 50218730-50218730 on Build GRCh38
rs771572928 Chr.22: 50219064-50219064 on Build GRCh38
rs138735427 Chr.22: 50218808-50218808 on Build GRCh38
rs778114333 Chr.22: 50218803-50218803 on Build GRCh38
rs949561836 Chr.22: 50218979-50218979 on Build GRCh38
rs34562250 Chr.22: 50218796-50218796 on Build GRCh38
rs766836537 Chr.22: 50218748-50218748 on Build GRCh38
rs560215964 Chr.22: 50218955-50218955 on Build GRCh38
rs372126473 Chr.22: 50218735-50218735 on Build GRCh38
rs750660343 Chr.22: 50219042-50219042 on Build GRCh38
rs7291133 Chr.22: 50218684-50218684 on Build GRCh38
rs781246922 Chr.22: 50218699-50218699 on Build GRCh38
rs745746422 Chr.22: 50218711-50218711 on Build GRCh38
rs746996997 Chr.22: 50218771-50218771 on Build GRCh38
rs745356037 Chr.22: 50218940-50218940 on Build GRCh38
rs371125566 Chr.22: 50218851-50218851 on Build GRCh38
rs773942283 Chr.22: 50218939-50218939 on Build GRCh38
rs780407364 Chr.22: 50218936-50218946 on Build GRCh38
rs931875371 Chr.22: 50218782-50218782 on Build GRCh38
rs78122876 Chr.22: 50218760-50218760 on Build GRCh38
rs895319593 Chr.22: 50218774-50218774 on Build GRCh38
rs751972366 Chr.22: 50218799-50218799 on Build GRCh38
rs779186476 Chr.22: 50218632-50218632 on Build GRCh38
rs150285388 Chr.22: 50218990-50218990 on Build GRCh38
rs771176657 Chr.22: 50218766-50218766 on Build GRCh38
rs148158491 Chr.22: 50218952-50218952 on Build GRCh38
rs754165072 Chr.22: 50218805-50218805 on Build GRCh38
rs768068414 Chr.22: 50218865-50218865 on Build GRCh38
rs374770901 Chr.22: 50219058-50219058 on Build GRCh38
rs931471854 Chr.22: 50218753-50218753 on Build GRCh38
rs756422159 Chr.22: 50218761-50218761 on Build GRCh38
rs376881715 Chr.22: 50218826-50218826 on Build GRCh38
rs531388861 Chr.22: 50218999-50218999 on Build GRCh38
rs137904544 Chr.22: 50219040-50219040 on Build GRCh38
rs761198452 Chr.22: 50218668-50218669 on Build GRCh38
rs747172446 Chr.22: 50218622-50218622 on Build GRCh38
rs571498246 Chr.22: 50219023-50219023 on Build GRCh38
rs533237937 Chr.22: 50218649-50218649 on Build GRCh38
rs755407411 Chr.22: 50218697-50218697 on Build GRCh38
rs756618208 Chr.22: 50218802-50218802 on Build GRCh38
rs909092761 Chr.22: 50218998-50218998 on Build GRCh38
rs375713039 Chr.22: 50218633-50218633 on Build GRCh38
rs781165862 Chr.22: 50218910-50218910 on Build GRCh38
rs1049491393 Chr.22: 50219039-50219039 on Build GRCh38
rs745462766 Chr.22: 50218845-50218845 on Build GRCh38
rs34104775 Chr.22: 50218790-50218790 on Build GRCh38
rs200158674 Chr.22: 50218630-50218630 on Build GRCh38
rs370418951 Chr.22: 50218880-50218880 on Build GRCh38
rs778198550 Chr.22: 50218762-50218762 on Build GRCh38
rs934328468 Chr.22: 50219006-50219006 on Build GRCh38
rs753092876 Chr.22: 50218801-50218801 on Build GRCh38

View Full Product Details