Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50252818-50253311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPK12
Entrez Gene ID
6300
Gene Name
mitogen-activated protein kinase 12
Gene Aliases
ERK-6, ERK3, ERK6, MAPK 12, P38GAMMA, PRKM12, SAPK-3, SAPK3
UniGene
Hs.432642
Gene Chromosome Location
Chr.22: 50252902-50261759 on Build GRCh38
Transcripts
NM_001303252.1 NM_002969.4

SNP Detail

SNP ID Chromosome Location
rs990235219 Chr.22: 50252864-50252864 on Build GRCh38
rs5771274 Chr.22: 50252868-50252868 on Build GRCh38
rs146012738 Chr.22: 50252996-50252996 on Build GRCh38
rs891271575 Chr.22: 50252898-50252898 on Build GRCh38
rs1037291486 Chr.22: 50253144-50253144 on Build GRCh38
rs921345995 Chr.22: 50252888-50252888 on Build GRCh38
rs756864268 Chr.22: 50253025-50253025 on Build GRCh38
rs368181693 Chr.22: 50253248-50253248 on Build GRCh38
rs528492513 Chr.22: 50253007-50253007 on Build GRCh38
rs530689766 Chr.22: 50253138-50253138 on Build GRCh38
rs983902318 Chr.22: 50252917-50252917 on Build GRCh38
rs763826882 Chr.22: 50252882-50252882 on Build GRCh38
rs1037661873 Chr.22: 50252940-50252940 on Build GRCh38
rs957365311 Chr.22: 50253059-50253059 on Build GRCh38
rs894130479 Chr.22: 50253191-50253191 on Build GRCh38
rs1028353078 Chr.22: 50253168-50253168 on Build GRCh38
rs5771275 Chr.22: 50252968-50252968 on Build GRCh38
rs965696480 Chr.22: 50253127-50253127 on Build GRCh38
rs1041249879 Chr.22: 50253281-50253281 on Build GRCh38
rs984183780 Chr.22: 50253170-50253170 on Build GRCh38
rs556915887 Chr.22: 50252938-50252938 on Build GRCh38
rs750232984 Chr.22: 50253122-50253122 on Build GRCh38
rs908349518 Chr.22: 50253174-50253174 on Build GRCh38
rs952860002 Chr.22: 50252889-50252889 on Build GRCh38
rs532091212 Chr.22: 50253259-50253259 on Build GRCh38
rs1056985963 Chr.22: 50252982-50252982 on Build GRCh38
rs1031966984 Chr.22: 50253035-50253035 on Build GRCh38
rs549793996 Chr.22: 50253277-50253277 on Build GRCh38
rs550861394 Chr.22: 50253068-50253068 on Build GRCh38
rs1020288965 Chr.22: 50253097-50253097 on Build GRCh38
rs997245850 Chr.22: 50253137-50253137 on Build GRCh38
rs879186674 Chr.22: 50253108-50253108 on Build GRCh38
rs562079602 Chr.22: 50253036-50253036 on Build GRCh38
rs931560203 Chr.22: 50253173-50253173 on Build GRCh38
rs570738798 Chr.22: 50253183-50253183 on Build GRCh38
rs557541989 Chr.22: 50252878-50252878 on Build GRCh38
rs992661651 Chr.22: 50253190-50253190 on Build GRCh38
rs904259436 Chr.22: 50253000-50253000 on Build GRCh38
rs2235354 Chr.22: 50253280-50253280 on Build GRCh38
rs575704674 Chr.22: 50252995-50252995 on Build GRCh38
rs952883933 Chr.22: 50253169-50253169 on Build GRCh38
rs1052861023 Chr.22: 50253268-50253268 on Build GRCh38
rs374298606 Chr.22: 50253229-50253229 on Build GRCh38
rs939912802 Chr.22: 50252940-50252940 on Build GRCh38
rs752758155 Chr.22: 50253230-50253230 on Build GRCh38
rs948695483 Chr.22: 50253221-50253221 on Build GRCh38
rs762620959 Chr.22: 50252881-50252881 on Build GRCh38
rs767293245 Chr.22: 50253053-50253053 on Build GRCh38
rs112556228 Chr.22: 50252999-50252999 on Build GRCh38
rs1012679542 Chr.22: 50253220-50253220 on Build GRCh38
rs956922981 Chr.22: 50252853-50252853 on Build GRCh38
rs921263883 Chr.22: 50253071-50253071 on Build GRCh38
rs892649281 Chr.22: 50253064-50253064 on Build GRCh38
rs980159101 Chr.22: 50253223-50253223 on Build GRCh38
rs183920132 Chr.22: 50253274-50253274 on Build GRCh38
rs894266172 Chr.22: 50252998-50252998 on Build GRCh38
rs550750575 Chr.22: 50253103-50253103 on Build GRCh38
rs563518592 Chr.22: 50253131-50253131 on Build GRCh38
rs987277336 Chr.22: 50253102-50253102 on Build GRCh38
rs540227121 Chr.22: 50253032-50253032 on Build GRCh38
rs932748169 Chr.22: 50253080-50253080 on Build GRCh38
rs77032604 Chr.22: 50252856-50252856 on Build GRCh38
rs994425533 Chr.22: 50252966-50252966 on Build GRCh38
rs552533135 Chr.22: 50253182-50253182 on Build GRCh38
rs756012829 Chr.22: 50253130-50253130 on Build GRCh38
rs917148814 Chr.22: 50253202-50253202 on Build GRCh38
rs908402917 Chr.22: 50252939-50252939 on Build GRCh38
rs1032542475 Chr.22: 50252846-50252846 on Build GRCh38
rs780030895 Chr.22: 50253214-50253214 on Build GRCh38
rs142563172 Chr.22: 50252857-50252857 on Build GRCh38

View Full Product Details