Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50446409-50446915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SBF1
Entrez Gene ID
6305
Gene Name
SET binding factor 1
Gene Aliases
CMT4B3, DENND7A, MTMR5
UniGene
Hs.589924
Gene Chromosome Location
Chr.22: 50445000-50475071 on Build GRCh38
Transcripts
NM_002972.3 XM_005261935.2 XM_011530709.2 XM_011530710.2 XM_017028905.1

SNP Detail

SNP ID Chromosome Location
rs908143821 Chr.22: 50446696-50446696 on Build GRCh38
rs757856395 Chr.22: 50446855-50446855 on Build GRCh38
rs762787190 Chr.22: 50446787-50446787 on Build GRCh38
rs149676235 Chr.22: 50446475-50446475 on Build GRCh38
rs905854095 Chr.22: 50446647-50446647 on Build GRCh38
rs750851244 Chr.22: 50446859-50446859 on Build GRCh38
rs756909602 Chr.22: 50446801-50446801 on Build GRCh38
rs764645143 Chr.22: 50446850-50446850 on Build GRCh38
rs372070515 Chr.22: 50446828-50446828 on Build GRCh38
rs929112019 Chr.22: 50446611-50446611 on Build GRCh38
rs1047991016 Chr.22: 50446480-50446480 on Build GRCh38
rs764545573 Chr.22: 50446809-50446809 on Build GRCh38
rs963789457 Chr.22: 50446447-50446447 on Build GRCh38
rs925361936 Chr.22: 50446868-50446868 on Build GRCh38
rs956150871 Chr.22: 50446566-50446566 on Build GRCh38
rs999334552 Chr.22: 50446554-50446554 on Build GRCh38
rs887876787 Chr.22: 50446544-50446552 on Build GRCh38
rs750836085 Chr.22: 50446697-50446697 on Build GRCh38
rs1026323014 Chr.22: 50446530-50446530 on Build GRCh38
rs910197605 Chr.22: 50446586-50446586 on Build GRCh38
rs770645639 Chr.22: 50446837-50446837 on Build GRCh38
rs770876939 Chr.22: 50446880-50446880 on Build GRCh38
rs774090168 Chr.22: 50446881-50446881 on Build GRCh38
rs367882692 Chr.22: 50446826-50446826 on Build GRCh38
rs1051956913 Chr.22: 50446483-50446483 on Build GRCh38
rs557271625 Chr.22: 50446619-50446619 on Build GRCh38
rs747101219 Chr.22: 50446874-50446874 on Build GRCh38
rs941826937 Chr.22: 50446888-50446888 on Build GRCh38
rs968944212 Chr.22: 50446708-50446708 on Build GRCh38
rs758618812 Chr.22: 50446860-50446860 on Build GRCh38
rs2273261 Chr.22: 50446762-50446762 on Build GRCh38
rs369419850 Chr.22: 50446867-50446867 on Build GRCh38
rs903907695 Chr.22: 50446736-50446736 on Build GRCh38
rs975785874 Chr.22: 50446729-50446729 on Build GRCh38
rs555750779 Chr.22: 50446531-50446531 on Build GRCh38
rs1040320148 Chr.22: 50446512-50446512 on Build GRCh38
rs919659486 Chr.22: 50446650-50446650 on Build GRCh38
rs983557245 Chr.22: 50446688-50446688 on Build GRCh38
rs995192909 Chr.22: 50446894-50446894 on Build GRCh38
rs895225115 Chr.22: 50446518-50446518 on Build GRCh38
rs747177637 Chr.22: 50446598-50446598 on Build GRCh38
rs771730295 Chr.22: 50446885-50446885 on Build GRCh38
rs765651964 Chr.22: 50446856-50446856 on Build GRCh38
rs754521068 Chr.22: 50446817-50446818 on Build GRCh38
rs901776143 Chr.22: 50446602-50446602 on Build GRCh38
rs756602489 Chr.22: 50446805-50446805 on Build GRCh38
rs751014871 Chr.22: 50446777-50446777 on Build GRCh38
rs1057116592 Chr.22: 50446703-50446703 on Build GRCh38
rs574091806 Chr.22: 50446578-50446578 on Build GRCh38
rs542111425 Chr.22: 50446618-50446618 on Build GRCh38
rs987883606 Chr.22: 50446433-50446433 on Build GRCh38
rs964043238 Chr.22: 50446627-50446627 on Build GRCh38
rs113982913 Chr.22: 50446816-50446817 on Build GRCh38
rs764944316 Chr.22: 50446819-50446821 on Build GRCh38
rs1035986959 Chr.22: 50446563-50446563 on Build GRCh38
rs759178211 Chr.22: 50446847-50446847 on Build GRCh38
rs970114219 Chr.22: 50446555-50446555 on Build GRCh38
rs766003271 Chr.22: 50446790-50446790 on Build GRCh38
rs1006702202 Chr.22: 50446624-50446624 on Build GRCh38
rs1044184609 Chr.22: 50446549-50446549 on Build GRCh38
rs569083728 Chr.22: 50446455-50446455 on Build GRCh38
rs542583044 Chr.22: 50446531-50446531 on Build GRCh38
rs767105633 Chr.22: 50446848-50446848 on Build GRCh38
rs746032681 Chr.22: 50446825-50446825 on Build GRCh38
rs763413647 Chr.22: 50446716-50446716 on Build GRCh38
rs1030934259 Chr.22: 50446560-50446560 on Build GRCh38
rs567594375 Chr.22: 50446759-50446759 on Build GRCh38
rs368562661 Chr.22: 50446821-50446822 on Build GRCh38
rs988984918 Chr.22: 50446571-50446571 on Build GRCh38
rs975826386 Chr.22: 50446833-50446833 on Build GRCh38
rs868397648 Chr.22: 50446778-50446778 on Build GRCh38
rs539950812 Chr.22: 50446810-50446810 on Build GRCh38
rs867549822 Chr.22: 50446502-50446502 on Build GRCh38
rs139400090 Chr.22: 50446463-50446463 on Build GRCh38
rs749078847 Chr.22: 50446836-50446836 on Build GRCh38
rs187386678 Chr.22: 50446493-50446493 on Build GRCh38
rs757750146 Chr.22: 50446817-50446817 on Build GRCh38
rs190647894 Chr.22: 50446661-50446661 on Build GRCh38
rs943262003 Chr.22: 50446494-50446494 on Build GRCh38
rs752874940 Chr.22: 50446832-50446833 on Build GRCh38
rs999367363 Chr.22: 50446753-50446753 on Build GRCh38
rs145443464 Chr.22: 50446717-50446717 on Build GRCh38
rs755081050 Chr.22: 50446877-50446877 on Build GRCh38
rs766994616 Chr.22: 50446804-50446804 on Build GRCh38
rs901676557 Chr.22: 50446515-50446515 on Build GRCh38
rs775031992 Chr.22: 50446849-50446849 on Build GRCh38
rs779269509 Chr.22: 50446824-50446824 on Build GRCh38
rs550186623 Chr.22: 50446890-50446890 on Build GRCh38
rs1040691856 Chr.22: 50446599-50446599 on Build GRCh38
rs923167352 Chr.22: 50446731-50446731 on Build GRCh38
rs913627728 Chr.22: 50446577-50446577 on Build GRCh38
rs903709868 Chr.22: 50446552-50446552 on Build GRCh38
rs374844330 Chr.22: 50446891-50446891 on Build GRCh38
rs752406108 Chr.22: 50446820-50446823 on Build GRCh38
rs1043849520 Chr.22: 50446726-50446726 on Build GRCh38
rs973880049 Chr.22: 50446644-50446644 on Build GRCh38
rs373741978 Chr.22: 50446879-50446879 on Build GRCh38
rs941911061 Chr.22: 50446763-50446763 on Build GRCh38
rs1008310509 Chr.22: 50446580-50446580 on Build GRCh38
rs757983848 Chr.22: 50446823-50446824 on Build GRCh38
rs905957561 Chr.22: 50446784-50446784 on Build GRCh38
rs775905931 Chr.22: 50446671-50446671 on Build GRCh38
rs765609195 Chr.22: 50446659-50446659 on Build GRCh38
rs956290940 Chr.22: 50446685-50446685 on Build GRCh38
rs909133779 Chr.22: 50446886-50446886 on Build GRCh38
rs774145195 Chr.22: 50446840-50446840 on Build GRCh38
rs767144189 Chr.22: 50446854-50446854 on Build GRCh38
rs565245585 Chr.22: 50446767-50446767 on Build GRCh38
rs745897282 Chr.22: 50446862-50446863 on Build GRCh38
rs745689851 Chr.22: 50446884-50446884 on Build GRCh38
rs773192469 Chr.22: 50446835-50446835 on Build GRCh38
rs943083451 Chr.22: 50446594-50446597 on Build GRCh38
rs1012373845 Chr.22: 50446522-50446522 on Build GRCh38
rs759191780 Chr.22: 50446796-50446796 on Build GRCh38
rs1008179555 Chr.22: 50446765-50446765 on Build GRCh38
rs549319267 Chr.22: 50446481-50446481 on Build GRCh38
rs751279641 Chr.22: 50446792-50446792 on Build GRCh38
rs781254344 Chr.22: 50446878-50446878 on Build GRCh38

View Full Product Details