Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50783581-50784105 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABL2B
Entrez Gene ID
11158
Gene Name
RAB, member of RAS oncogene family-like 2B
Gene Aliases
-
UniGene
Hs.584862
Gene Chromosome Location
Chr.22: 50767492-50783705 on Build GRCh38
Transcripts
NM_001003789.1 NM_001130919.1 NM_001130920.1 NM_001130921.1 NM_001130922.1 NM_001130923.1 NM_007081.2 XM_005261899.2 XM_011530668.1 XM_011530669.2 XM_011530670.1 XM_011530671.1 XM_011530673.1 XM_011530675.1 XM_017028543.1 XM_017028544.1 XM_017028545.1 XM_017028546.1 XM_017028547.1 XM_017028548.1 XM_017028549.1 XM_017028550.1 XM_017028552.1 XM_017028553.1 XM_017028554.1 XM_017028555.1 XM_017028556.1 XM_017028557.1 XM_017028558.1 NR_026981.1 NR_026982.1

Gene Symbol
RPL23AP82
Entrez Gene ID
284942
Gene Name
ribosomal protein L23a pseudogene 82
Gene Aliases
RPL23A_43_1761
UniGene
Hs.406135
Gene Chromosome Location
Chr.22: 50757086-50799637 on Build GRCh38
Transcripts
NM_001003789.1 NM_001130919.1 NM_001130920.1 NM_001130921.1 NM_001130922.1 NM_001130923.1 NM_007081.2 XM_005261899.2 XM_011530668.1 XM_011530669.2 XM_011530670.1 XM_011530671.1 XM_011530673.1 XM_011530675.1 XM_017028543.1 XM_017028544.1 XM_017028545.1 XM_017028546.1 XM_017028547.1 XM_017028548.1 XM_017028549.1 XM_017028550.1 XM_017028552.1 XM_017028553.1 XM_017028554.1 XM_017028555.1 XM_017028556.1 XM_017028557.1 XM_017028558.1 NR_026981.1 NR_026982.1

SNP Detail

SNP ID Chromosome Location
rs527778704 Chr.22: 50783645-50783645 on Build GRCh38
rs2879915 Chr.22: 50783624-50783624 on Build GRCh38
rs550865986 Chr.22: 50783743-50783743 on Build GRCh38
rs374739078 Chr.22: 50783678-50783678 on Build GRCh38
rs760935913 Chr.22: 50783924-50783925 on Build GRCh38
rs551276100 Chr.22: 50783867-50783867 on Build GRCh38
rs569151671 Chr.22: 50783785-50783785 on Build GRCh38
rs147301669 Chr.22: 50783605-50783605 on Build GRCh38
rs562765971 Chr.22: 50783604-50783604 on Build GRCh38
rs3217568 Chr.22: 50783597-50783597 on Build GRCh38
rs539678603 Chr.22: 50783791-50783791 on Build GRCh38
rs544469444 Chr.22: 50783601-50783601 on Build GRCh38
rs569235293 Chr.22: 50783638-50783638 on Build GRCh38
rs774100236 Chr.22: 50783654-50783654 on Build GRCh38
rs566566146 Chr.22: 50783951-50783951 on Build GRCh38
rs545257994 Chr.22: 50783617-50783617 on Build GRCh38
rs549360228 Chr.22: 50783652-50783652 on Build GRCh38
rs555358604 Chr.22: 50784037-50784037 on Build GRCh38
rs144822152 Chr.22: 50783823-50783823 on Build GRCh38
rs533613154 Chr.22: 50784027-50784027 on Build GRCh38

View Full Product Details