Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3153332-3153833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
UniGene
Hs.18925
Gene Chromosome Location
Chr.3: 3148490-3179717 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2 XM_011533778.2

Gene Symbol
TRNT1
Entrez Gene ID
51095
Gene Name
tRNA nucleotidyl transferase 1
Gene Aliases
CCA1, CGI-47, MtCCA, RPEM, SIFD
UniGene
Hs.732725
Gene Chromosome Location
Chr.3: 3126916-3153435 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2 XM_011533778.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1043613 c.997A>G p.T333A Chr.3: 3153443-3153443 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775474416 Chr.3: 3153385-3153385 on Build GRCh38
rs752097742 Chr.3: 3153447-3153447 on Build GRCh38
rs972504554 Chr.3: 3153757-3153757 on Build GRCh38
rs918628526 Chr.3: 3153473-3153473 on Build GRCh38
rs531432826 Chr.3: 3153367-3153367 on Build GRCh38
rs753929515 Chr.3: 3153395-3153395 on Build GRCh38
rs865869655 Chr.3: 3153585-3153585 on Build GRCh38
rs6793531 Chr.3: 3153667-3153667 on Build GRCh38
rs936385112 Chr.3: 3153722-3153722 on Build GRCh38
rs920068984 Chr.3: 3153631-3153631 on Build GRCh38
rs770665841 Chr.3: 3153470-3153470 on Build GRCh38
rs370167783 Chr.3: 3153491-3153491 on Build GRCh38
rs563428675 Chr.3: 3153363-3153363 on Build GRCh38
rs547623806 Chr.3: 3153408-3153408 on Build GRCh38
rs567197193 Chr.3: 3153459-3153459 on Build GRCh38
rs766562089 Chr.3: 3153408-3153408 on Build GRCh38
rs373754376 Chr.3: 3153407-3153407 on Build GRCh38
rs148975864 Chr.3: 3153481-3153481 on Build GRCh38
rs921018060 Chr.3: 3153702-3153702 on Build GRCh38
rs767533718 Chr.3: 3153401-3153401 on Build GRCh38
rs979468075 Chr.3: 3153370-3153370 on Build GRCh38
rs918451673 Chr.3: 3153799-3153799 on Build GRCh38
rs376902894 Chr.3: 3153411-3153411 on Build GRCh38
rs761957765 Chr.3: 3153398-3153398 on Build GRCh38
rs763155197 Chr.3: 3153530-3153530 on Build GRCh38
rs556328468 Chr.3: 3153483-3153483 on Build GRCh38
rs1041910901 Chr.3: 3153650-3153650 on Build GRCh38
rs753497042 Chr.3: 3153649-3153649 on Build GRCh38
rs778647676 Chr.3: 3153472-3153472 on Build GRCh38
rs1009525432 Chr.3: 3153554-3153554 on Build GRCh38
rs776654473 Chr.3: 3153516-3153516 on Build GRCh38
rs73005226 Chr.3: 3153685-3153685 on Build GRCh38
rs892379739 Chr.3: 3153541-3153541 on Build GRCh38
rs931420102 Chr.3: 3153490-3153490 on Build GRCh38
rs867924924 Chr.3: 3153721-3153721 on Build GRCh38
rs974246393 Chr.3: 3153629-3153629 on Build GRCh38
rs551310625 Chr.3: 3153383-3153383 on Build GRCh38
rs905902110 Chr.3: 3153576-3153576 on Build GRCh38
rs755596263 Chr.3: 3153449-3153449 on Build GRCh38
rs545194008 Chr.3: 3153386-3153386 on Build GRCh38
rs17027577 Chr.3: 3153641-3153641 on Build GRCh38
rs958749374 Chr.3: 3153615-3153615 on Build GRCh38
rs937570386 Chr.3: 3153602-3153602 on Build GRCh38
rs750707474 Chr.3: 3153404-3153404 on Build GRCh38
rs760603312 Chr.3: 3153501-3153501 on Build GRCh38
rs773456257 Chr.3: 3153525-3153525 on Build GRCh38
rs202139535 Chr.3: 3153391-3153391 on Build GRCh38
rs902227431 Chr.3: 3153653-3153653 on Build GRCh38
rs760397636 Chr.3: 3153436-3153436 on Build GRCh38
rs780326607 Chr.3: 3153445-3153445 on Build GRCh38
rs748809085 Chr.3: 3153465-3153465 on Build GRCh38
rs1047175338 Chr.3: 3153374-3153374 on Build GRCh38
rs569745851 Chr.3: 3153519-3153519 on Build GRCh38
rs1019464627 Chr.3: 3153727-3153727 on Build GRCh38
rs759405801 Chr.3: 3153377-3153377 on Build GRCh38
rs533854593 Chr.3: 3153402-3153402 on Build GRCh38
rs546193693 Chr.3: 3153642-3153642 on Build GRCh38
rs376683996 Chr.3: 3153520-3153520 on Build GRCh38
rs952904250 Chr.3: 3153804-3153804 on Build GRCh38
rs930212105 Chr.3: 3153368-3153368 on Build GRCh38
rs965266932 Chr.3: 3153749-3153749 on Build GRCh38
rs1048867153 Chr.3: 3153561-3153561 on Build GRCh38
rs953762977 Chr.3: 3153710-3153710 on Build GRCh38
rs538707484 Chr.3: 3153569-3153569 on Build GRCh38

View Full Product Details