Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 7452487-7453020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM7
Entrez Gene ID
2917
Gene Name
glutamate metabotropic receptor 7
Gene Aliases
GLUR7, GPRC1G, MGLU7, MGLUR7, PPP1R87
UniGene
Hs.606393
Gene Chromosome Location
Chr.3: 6861097-7743032 on Build GRCh38
Transcripts
NM_000844.3 NM_181874.2 XM_017006272.1 XM_017006273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4327227 c.1199C>T p.S400F Chr.3: 7452631-7452631 on Build GRCh38
1048470 c.1184G>T p.R395I Chr.3: 7452616-7452616 on Build GRCh38
5821621 c.1261G>A p.A421T Chr.3: 7452693-7452693 on Build GRCh38
6165689 c.1331G>C p.G444A Chr.3: 7452763-7452763 on Build GRCh38
3597437 c.1184G>A p.R395K Chr.3: 7452616-7452616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751565946 Chr.3: 7452824-7452824 on Build GRCh38
rs750104645 Chr.3: 7452559-7452568 on Build GRCh38
rs775493357 Chr.3: 7452647-7452647 on Build GRCh38
rs138874035 Chr.3: 7452788-7452788 on Build GRCh38
rs888669895 Chr.3: 7452871-7452871 on Build GRCh38
rs750120625 Chr.3: 7452797-7452797 on Build GRCh38
rs771348052 Chr.3: 7452847-7452847 on Build GRCh38
rs750951751 Chr.3: 7452574-7452574 on Build GRCh38
rs34067965 Chr.3: 7452611-7452611 on Build GRCh38
rs1040152824 Chr.3: 7452987-7452987 on Build GRCh38
rs377452581 Chr.3: 7452887-7452887 on Build GRCh38
rs764466911 Chr.3: 7452584-7452585 on Build GRCh38
rs767039541 Chr.3: 7452679-7452679 on Build GRCh38
rs761621971 Chr.3: 7452790-7452790 on Build GRCh38
rs752088753 Chr.3: 7452684-7452684 on Build GRCh38
rs201868190 Chr.3: 7452732-7452732 on Build GRCh38
rs778942212 Chr.3: 7452601-7452601 on Build GRCh38
rs749023869 Chr.3: 7452598-7452598 on Build GRCh38
rs754109879 Chr.3: 7452587-7452588 on Build GRCh38
rs754981300 Chr.3: 7452825-7452825 on Build GRCh38
rs144471689 Chr.3: 7452872-7452872 on Build GRCh38
rs763387881 Chr.3: 7452758-7452758 on Build GRCh38
rs770193953 Chr.3: 7452667-7452667 on Build GRCh38
rs369533386 Chr.3: 7452692-7452692 on Build GRCh38
rs765674409 Chr.3: 7452591-7452592 on Build GRCh38
rs773180357 Chr.3: 7452559-7452570 on Build GRCh38
rs370004780 Chr.3: 7452575-7452575 on Build GRCh38
rs762174725 Chr.3: 7452562-7452562 on Build GRCh38
rs773626333 Chr.3: 7452746-7452746 on Build GRCh38
rs771275844 Chr.3: 7452770-7452770 on Build GRCh38
rs776426278 Chr.3: 7452557-7452558 on Build GRCh38
rs770141077 Chr.3: 7452744-7452744 on Build GRCh38
rs755778338 Chr.3: 7452590-7452590 on Build GRCh38
rs764998178 Chr.3: 7452794-7452794 on Build GRCh38
rs748409723 Chr.3: 7452743-7452743 on Build GRCh38
rs1041573031 Chr.3: 7452908-7452908 on Build GRCh38
rs909879833 Chr.3: 7452556-7452556 on Build GRCh38
rs752389621 Chr.3: 7452588-7452588 on Build GRCh38
rs758214571 Chr.3: 7452731-7452731 on Build GRCh38
rs888798261 Chr.3: 7452985-7452985 on Build GRCh38
rs781371497 Chr.3: 7452736-7452736 on Build GRCh38
rs747998584 Chr.3: 7452559-7452562 on Build GRCh38
rs765696371 Chr.3: 7452570-7452570 on Build GRCh38
rs929999266 Chr.3: 7452545-7452545 on Build GRCh38
rs537011473 Chr.3: 7452772-7452772 on Build GRCh38
rs547700677 Chr.3: 7452527-7452527 on Build GRCh38
rs1054359491 Chr.3: 7452954-7452954 on Build GRCh38
rs964784976 Chr.3: 7452585-7452586 on Build GRCh38
rs758248021 Chr.3: 7452798-7452798 on Build GRCh38
rs1007743861 Chr.3: 7452750-7452750 on Build GRCh38
rs1015979958 Chr.3: 7452514-7452514 on Build GRCh38
rs749446984 Chr.3: 7452836-7452836 on Build GRCh38
rs778889222 Chr.3: 7452728-7452728 on Build GRCh38
rs974891778 Chr.3: 7452930-7452930 on Build GRCh38
rs578189404 Chr.3: 7452848-7452848 on Build GRCh38
rs779809743 Chr.3: 7452818-7452818 on Build GRCh38
rs2229902 Chr.3: 7452730-7452730 on Build GRCh38
rs746422532 Chr.3: 7452849-7452849 on Build GRCh38
rs763439420 Chr.3: 7452672-7452672 on Build GRCh38
rs865838895 Chr.3: 7452748-7452748 on Build GRCh38
rs772157224 Chr.3: 7452633-7452633 on Build GRCh38
rs895792774 Chr.3: 7452805-7452805 on Build GRCh38
rs565859845 Chr.3: 7452554-7452554 on Build GRCh38
rs760819535 Chr.3: 7452578-7452579 on Build GRCh38
rs767522388 Chr.3: 7452558-7452559 on Build GRCh38
rs774952581 Chr.3: 7452784-7452784 on Build GRCh38
rs757092015 Chr.3: 7452726-7452726 on Build GRCh38
rs781318433 Chr.3: 7452827-7452827 on Build GRCh38
rs143220145 Chr.3: 7452662-7452662 on Build GRCh38
rs541082898 Chr.3: 7452833-7452833 on Build GRCh38
rs935944311 Chr.3: 7452943-7452943 on Build GRCh38
rs149410668 Chr.3: 7452867-7452867 on Build GRCh38
rs753563518 Chr.3: 7452722-7452722 on Build GRCh38
rs777495534 Chr.3: 7452597-7452597 on Build GRCh38
rs561648918 Chr.3: 7452940-7452940 on Build GRCh38
rs796298391 Chr.3: 7452587-7452588 on Build GRCh38
rs954544122 Chr.3: 7452955-7452955 on Build GRCh38
rs1007203033 Chr.3: 7452986-7452986 on Build GRCh38
rs139571456 Chr.3: 7452904-7452904 on Build GRCh38
rs111752136 Chr.3: 7452559-7452560 on Build GRCh38
rs144288627 Chr.3: 7452651-7452651 on Build GRCh38
rs547860504 Chr.3: 7452537-7452537 on Build GRCh38
rs553602809 Chr.3: 7452839-7452839 on Build GRCh38
rs34555823 Chr.3: 7452818-7452818 on Build GRCh38
rs369592692 Chr.3: 7452733-7452733 on Build GRCh38
rs748279203 Chr.3: 7452713-7452713 on Build GRCh38
rs769944164 Chr.3: 7452834-7452834 on Build GRCh38
rs776873783 Chr.3: 7452656-7452656 on Build GRCh38
rs903009731 Chr.3: 7452931-7452931 on Build GRCh38
rs763798098 Chr.3: 7452709-7452709 on Build GRCh38
rs758361455 Chr.3: 7452567-7452568 on Build GRCh38
rs766978646 Chr.3: 7452577-7452577 on Build GRCh38
rs773679767 Chr.3: 7452671-7452671 on Build GRCh38
rs760052014 Chr.3: 7452785-7452785 on Build GRCh38
rs201779962 Chr.3: 7452600-7452600 on Build GRCh38
rs768274394 Chr.3: 7452856-7452856 on Build GRCh38

View Full Product Details