Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 8878605-8879129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD18
Entrez Gene ID
56852
Gene Name
RAD18, E3 ubiquitin protein ligase
Gene Aliases
RNF73
UniGene
Hs.375684
Gene Chromosome Location
Chr.3: 8877196-8969249 on Build GRCh38
Transcripts
NM_020165.3

SNP Detail

SNP ID Chromosome Location
rs1028744383 Chr.3: 8878698-8878698 on Build GRCh38
rs760994754 Chr.3: 8878805-8878805 on Build GRCh38
rs1032552728 Chr.3: 8879060-8879060 on Build GRCh38
rs528936305 Chr.3: 8878709-8878709 on Build GRCh38
rs887727566 Chr.3: 8878637-8878637 on Build GRCh38
rs184332893 Chr.3: 8878955-8878955 on Build GRCh38
rs1045519387 Chr.3: 8878783-8878783 on Build GRCh38
rs563742954 Chr.3: 8878964-8878964 on Build GRCh38
rs73810710 Chr.3: 8878779-8878779 on Build GRCh38
rs549702729 Chr.3: 8878638-8878638 on Build GRCh38
rs576256643 Chr.3: 8879026-8879026 on Build GRCh38
rs774456197 Chr.3: 8878633-8878633 on Build GRCh38
rs762640510 Chr.3: 8878700-8878703 on Build GRCh38
rs1014917345 Chr.3: 8878826-8878826 on Build GRCh38
rs45538936 Chr.3: 8879027-8879027 on Build GRCh38
rs988138845 Chr.3: 8878754-8878754 on Build GRCh38
rs561327719 Chr.3: 8878909-8878909 on Build GRCh38
rs574311563 Chr.3: 8878819-8878819 on Build GRCh38
rs537993456 Chr.3: 8878739-8878739 on Build GRCh38
rs182520863 Chr.3: 8878731-8878731 on Build GRCh38
rs187160373 Chr.3: 8878737-8878737 on Build GRCh38
rs193920 Chr.3: 8878821-8878821 on Build GRCh38
rs1001423983 Chr.3: 8879047-8879047 on Build GRCh38
rs766830843 Chr.3: 8878781-8878781 on Build GRCh38
rs767777766 Chr.3: 8878689-8878689 on Build GRCh38
rs527973429 Chr.3: 8878697-8878697 on Build GRCh38
rs887691983 Chr.3: 8878881-8878881 on Build GRCh38
rs111345608 Chr.3: 8878774-8878774 on Build GRCh38
rs935949743 Chr.3: 8878743-8878743 on Build GRCh38
rs560794661 Chr.3: 8878696-8878696 on Build GRCh38
rs973693823 Chr.3: 8878908-8878908 on Build GRCh38
rs935630755 Chr.3: 8879032-8879032 on Build GRCh38
rs761858726 Chr.3: 8878673-8878673 on Build GRCh38
rs963685383 Chr.3: 8878891-8878891 on Build GRCh38
rs997210977 Chr.3: 8878968-8878968 on Build GRCh38
rs1027954551 Chr.3: 8878631-8878631 on Build GRCh38
rs904462160 Chr.3: 8878741-8878741 on Build GRCh38
rs372209221 Chr.3: 8878822-8878822 on Build GRCh38
rs1014581137 Chr.3: 8878659-8878659 on Build GRCh38
rs1055716995 Chr.3: 8879048-8879048 on Build GRCh38
rs150735038 Chr.3: 8878790-8878790 on Build GRCh38
rs142774791 Chr.3: 8878744-8878744 on Build GRCh38
rs965758732 Chr.3: 8878675-8878675 on Build GRCh38

View Full Product Details