Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 8912089-8912601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAD18
Entrez Gene ID
56852
Gene Name
RAD18, E3 ubiquitin protein ligase
Gene Aliases
RNF73
UniGene
Hs.375684
Gene Chromosome Location
Chr.3: 8877196-8969249 on Build GRCh38
Transcripts
NM_020165.3 XM_017006873.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1582967 c.1016A>G p.H339R Chr.3: 8912323-8912323 on Build GRCh38
5355407 c.1027+1G>A p.? Chr.3: 8912311-8912311 on Build GRCh38
238055 c.1017C>T p.H339H Chr.3: 8912322-8912322 on Build GRCh38
5834751 c.983delA p.K328fs*8 Chr.3: 8912356-8912356 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775598102 Chr.3: 8912405-8912405 on Build GRCh38
rs575494131 Chr.3: 8912488-8912488 on Build GRCh38
rs751740905 Chr.3: 8912412-8912412 on Build GRCh38
rs749788340 Chr.3: 8912401-8912402 on Build GRCh38
rs184558744 Chr.3: 8912531-8912531 on Build GRCh38
rs933230040 Chr.3: 8912257-8912257 on Build GRCh38
rs778461934 Chr.3: 8912383-8912384 on Build GRCh38
rs756750695 Chr.3: 8912347-8912347 on Build GRCh38
rs45472192 Chr.3: 8912554-8912554 on Build GRCh38
rs920499736 Chr.3: 8912307-8912307 on Build GRCh38
rs769662100 Chr.3: 8912380-8912380 on Build GRCh38
rs764026833 Chr.3: 8912301-8912301 on Build GRCh38
rs775322622 Chr.3: 8912295-8912295 on Build GRCh38
rs924053242 Chr.3: 8912210-8912210 on Build GRCh38
rs1019095774 Chr.3: 8912177-8912177 on Build GRCh38
rs34098429 Chr.3: 8912360-8912361 on Build GRCh38
rs778433947 Chr.3: 8912348-8912348 on Build GRCh38
rs1002236200 Chr.3: 8912383-8912383 on Build GRCh38
rs968111766 Chr.3: 8912200-8912200 on Build GRCh38
rs374743075 Chr.3: 8912277-8912277 on Build GRCh38
rs45603638 Chr.3: 8912147-8912148 on Build GRCh38
rs748622829 Chr.3: 8912343-8912343 on Build GRCh38
rs753735730 Chr.3: 8912306-8912306 on Build GRCh38
rs917044026 Chr.3: 8912252-8912252 on Build GRCh38
rs780548862 Chr.3: 8912322-8912322 on Build GRCh38
rs746761762 Chr.3: 8912370-8912370 on Build GRCh38
rs766639371 Chr.3: 8912403-8912403 on Build GRCh38
rs1026098304 Chr.3: 8912138-8912138 on Build GRCh38
rs886174795 Chr.3: 8912161-8912161 on Build GRCh38
rs6772242 Chr.3: 8912522-8912522 on Build GRCh38
rs961023452 Chr.3: 8912431-8912431 on Build GRCh38
rs964798092 Chr.3: 8912542-8912542 on Build GRCh38
rs781681255 Chr.3: 8912338-8912338 on Build GRCh38
rs988729156 Chr.3: 8912570-8912570 on Build GRCh38
rs752025834 Chr.3: 8912331-8912331 on Build GRCh38
rs771822989 Chr.3: 8912294-8912294 on Build GRCh38
rs992632029 Chr.3: 8912239-8912239 on Build GRCh38
rs199978402 Chr.3: 8912385-8912385 on Build GRCh38
rs758739014 Chr.3: 8912321-8912321 on Build GRCh38
rs750646650 Chr.3: 8912316-8912316 on Build GRCh38
rs537791255 Chr.3: 8912205-8912205 on Build GRCh38
rs765443201 Chr.3: 8912312-8912312 on Build GRCh38
rs554623610 Chr.3: 8912255-8912255 on Build GRCh38
rs368820398 Chr.3: 8912311-8912311 on Build GRCh38
rs189343842 Chr.3: 8912567-8912567 on Build GRCh38
rs761699400 Chr.3: 8912382-8912382 on Build GRCh38
rs545294411 Chr.3: 8912436-8912436 on Build GRCh38
rs558639766 Chr.3: 8912362-8912362 on Build GRCh38
rs45598638 Chr.3: 8912227-8912227 on Build GRCh38
rs763259767 Chr.3: 8912402-8912402 on Build GRCh38
rs775129950 Chr.3: 8912369-8912369 on Build GRCh38
rs770360102 Chr.3: 8912269-8912269 on Build GRCh38
rs980330733 Chr.3: 8912570-8912570 on Build GRCh38
rs768361215 Chr.3: 8912372-8912372 on Build GRCh38
rs1022967175 Chr.3: 8912196-8912196 on Build GRCh38
rs756849157 Chr.3: 8912402-8912403 on Build GRCh38
rs773943026 Chr.3: 8912272-8912272 on Build GRCh38
rs769048598 Chr.3: 8912125-8912125 on Build GRCh38
rs540613480 Chr.3: 8912349-8912349 on Build GRCh38
rs561033027 Chr.3: 8912525-8912525 on Build GRCh38
rs1009092633 Chr.3: 8912170-8912170 on Build GRCh38
rs1011374416 Chr.3: 8912332-8912332 on Build GRCh38
rs76654438 Chr.3: 8912461-8912461 on Build GRCh38
rs200667948 Chr.3: 8912344-8912344 on Build GRCh38
rs773479743 Chr.3: 8912391-8912391 on Build GRCh38
rs537284462 Chr.3: 8912145-8912145 on Build GRCh38
rs573304645 Chr.3: 8912259-8912259 on Build GRCh38
rs992162304 Chr.3: 8912533-8912533 on Build GRCh38
rs1040567099 Chr.3: 8912379-8912379 on Build GRCh38
rs999553377 Chr.3: 8912501-8912501 on Build GRCh38
rs974983642 Chr.3: 8912183-8912183 on Build GRCh38
rs748762707 Chr.3: 8912268-8912268 on Build GRCh38
rs760526324 Chr.3: 8912300-8912300 on Build GRCh38
rs9312029 Chr.3: 8912182-8912182 on Build GRCh38
rs111253431 Chr.3: 8912287-8912287 on Build GRCh38

View Full Product Details