Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9759249-9759751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK1
Entrez Gene ID
8536
Gene Name
calcium/calmodulin dependent protein kinase I
Gene Aliases
CAMKI
UniGene
Hs.434875
Gene Chromosome Location
Chr.3: 9757345-9769987 on Build GRCh38
Transcripts
NM_003656.4 XM_005265516.1 XM_005265517.2 XM_017007354.1 NM_016821.2 NM_016826.2 NM_016827.2 NM_016828.2 NM_016829.2 XM_011533760.1 XM_017006494.1 XM_017006496.1 XM_017006497.1 XM_017006498.1 XM_017006499.1

Gene Symbol
OGG1
Entrez Gene ID
4968
Gene Name
8-oxoguanine DNA glycosylase
Gene Aliases
HMMH, HOGG1, MUTM, OGH1
UniGene
Hs.380271
Gene Chromosome Location
Chr.3: 9749944-9788246 on Build GRCh38
Transcripts
NM_003656.4 XM_005265516.1 XM_005265517.2 XM_017007354.1 NM_016821.2 NM_016826.2 NM_016827.2 NM_016828.2 NM_016829.2 XM_011533760.1 XM_017006494.1 XM_017006496.1 XM_017006497.1 XM_017006498.1 XM_017006499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1692345 c.751G>A p.D251N Chr.3: 9759745-9759745 on Build GRCh38
1049184 c.762G>A p.R254R Chr.3: 9759734-9759734 on Build GRCh38
7261024 c.888_890delGAA p.K296delK Chr.3: 9759510-9759512 on Build GRCh38
1753418 c.759C>T p.I253I Chr.3: 9759737-9759737 on Build GRCh38
3916803 c.778G>A p.D260N Chr.3: 9759718-9759718 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748549507 Chr.3: 9759631-9759631 on Build GRCh38
rs534136001 Chr.3: 9759463-9759463 on Build GRCh38
rs142058092 Chr.3: 9759309-9759309 on Build GRCh38
rs377182467 Chr.3: 9759493-9759493 on Build GRCh38
rs924785759 Chr.3: 9759322-9759322 on Build GRCh38
rs370723271 Chr.3: 9759633-9759633 on Build GRCh38
rs758465129 Chr.3: 9759521-9759521 on Build GRCh38
rs774836718 Chr.3: 9759585-9759585 on Build GRCh38
rs745947683 Chr.3: 9759518-9759518 on Build GRCh38
rs771326810 Chr.3: 9759642-9759642 on Build GRCh38
rs778754920 Chr.3: 9759515-9759515 on Build GRCh38
rs200157460 Chr.3: 9759702-9759702 on Build GRCh38
rs760109896 Chr.3: 9759653-9759653 on Build GRCh38
rs761538555 Chr.3: 9759593-9759593 on Build GRCh38
rs767669718 Chr.3: 9759297-9759298 on Build GRCh38
rs755688457 Chr.3: 9759483-9759483 on Build GRCh38
rs1005315104 Chr.3: 9759277-9759277 on Build GRCh38
rs758422677 Chr.3: 9759605-9759605 on Build GRCh38
rs753634244 Chr.3: 9759591-9759591 on Build GRCh38
rs747307663 Chr.3: 9759526-9759526 on Build GRCh38
rs748829054 Chr.3: 9759277-9759278 on Build GRCh38
rs796304481 Chr.3: 9759460-9759460 on Build GRCh38
rs56046762 Chr.3: 9759641-9759641 on Build GRCh38
rs897683532 Chr.3: 9759475-9759476 on Build GRCh38
rs982289547 Chr.3: 9759598-9759598 on Build GRCh38
rs150806322 Chr.3: 9759524-9759524 on Build GRCh38
rs775733732 Chr.3: 9759321-9759321 on Build GRCh38
rs970740599 Chr.3: 9759668-9759668 on Build GRCh38
rs938290261 Chr.3: 9759610-9759610 on Build GRCh38
rs913984992 Chr.3: 9759529-9759529 on Build GRCh38
rs778632031 Chr.3: 9759337-9759340 on Build GRCh38
rs766768473 Chr.3: 9759447-9759447 on Build GRCh38
rs888116881 Chr.3: 9759630-9759630 on Build GRCh38
rs760975814 Chr.3: 9759444-9759444 on Build GRCh38
rs187470736 Chr.3: 9759640-9759640 on Build GRCh38
rs377602451 Chr.3: 9759617-9759617 on Build GRCh38
rs566055751 Chr.3: 9759616-9759616 on Build GRCh38
rs758249202 Chr.3: 9759603-9759603 on Build GRCh38
rs765190978 Chr.3: 9759594-9759594 on Build GRCh38
rs148790679 Chr.3: 9759676-9759676 on Build GRCh38
rs560292298 Chr.3: 9759682-9759682 on Build GRCh38
rs139923053 Chr.3: 9759533-9759533 on Build GRCh38
rs752350819 Chr.3: 9759482-9759482 on Build GRCh38
rs562375406 Chr.3: 9759595-9759595 on Build GRCh38
rs536390884 Chr.3: 9759393-9759393 on Build GRCh38
rs775400164 Chr.3: 9759714-9759714 on Build GRCh38
rs1015314553 Chr.3: 9759324-9759324 on Build GRCh38
rs767095223 Chr.3: 9759465-9759465 on Build GRCh38
rs370718078 Chr.3: 9759679-9759679 on Build GRCh38
rs367901510 Chr.3: 9759701-9759701 on Build GRCh38
rs558795108 Chr.3: 9759464-9759464 on Build GRCh38
rs773723897 Chr.3: 9759549-9759549 on Build GRCh38
rs199997024 Chr.3: 9759684-9759684 on Build GRCh38
rs916163430 Chr.3: 9759651-9759651 on Build GRCh38
rs765837789 Chr.3: 9759459-9759459 on Build GRCh38
rs939127948 Chr.3: 9759440-9759440 on Build GRCh38
rs573056321 Chr.3: 9759429-9759429 on Build GRCh38
rs372987286 Chr.3: 9759571-9759571 on Build GRCh38
rs2069058 Chr.3: 9759339-9759339 on Build GRCh38
rs763922235 Chr.3: 9759488-9759488 on Build GRCh38
rs762470544 Chr.3: 9759449-9759449 on Build GRCh38
rs745955867 Chr.3: 9759297-9759297 on Build GRCh38
rs745642487 Chr.3: 9759510-9759512 on Build GRCh38
rs771650402 Chr.3: 9759573-9759573 on Build GRCh38
rs879772293 Chr.3: 9759392-9759392 on Build GRCh38
rs544171008 Chr.3: 9759586-9759586 on Build GRCh38
rs770441132 Chr.3: 9759542-9759542 on Build GRCh38
rs779020582 Chr.3: 9759283-9759283 on Build GRCh38
rs747365546 Chr.3: 9759331-9759331 on Build GRCh38
rs768987307 Chr.3: 9759530-9759530 on Build GRCh38
rs2304277 Chr.3: 9759396-9759396 on Build GRCh38
rs781511507 Chr.3: 9759623-9759623 on Build GRCh38
rs757250734 Chr.3: 9759504-9759504 on Build GRCh38
rs184042619 Chr.3: 9759387-9759387 on Build GRCh38
rs994677617 Chr.3: 9759491-9759491 on Build GRCh38
rs547741149 Chr.3: 9759584-9759584 on Build GRCh38
rs748459644 Chr.3: 9759539-9759539 on Build GRCh38
rs755257698 Chr.3: 9759622-9759622 on Build GRCh38
rs776833307 Chr.3: 9759534-9759534 on Build GRCh38
rs772039812 Chr.3: 9759701-9759702 on Build GRCh38
rs373982598 Chr.3: 9759337-9759337 on Build GRCh38
rs775132172 Chr.3: 9759650-9759650 on Build GRCh38

View Full Product Details