Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 9878499-9879009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIDEC
Entrez Gene ID
63924
Gene Name
cell death inducing DFFA like effector c
Gene Aliases
CIDE-3, CIDE3, FPLD5, FSP27
UniGene
Hs.567562
Gene Chromosome Location
Chr.3: 9866710-9880254 on Build GRCh38
Transcripts
NM_001199551.1 NM_001199552.1 NM_001199623.1 NM_001321142.1 NM_001321143.1 NM_001321144.1 NM_022094.3

SNP Detail

SNP ID Chromosome Location
rs768704792 Chr.3: 9878556-9878556 on Build GRCh38
rs970362914 Chr.3: 9878606-9878606 on Build GRCh38
rs1009947675 Chr.3: 9878796-9878796 on Build GRCh38
rs370106080 Chr.3: 9878586-9878586 on Build GRCh38
rs1017397029 Chr.3: 9878878-9878878 on Build GRCh38
rs180765357 Chr.3: 9878881-9878881 on Build GRCh38
rs957117903 Chr.3: 9878526-9878526 on Build GRCh38
rs557142406 Chr.3: 9878981-9878981 on Build GRCh38
rs777278439 Chr.3: 9878521-9878521 on Build GRCh38
rs918668897 Chr.3: 9878986-9878986 on Build GRCh38
rs745695964 Chr.3: 9878540-9878540 on Build GRCh38
rs933978181 Chr.3: 9878711-9878711 on Build GRCh38
rs756925680 Chr.3: 9878532-9878532 on Build GRCh38
rs746959494 Chr.3: 9878547-9878547 on Build GRCh38
rs58940229 Chr.3: 9878982-9878982 on Build GRCh38
rs538739404 Chr.3: 9878930-9878930 on Build GRCh38
rs763048519 Chr.3: 9878774-9878774 on Build GRCh38
rs11915736 Chr.3: 9878722-9878722 on Build GRCh38
rs943353452 Chr.3: 9878529-9878529 on Build GRCh38
rs548162516 Chr.3: 9878746-9878746 on Build GRCh38
rs377320161 Chr.3: 9878794-9878794 on Build GRCh38
rs373033506 Chr.3: 9878757-9878757 on Build GRCh38
rs142220074 Chr.3: 9878851-9878851 on Build GRCh38
rs771920426 Chr.3: 9878542-9878542 on Build GRCh38
rs958625387 Chr.3: 9878886-9878886 on Build GRCh38
rs765152542 Chr.3: 9878760-9878760 on Build GRCh38
rs189509177 Chr.3: 9878668-9878668 on Build GRCh38
rs897120683 Chr.3: 9878701-9878701 on Build GRCh38
rs909012755 Chr.3: 9878931-9878931 on Build GRCh38
rs1035959620 Chr.3: 9878824-9878824 on Build GRCh38
rs143766859 Chr.3: 9878908-9878908 on Build GRCh38
rs765427479 Chr.3: 9878763-9878763 on Build GRCh38
rs759708837 Chr.3: 9878872-9878872 on Build GRCh38
rs923240892 Chr.3: 9878726-9878726 on Build GRCh38
rs570191450 Chr.3: 9878594-9878594 on Build GRCh38
rs981059361 Chr.3: 9878880-9878880 on Build GRCh38
rs775104397 Chr.3: 9878544-9878544 on Build GRCh38
rs1012144171 Chr.3: 9878766-9878766 on Build GRCh38
rs369738978 Chr.3: 9878932-9878932 on Build GRCh38
rs562660985 Chr.3: 9878700-9878700 on Build GRCh38
rs893300524 Chr.3: 9878793-9878793 on Build GRCh38
rs766772053 Chr.3: 9878799-9878799 on Build GRCh38
rs565006408 Chr.3: 9878667-9878667 on Build GRCh38
rs546870625 Chr.3: 9878562-9878563 on Build GRCh38
rs1026056674 Chr.3: 9878868-9878868 on Build GRCh38
rs1037530985 Chr.3: 9878695-9878695 on Build GRCh38
rs909058055 Chr.3: 9878681-9878681 on Build GRCh38
rs1044601657 Chr.3: 9878767-9878767 on Build GRCh38
rs773505472 Chr.3: 9878765-9878765 on Build GRCh38
rs960369790 Chr.3: 9878679-9878679 on Build GRCh38
rs374888110 Chr.3: 9878558-9878558 on Build GRCh38
rs927204533 Chr.3: 9878650-9878650 on Build GRCh38
rs951864084 Chr.3: 9878869-9878869 on Build GRCh38
rs1053106837 Chr.3: 9878749-9878749 on Build GRCh38
rs748783569 Chr.3: 9878528-9878528 on Build GRCh38
rs371554594 Chr.3: 9878536-9878536 on Build GRCh38
rs62247501 Chr.3: 9878563-9878563 on Build GRCh38
rs139025697 Chr.3: 9878689-9878689 on Build GRCh38
rs906030680 Chr.3: 9878776-9878776 on Build GRCh38
rs993186976 Chr.3: 9878867-9878867 on Build GRCh38
rs890031050 Chr.3: 9878739-9878739 on Build GRCh38

View Full Product Details