Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196802900-196803399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAK2
Entrez Gene ID
5062
Gene Name
p21 (RAC1) activated kinase 2
Gene Aliases
PAK65, PAKgamma
UniGene
Hs.518530
Gene Chromosome Location
Chr.3: 196739857-196832647 on Build GRCh38
Transcripts
NM_002577.4 XM_011512870.2 XM_017006501.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5634772 c.335C>T p.T112I Chr.3: 196803063-196803063 on Build GRCh38
7382257 c.384G>T p.K128N Chr.3: 196803112-196803112 on Build GRCh38
1422035 c.303G>C p.Q101H Chr.3: 196803031-196803031 on Build GRCh38
5648972 c.420G>C p.L140L Chr.3: 196803148-196803148 on Build GRCh38
1422037 c.436_436+1insA p.K147fs*28 Chr.3: 196803164-196803165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55926500 Chr.3: 196803043-196803043 on Build GRCh38
rs765399513 Chr.3: 196803175-196803175 on Build GRCh38
rs756303352 Chr.3: 196803097-196803097 on Build GRCh38
rs768588878 Chr.3: 196803188-196803188 on Build GRCh38
rs757566107 Chr.3: 196803109-196803109 on Build GRCh38
rs76649479 Chr.3: 196803275-196803275 on Build GRCh38
rs73205842 Chr.3: 196803142-196803142 on Build GRCh38
rs926621438 Chr.3: 196803352-196803352 on Build GRCh38
rs752800616 Chr.3: 196803093-196803093 on Build GRCh38
rs746894928 Chr.3: 196803186-196803186 on Build GRCh38
rs1045856583 Chr.3: 196803362-196803362 on Build GRCh38
rs774766455 Chr.3: 196803148-196803148 on Build GRCh38
rs772030520 Chr.3: 196803115-196803115 on Build GRCh38
rs755457188 Chr.3: 196802970-196802970 on Build GRCh38
rs970822924 Chr.3: 196803296-196803296 on Build GRCh38
rs756162157 Chr.3: 196803165-196803167 on Build GRCh38
rs781268182 Chr.3: 196802972-196802972 on Build GRCh38
rs373906638 Chr.3: 196803174-196803174 on Build GRCh38
rs183020057 Chr.3: 196803046-196803046 on Build GRCh38
rs115224945 Chr.3: 196803151-196803151 on Build GRCh38
rs755506544 Chr.3: 196803081-196803081 on Build GRCh38
rs549385623 Chr.3: 196802945-196802945 on Build GRCh38
rs908596437 Chr.3: 196802956-196802956 on Build GRCh38
rs147476390 Chr.3: 196803014-196803014 on Build GRCh38
rs753970309 Chr.3: 196803165-196803165 on Build GRCh38
rs775335636 Chr.3: 196803118-196803118 on Build GRCh38
rs768110153 Chr.3: 196803133-196803133 on Build GRCh38
rs922187489 Chr.3: 196803235-196803235 on Build GRCh38
rs745649967 Chr.3: 196803171-196803171 on Build GRCh38
rs763365578 Chr.3: 196803267-196803267 on Build GRCh38
rs753242485 Chr.3: 196803164-196803165 on Build GRCh38
rs752060550 Chr.3: 196802968-196802968 on Build GRCh38
rs746983645 Chr.3: 196802946-196802946 on Build GRCh38
rs757545981 Chr.3: 196803166-196803166 on Build GRCh38
rs751408265 Chr.3: 196803106-196803106 on Build GRCh38
rs866264341 Chr.3: 196803373-196803373 on Build GRCh38
rs780679132 Chr.3: 196803193-196803193 on Build GRCh38
rs375638470 Chr.3: 196803008-196803008 on Build GRCh38
rs982890959 Chr.3: 196802951-196802951 on Build GRCh38
rs773359514 Chr.3: 196803260-196803260 on Build GRCh38
rs770128324 Chr.3: 196802978-196802978 on Build GRCh38
rs747716302 Chr.3: 196803203-196803203 on Build GRCh38
rs749060657 Chr.3: 196802989-196802989 on Build GRCh38
rs994785146 Chr.3: 196803220-196803220 on Build GRCh38
rs200828622 Chr.3: 196803124-196803124 on Build GRCh38
rs778190333 Chr.3: 196802987-196802987 on Build GRCh38
rs764211225 Chr.3: 196803163-196803163 on Build GRCh38
rs1027947867 Chr.3: 196803237-196803237 on Build GRCh38
rs1018527689 Chr.3: 196803170-196803170 on Build GRCh38
rs756710402 Chr.3: 196803161-196803161 on Build GRCh38
rs78187178 Chr.3: 196802950-196802950 on Build GRCh38
rs201816624 Chr.3: 196803183-196803183 on Build GRCh38
rs775058686 Chr.3: 196803012-196803012 on Build GRCh38
rs753216350 Chr.3: 196803156-196803156 on Build GRCh38
rs557616773 Chr.3: 196803152-196803152 on Build GRCh38
rs748244692 Chr.3: 196802976-196802976 on Build GRCh38
rs982177941 Chr.3: 196803351-196803351 on Build GRCh38
rs764867800 Chr.3: 196803146-196803146 on Build GRCh38
rs139723099 Chr.3: 196803269-196803269 on Build GRCh38
rs759985265 Chr.3: 196803001-196803002 on Build GRCh38
rs766696285 Chr.3: 196802970-196802971 on Build GRCh38
rs761391530 Chr.3: 196803145-196803145 on Build GRCh38
rs775482488 Chr.3: 196802928-196802928 on Build GRCh38
rs79726945 Chr.3: 196803107-196803107 on Build GRCh38
rs78043821 Chr.3: 196803111-196803111 on Build GRCh38
rs77470308 Chr.3: 196803055-196803055 on Build GRCh38
rs768040650 Chr.3: 196803156-196803158 on Build GRCh38
rs771905631 Chr.3: 196803011-196803011 on Build GRCh38
rs539574340 Chr.3: 196803119-196803119 on Build GRCh38
rs761898736 Chr.3: 196803049-196803049 on Build GRCh38
rs201465227 Chr.3: 196803031-196803031 on Build GRCh38
rs776632073 Chr.3: 196803039-196803039 on Build GRCh38
rs767968693 Chr.3: 196803082-196803082 on Build GRCh38
rs770808918 Chr.3: 196802991-196802991 on Build GRCh38
rs112793531 Chr.3: 196803004-196803004 on Build GRCh38
rs200335533 Chr.3: 196803213-196803213 on Build GRCh38
rs975374641 Chr.3: 196803198-196803198 on Build GRCh38
rs774037198 Chr.3: 196803003-196803003 on Build GRCh38
rs1055064944 Chr.3: 196803009-196803009 on Build GRCh38
rs751930141 Chr.3: 196802980-196802981 on Build GRCh38

View Full Product Details