Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 196946260-196946757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGZ
Entrez Gene ID
80235
Gene Name
phosphatidylinositol glycan anchor biosynthesis class Z
Gene Aliases
GPI-MT-IV, PIG-Z, SMP3
UniGene
Hs.518403
Gene Chromosome Location
Chr.3: 196946343-196969411 on Build GRCh38
Transcripts
NM_025163.3 XM_011513190.1 XM_011513192.2 XM_017007240.1 XM_017007241.1 XM_017007242.1 XM_017007243.1

SNP Detail

SNP ID Chromosome Location
rs976991378 Chr.3: 196946683-196946683 on Build GRCh38
rs957308210 Chr.3: 196946674-196946674 on Build GRCh38
rs540008018 Chr.3: 196946688-196946688 on Build GRCh38
rs775693898 Chr.3: 196946420-196946420 on Build GRCh38
rs913149775 Chr.3: 196946430-196946430 on Build GRCh38
rs546982626 Chr.3: 196946726-196946726 on Build GRCh38
rs906497840 Chr.3: 196946721-196946721 on Build GRCh38
rs564807419 Chr.3: 196946543-196946543 on Build GRCh38
rs116262443 Chr.3: 196946627-196946627 on Build GRCh38
rs928472206 Chr.3: 196946339-196946339 on Build GRCh38
rs187160686 Chr.3: 196946505-196946505 on Build GRCh38
rs979855894 Chr.3: 196946636-196946636 on Build GRCh38
rs1020126073 Chr.3: 196946285-196946285 on Build GRCh38
rs893820398 Chr.3: 196946481-196946481 on Build GRCh38
rs530720868 Chr.3: 196946696-196946696 on Build GRCh38
rs900538222 Chr.3: 196946506-196946506 on Build GRCh38
rs944295379 Chr.3: 196946663-196946663 on Build GRCh38
rs1035937056 Chr.3: 196946601-196946601 on Build GRCh38
rs576466607 Chr.3: 196946720-196946720 on Build GRCh38
rs527449610 Chr.3: 196946357-196946357 on Build GRCh38
rs551875828 Chr.3: 196946385-196946385 on Build GRCh38
rs921449393 Chr.3: 196946687-196946687 on Build GRCh38
rs912427406 Chr.3: 196946710-196946710 on Build GRCh38
rs982485003 Chr.3: 196946642-196946642 on Build GRCh38
rs549742562 Chr.3: 196946470-196946470 on Build GRCh38
rs886702556 Chr.3: 196946558-196946558 on Build GRCh38
rs752861194 Chr.3: 196946525-196946525 on Build GRCh38
rs559927096 Chr.3: 196946306-196946306 on Build GRCh38
rs930930381 Chr.3: 196946360-196946360 on Build GRCh38
rs1046756888 Chr.3: 196946376-196946376 on Build GRCh38
rs981723833 Chr.3: 196946309-196946309 on Build GRCh38
rs1005064008 Chr.3: 196946620-196946621 on Build GRCh38
rs145585261 Chr.3: 196946439-196946439 on Build GRCh38
rs945459439 Chr.3: 196946714-196946714 on Build GRCh38
rs1050927520 Chr.3: 196946449-196946449 on Build GRCh38
rs138016717 Chr.3: 196946697-196946697 on Build GRCh38
rs967303559 Chr.3: 196946584-196946584 on Build GRCh38
rs991661886 Chr.3: 196946610-196946610 on Build GRCh38
rs1009007062 Chr.3: 196946471-196946471 on Build GRCh38
rs1002761449 Chr.3: 196946596-196946596 on Build GRCh38
rs908200359 Chr.3: 196946644-196946644 on Build GRCh38
rs935672486 Chr.3: 196946403-196946403 on Build GRCh38
rs952422104 Chr.3: 196946628-196946628 on Build GRCh38
rs767536082 Chr.3: 196946467-196946467 on Build GRCh38
rs369571713 Chr.3: 196946394-196946394 on Build GRCh38
rs991228050 Chr.3: 196946354-196946354 on Build GRCh38
rs566289287 Chr.3: 196946638-196946638 on Build GRCh38
rs373210332 Chr.3: 196946535-196946535 on Build GRCh38
rs774187157 Chr.3: 196946415-196946415 on Build GRCh38
rs997358542 Chr.3: 196946455-196946455 on Build GRCh38
rs890937979 Chr.3: 196946459-196946459 on Build GRCh38
rs1028031399 Chr.3: 196946546-196946546 on Build GRCh38
rs944284516 Chr.3: 196946406-196946406 on Build GRCh38
rs958711460 Chr.3: 196946346-196946346 on Build GRCh38
rs536156161 Chr.3: 196946603-196946603 on Build GRCh38

View Full Product Details