Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 271160-271661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF732
Entrez Gene ID
654254
Gene Name
zinc finger protein 732
Gene Aliases
-
UniGene
Hs.698668
Gene Chromosome Location
Chr.4: 270675-305321 on Build GRCh38
Transcripts
NM_001137608.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6235540 c.1253+2T>G p.? Chr.4: 271431-271431 on Build GRCh38
1328549 c.1268G>C p.C423S Chr.4: 271589-271589 on Build GRCh38
351013 c.1179G>A p.E393E Chr.4: 271507-271507 on Build GRCh38
6291580 c.1266G>A p.E422E Chr.4: 271591-271591 on Build GRCh38
4744853 c.1581_1582delGA p.K528fs*3 Chr.4: 271275-271276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782809336 Chr.4: 271308-271308 on Build GRCh38
rs376489653 Chr.4: 271565-271565 on Build GRCh38
rs782768494 Chr.4: 271199-271199 on Build GRCh38
rs782679142 Chr.4: 271541-271542 on Build GRCh38
rs782276416 Chr.4: 271578-271579 on Build GRCh38
rs782422402 Chr.4: 271430-271430 on Build GRCh38
rs200663767 Chr.4: 271235-271235 on Build GRCh38
rs782021676 Chr.4: 271508-271508 on Build GRCh38
rs782509381 Chr.4: 271278-271278 on Build GRCh38
rs782252556 Chr.4: 271361-271362 on Build GRCh38
rs979029918 Chr.4: 271285-271285 on Build GRCh38
rs61792065 Chr.4: 271566-271566 on Build GRCh38
rs781828171 Chr.4: 271458-271458 on Build GRCh38
rs782037753 Chr.4: 271432-271433 on Build GRCh38
rs1029881076 Chr.4: 271218-271218 on Build GRCh38
rs61792073 Chr.4: 271622-271622 on Build GRCh38
rs371788816 Chr.4: 271562-271562 on Build GRCh38
rs782045603 Chr.4: 271340-271340 on Build GRCh38
rs782080556 Chr.4: 271426-271427 on Build GRCh38
rs782392167 Chr.4: 271479-271479 on Build GRCh38
rs782061740 Chr.4: 271538-271538 on Build GRCh38
rs782168045 Chr.4: 271510-271510 on Build GRCh38
rs782807148 Chr.4: 271236-271236 on Build GRCh38
rs61792068 Chr.4: 271591-271591 on Build GRCh38
rs782474866 Chr.4: 271459-271459 on Build GRCh38
rs61792074 Chr.4: 271625-271625 on Build GRCh38
rs782236186 Chr.4: 271524-271524 on Build GRCh38
rs782538668 Chr.4: 271368-271368 on Build GRCh38
rs967676985 Chr.4: 271276-271276 on Build GRCh38
rs543518794 Chr.4: 271573-271573 on Build GRCh38
rs61792061 Chr.4: 271535-271535 on Build GRCh38
rs782729531 Chr.4: 271477-271478 on Build GRCh38
rs782798271 Chr.4: 271440-271440 on Build GRCh38
rs781902406 Chr.4: 271541-271541 on Build GRCh38
rs374403050 Chr.4: 271433-271433 on Build GRCh38
rs980226947 Chr.4: 271597-271597 on Build GRCh38
rs782084365 Chr.4: 271631-271631 on Build GRCh38
rs781784647 Chr.4: 271231-271231 on Build GRCh38
rs781868973 Chr.4: 271352-271352 on Build GRCh38
rs1017932893 Chr.4: 271272-271272 on Build GRCh38
rs890949954 Chr.4: 271483-271483 on Build GRCh38
rs782774524 Chr.4: 271449-271449 on Build GRCh38
rs782773527 Chr.4: 271599-271599 on Build GRCh38
rs782677287 Chr.4: 271439-271439 on Build GRCh38
rs782233438 Chr.4: 271284-271284 on Build GRCh38
rs781990551 Chr.4: 271312-271312 on Build GRCh38
rs781999604 Chr.4: 271208-271208 on Build GRCh38
rs577455458 Chr.4: 271558-271558 on Build GRCh38
rs61792064 Chr.4: 271560-271560 on Build GRCh38
rs549742787 Chr.4: 271215-271215 on Build GRCh38
rs61792075 Chr.4: 271638-271638 on Build GRCh38
rs782574291 Chr.4: 271460-271460 on Build GRCh38
rs782012955 Chr.4: 271577-271577 on Build GRCh38
rs782680241 Chr.4: 271472-271472 on Build GRCh38
rs34359434 Chr.4: 271511-271512 on Build GRCh38
rs782244153 Chr.4: 271474-271474 on Build GRCh38
rs781812000 Chr.4: 271512-271512 on Build GRCh38
rs61792067 Chr.4: 271580-271580 on Build GRCh38
rs781911985 Chr.4: 271441-271441 on Build GRCh38
rs782586534 Chr.4: 271292-271292 on Build GRCh38
rs61792072 Chr.4: 271610-271610 on Build GRCh38
rs369875361 Chr.4: 271544-271544 on Build GRCh38
rs994032603 Chr.4: 271188-271188 on Build GRCh38
rs61792070 Chr.4: 271602-271602 on Build GRCh38
rs956506823 Chr.4: 271551-271551 on Build GRCh38
rs566267637 Chr.4: 271350-271350 on Build GRCh38
rs144087243 Chr.4: 271439-271439 on Build GRCh38
rs782782303 Chr.4: 271540-271540 on Build GRCh38
rs781969293 Chr.4: 271315-271315 on Build GRCh38
rs782704448 Chr.4: 271366-271366 on Build GRCh38
rs573513849 Chr.4: 271442-271442 on Build GRCh38
rs782379870 Chr.4: 271511-271511 on Build GRCh38
rs782605604 Chr.4: 271283-271283 on Build GRCh38
rs561907401 Chr.4: 271226-271226 on Build GRCh38
rs782433261 Chr.4: 271553-271553 on Build GRCh38
rs781783458 Chr.4: 271536-271536 on Build GRCh38
rs201614981 Chr.4: 271406-271406 on Build GRCh38
rs782435737 Chr.4: 271287-271287 on Build GRCh38
rs370745325 Chr.4: 271399-271399 on Build GRCh38
rs182419378 Chr.4: 271519-271519 on Build GRCh38
rs782367086 Chr.4: 271385-271386 on Build GRCh38
rs61792063 Chr.4: 271552-271552 on Build GRCh38
rs34590377 Chr.4: 271509-271510 on Build GRCh38
rs781802433 Chr.4: 271465-271465 on Build GRCh38
rs782009847 Chr.4: 271514-271514 on Build GRCh38
rs782692142 Chr.4: 271369-271369 on Build GRCh38
rs782177730 Chr.4: 271501-271501 on Build GRCh38
rs782224334 Chr.4: 271624-271624 on Build GRCh38
rs61792071 Chr.4: 271608-271608 on Build GRCh38
rs782649660 Chr.4: 271547-271547 on Build GRCh38
rs200469451 Chr.4: 271600-271610 on Build GRCh38
rs782709395 Chr.4: 271343-271343 on Build GRCh38
rs369273736 Chr.4: 271575-271575 on Build GRCh38
rs782551745 Chr.4: 271224-271224 on Build GRCh38
rs376120998 Chr.4: 271370-271370 on Build GRCh38
rs61792066 Chr.4: 271570-271570 on Build GRCh38
rs782175929 Chr.4: 271574-271574 on Build GRCh38
rs1024510867 Chr.4: 271556-271556 on Build GRCh38
rs782222740 Chr.4: 271395-271395 on Build GRCh38
rs903758476 Chr.4: 271518-271518 on Build GRCh38
rs782280449 Chr.4: 271306-271306 on Build GRCh38
rs781922657 Chr.4: 271437-271437 on Build GRCh38
rs782014315 Chr.4: 271344-271344 on Build GRCh38
rs61792069 Chr.4: 271598-271598 on Build GRCh38
rs782154428 Chr.4: 271594-271594 on Build GRCh38
rs782080337 Chr.4: 271213-271213 on Build GRCh38
rs781897945 Chr.4: 271250-271250 on Build GRCh38
rs142565151 Chr.4: 271365-271365 on Build GRCh38
rs782635418 Chr.4: 271605-271605 on Build GRCh38
rs61792062 Chr.4: 271550-271550 on Build GRCh38
rs547735121 Chr.4: 271245-271245 on Build GRCh38
rs782279252 Chr.4: 271427-271427 on Build GRCh38
rs782689531 Chr.4: 271581-271581 on Build GRCh38
rs782531289 Chr.4: 271470-271470 on Build GRCh38
rs927495434 Chr.4: 271423-271423 on Build GRCh38
rs781807972 Chr.4: 271367-271367 on Build GRCh38
rs782001523 Chr.4: 271628-271628 on Build GRCh38
rs782534382 Chr.4: 271374-271374 on Build GRCh38
rs368791157 Chr.4: 271435-271435 on Build GRCh38
rs782322082 Chr.4: 271506-271506 on Build GRCh38
rs782712801 Chr.4: 271431-271432 on Build GRCh38

View Full Product Details