Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 851651-852150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAK
Entrez Gene ID
2580
Gene Name
cyclin G associated kinase
Gene Aliases
DNAJ26, DNAJC26
UniGene
Hs.369607
Gene Chromosome Location
Chr.4: 849275-932390 on Build GRCh38
Transcripts
NM_001318134.1 NM_005255.3 XM_005272268.2 XM_005272270.2 XM_011513425.2 XM_011513426.2 XM_011513427.2 XM_011513428.2 XM_011513429.2 XM_011513430.1 XM_011513431.2 XM_011513432.2 XM_011513434.2 XM_017007991.1 XM_017007992.1 XM_017007993.1 XM_017007994.1 XM_017007995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1431357 c.3368G>A p.R1123Q Chr.4: 851890-851890 on Build GRCh38
7369119 c.3393G>T p.W1131C Chr.4: 851865-851865 on Build GRCh38
1735495 c.3417delC p.A1141fs*15 Chr.4: 851841-851841 on Build GRCh38
7369120 c.3360G>C p.Q1120H Chr.4: 851898-851898 on Build GRCh38
275252 c.3453G>A p.S1151S Chr.4: 851805-851805 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs925282455 Chr.4: 852118-852118 on Build GRCh38
rs141041086 Chr.4: 851846-851846 on Build GRCh38
rs374362699 Chr.4: 852121-852121 on Build GRCh38
rs753861522 Chr.4: 851849-851849 on Build GRCh38
rs748803243 Chr.4: 851761-851761 on Build GRCh38
rs536094884 Chr.4: 852098-852098 on Build GRCh38
rs200146913 Chr.4: 851790-851790 on Build GRCh38
rs750564137 Chr.4: 851774-851774 on Build GRCh38
rs773687541 Chr.4: 851834-851834 on Build GRCh38
rs769191426 Chr.4: 851927-851928 on Build GRCh38
rs142037003 Chr.4: 851949-851949 on Build GRCh38
rs749986894 Chr.4: 851919-851919 on Build GRCh38
rs569597016 Chr.4: 852054-852054 on Build GRCh38
rs367691274 Chr.4: 851871-851871 on Build GRCh38
rs746338370 Chr.4: 851954-851954 on Build GRCh38
rs758315487 Chr.4: 851994-851994 on Build GRCh38
rs151123471 Chr.4: 851842-851842 on Build GRCh38
rs56326341 Chr.4: 851755-851755 on Build GRCh38
rs184983128 Chr.4: 852090-852090 on Build GRCh38
rs200295882 Chr.4: 852013-852013 on Build GRCh38
rs145100581 Chr.4: 851915-851915 on Build GRCh38
rs772120961 Chr.4: 851841-851841 on Build GRCh38
rs764561876 Chr.4: 851706-851706 on Build GRCh38
rs975720982 Chr.4: 851818-851818 on Build GRCh38
rs765265461 Chr.4: 851991-851991 on Build GRCh38
rs778123031 Chr.4: 851937-851937 on Build GRCh38
rs925425311 Chr.4: 852030-852030 on Build GRCh38
rs749711609 Chr.4: 852015-852015 on Build GRCh38
rs17165087 Chr.4: 852053-852053 on Build GRCh38
rs150254936 Chr.4: 851860-851860 on Build GRCh38
rs750349401 Chr.4: 851992-851992 on Build GRCh38
rs895674047 Chr.4: 852041-852041 on Build GRCh38
rs904036098 Chr.4: 851683-851683 on Build GRCh38
rs780799062 Chr.4: 851739-851739 on Build GRCh38
rs1043837313 Chr.4: 851864-851864 on Build GRCh38
rs1044251526 Chr.4: 851680-851680 on Build GRCh38
rs996562293 Chr.4: 852072-852072 on Build GRCh38
rs746370239 Chr.4: 852026-852026 on Build GRCh38
rs763401200 Chr.4: 851838-851838 on Build GRCh38
rs767829625 Chr.4: 851886-851886 on Build GRCh38
rs371404744 Chr.4: 851923-851923 on Build GRCh38
rs552544001 Chr.4: 852038-852038 on Build GRCh38
rs754693522 Chr.4: 852006-852006 on Build GRCh38
rs371554961 Chr.4: 852078-852078 on Build GRCh38
rs955227900 Chr.4: 852084-852084 on Build GRCh38
rs911502231 Chr.4: 851908-851908 on Build GRCh38
rs377323506 Chr.4: 852023-852023 on Build GRCh38
rs746694666 Chr.4: 851931-851931 on Build GRCh38
rs758673275 Chr.4: 851732-851732 on Build GRCh38
rs761107041 Chr.4: 851840-851841 on Build GRCh38
rs776792019 Chr.4: 851769-851769 on Build GRCh38
rs921212908 Chr.4: 851825-851825 on Build GRCh38
rs765365719 Chr.4: 852025-852025 on Build GRCh38
rs777242797 Chr.4: 851981-851981 on Build GRCh38
rs774558099 Chr.4: 851874-851874 on Build GRCh38
rs10031922 Chr.4: 851984-851984 on Build GRCh38
rs145818077 Chr.4: 851890-851890 on Build GRCh38
rs770560926 Chr.4: 851739-851739 on Build GRCh38
rs781056821 Chr.4: 851783-851783 on Build GRCh38
rs976924155 Chr.4: 852088-852088 on Build GRCh38
rs780314837 Chr.4: 851735-851735 on Build GRCh38
rs1003915327 Chr.4: 852059-852059 on Build GRCh38
rs774422172 Chr.4: 852042-852042 on Build GRCh38
rs56169884 Chr.4: 851848-851848 on Build GRCh38
rs777823118 Chr.4: 851789-851789 on Build GRCh38
rs751964862 Chr.4: 852069-852069 on Build GRCh38
rs986694971 Chr.4: 852009-852009 on Build GRCh38
rs774213026 Chr.4: 851767-851768 on Build GRCh38
rs368101195 Chr.4: 852000-852000 on Build GRCh38
rs55697914 Chr.4: 851952-851952 on Build GRCh38
rs774036539 Chr.4: 851764-851764 on Build GRCh38
rs1016626619 Chr.4: 851837-851837 on Build GRCh38
rs138890416 Chr.4: 851932-851932 on Build GRCh38
rs557028978 Chr.4: 852071-852071 on Build GRCh38
rs141977918 Chr.4: 851770-851770 on Build GRCh38
rs754778881 Chr.4: 851737-851737 on Build GRCh38
rs968369424 Chr.4: 852058-852058 on Build GRCh38
rs754394917 Chr.4: 851709-851709 on Build GRCh38
rs771564068 Chr.4: 851806-851806 on Build GRCh38
rs1002845993 Chr.4: 851714-851714 on Build GRCh38
rs752994019 Chr.4: 851887-851887 on Build GRCh38
rs564456843 Chr.4: 851948-851948 on Build GRCh38
rs142966472 Chr.4: 851780-851780 on Build GRCh38
rs888435249 Chr.4: 851999-851999 on Build GRCh38
rs756188963 Chr.4: 851785-851785 on Build GRCh38
rs539110746 Chr.4: 851754-851754 on Build GRCh38
rs757723323 Chr.4: 851711-851711 on Build GRCh38
rs770322584 Chr.4: 851827-851827 on Build GRCh38
rs955258979 Chr.4: 852001-852001 on Build GRCh38
rs748560870 Chr.4: 851863-851863 on Build GRCh38
rs201573243 Chr.4: 851985-851985 on Build GRCh38
rs771820165 Chr.4: 851862-851862 on Build GRCh38
rs377418158 Chr.4: 851805-851805 on Build GRCh38
rs777108426 Chr.4: 851809-851809 on Build GRCh38
rs763988520 Chr.4: 851891-851891 on Build GRCh38
rs770499898 Chr.4: 851797-851797 on Build GRCh38
rs964658199 Chr.4: 852082-852082 on Build GRCh38
rs750441811 Chr.4: 851726-851726 on Build GRCh38
rs17165089 Chr.4: 852061-852061 on Build GRCh38
rs576067675 Chr.4: 851762-851762 on Build GRCh38
rs755951124 Chr.4: 851847-851847 on Build GRCh38
rs886759211 Chr.4: 852057-852057 on Build GRCh38
rs555528243 Chr.4: 852083-852083 on Build GRCh38
rs367689402 Chr.4: 851720-851720 on Build GRCh38
rs1055264502 Chr.4: 852039-852039 on Build GRCh38
rs759504391 Chr.4: 851884-851884 on Build GRCh38
rs745811697 Chr.4: 851801-851801 on Build GRCh38
rs765716559 Chr.4: 851773-851773 on Build GRCh38
rs775648883 Chr.4: 851962-851962 on Build GRCh38
rs928410704 Chr.4: 851708-851708 on Build GRCh38
rs145713462 Chr.4: 851920-851920 on Build GRCh38
rs55801437 Chr.4: 851898-851898 on Build GRCh38
rs147555555 Chr.4: 851757-851757 on Build GRCh38
rs372104503 Chr.4: 851868-851868 on Build GRCh38
rs762158341 Chr.4: 851815-851815 on Build GRCh38
rs771331071 Chr.4: 851873-851873 on Build GRCh38
rs763518838 Chr.4: 851872-851872 on Build GRCh38
rs554365004 Chr.4: 852104-852104 on Build GRCh38
rs772313362 Chr.4: 851961-851961 on Build GRCh38
rs764401284 Chr.4: 851982-851982 on Build GRCh38
rs986663960 Chr.4: 852089-852089 on Build GRCh38
rs374621415 Chr.4: 851851-851851 on Build GRCh38
rs753631196 Chr.4: 851893-851893 on Build GRCh38
rs747944674 Chr.4: 851784-851784 on Build GRCh38
rs757585286 Chr.4: 852019-852019 on Build GRCh38
rs572653435 Chr.4: 852105-852105 on Build GRCh38
rs778502212 Chr.4: 851854-851854 on Build GRCh38
rs759064696 Chr.4: 851765-851765 on Build GRCh38
rs760994565 Chr.4: 851971-851971 on Build GRCh38
rs1037624284 Chr.4: 852018-852018 on Build GRCh38
rs766558665 Chr.4: 851778-851778 on Build GRCh38

View Full Product Details