Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 1723439-1723956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TACC3
Entrez Gene ID
10460
Gene Name
transforming acidic coiled-coil containing protein 3
Gene Aliases
ERIC-1, ERIC1
UniGene
Hs.104019
Gene Chromosome Location
Chr.4: 1721383-1745178 on Build GRCh38
Transcripts
NM_006342.2 XM_005247929.2 XM_005247930.2 XM_011513386.2 XM_017007653.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6796086 c.117A>G p.S39S Chr.4: 1723538-1723538 on Build GRCh38
4584922 c.287T>G p.V96G Chr.4: 1723852-1723852 on Build GRCh38
4435774 c.87T>G p.V29V Chr.4: 1723508-1723508 on Build GRCh38
1670847 c.250C>T p.Q84* Chr.4: 1723815-1723815 on Build GRCh38
1053217 c.74C>T p.S25L Chr.4: 1723495-1723495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3099555 Chr.4: 1723924-1723924 on Build GRCh38
rs751907182 Chr.4: 1723540-1723540 on Build GRCh38
rs545133849 Chr.4: 1723699-1723699 on Build GRCh38
rs750852524 Chr.4: 1723475-1723475 on Build GRCh38
rs771913700 Chr.4: 1723723-1723723 on Build GRCh38
rs751472230 Chr.4: 1723876-1723876 on Build GRCh38
rs767277016 Chr.4: 1723888-1723888 on Build GRCh38
rs762025550 Chr.4: 1723861-1723861 on Build GRCh38
rs112607903 Chr.4: 1723902-1723912 on Build GRCh38
rs746543897 Chr.4: 1723878-1723879 on Build GRCh38
rs1021398527 Chr.4: 1723490-1723490 on Build GRCh38
rs747489203 Chr.4: 1723921-1723921 on Build GRCh38
rs772723544 Chr.4: 1723747-1723747 on Build GRCh38
rs779924835 Chr.4: 1723709-1723709 on Build GRCh38
rs761443089 Chr.4: 1723656-1723656 on Build GRCh38
rs111586848 Chr.4: 1723907-1723907 on Build GRCh38
rs745629824 Chr.4: 1723612-1723612 on Build GRCh38
rs776870219 Chr.4: 1723860-1723860 on Build GRCh38
rs542100018 Chr.4: 1723463-1723463 on Build GRCh38
rs759940476 Chr.4: 1723544-1723544 on Build GRCh38
rs554173041 Chr.4: 1723850-1723850 on Build GRCh38
rs756373513 Chr.4: 1723578-1723578 on Build GRCh38
rs762871936 Chr.4: 1723538-1723538 on Build GRCh38
rs753842653 Chr.4: 1723694-1723694 on Build GRCh38
rs376371364 Chr.4: 1723698-1723698 on Build GRCh38
rs748197142 Chr.4: 1723496-1723496 on Build GRCh38
rs373800152 Chr.4: 1723530-1723530 on Build GRCh38
rs746597739 Chr.4: 1723618-1723618 on Build GRCh38
rs112671464 Chr.4: 1723477-1723477 on Build GRCh38
rs745729998 Chr.4: 1723675-1723683 on Build GRCh38
rs562391356 Chr.4: 1723506-1723506 on Build GRCh38
rs763301386 Chr.4: 1723535-1723535 on Build GRCh38
rs765740396 Chr.4: 1723474-1723474 on Build GRCh38
rs759970838 Chr.4: 1723749-1723749 on Build GRCh38
rs774800982 Chr.4: 1723670-1723670 on Build GRCh38
rs754928315 Chr.4: 1723918-1723918 on Build GRCh38
rs773412337 Chr.4: 1723867-1723867 on Build GRCh38
rs775823250 Chr.4: 1723758-1723758 on Build GRCh38
rs766481986 Chr.4: 1723813-1723813 on Build GRCh38
rs754020838 Chr.4: 1723588-1723588 on Build GRCh38
rs751957088 Chr.4: 1723487-1723487 on Build GRCh38
rs748859222 Chr.4: 1723848-1723848 on Build GRCh38
rs778311755 Chr.4: 1723919-1723919 on Build GRCh38
rs528006846 Chr.4: 1723511-1723511 on Build GRCh38
rs763459971 Chr.4: 1723531-1723531 on Build GRCh38
rs776102287 Chr.4: 1723909-1723910 on Build GRCh38
rs758404973 Chr.4: 1723804-1723804 on Build GRCh38
rs760309050 Chr.4: 1723464-1723464 on Build GRCh38
rs752501645 Chr.4: 1723897-1723897 on Build GRCh38
rs769012842 Chr.4: 1723859-1723859 on Build GRCh38
rs772633739 Chr.4: 1723750-1723750 on Build GRCh38
rs758420884 Chr.4: 1723707-1723707 on Build GRCh38
rs753064828 Chr.4: 1723677-1723677 on Build GRCh38
rs374704896 Chr.4: 1723928-1723928 on Build GRCh38
rs141706579 Chr.4: 1723819-1723819 on Build GRCh38
rs745719491 Chr.4: 1723516-1723516 on Build GRCh38
rs775050381 Chr.4: 1723803-1723804 on Build GRCh38
rs372966037 Chr.4: 1723873-1723873 on Build GRCh38
rs142473170 Chr.4: 1723520-1723520 on Build GRCh38
rs755187166 Chr.4: 1723489-1723489 on Build GRCh38
rs763174794 Chr.4: 1723868-1723868 on Build GRCh38
rs867940135 Chr.4: 1723647-1723647 on Build GRCh38
rs527928959 Chr.4: 1723593-1723594 on Build GRCh38
rs76574905 Chr.4: 1723926-1723926 on Build GRCh38
rs764438597 Chr.4: 1723685-1723685 on Build GRCh38
rs577000546 Chr.4: 1723824-1723824 on Build GRCh38
rs201821987 Chr.4: 1723608-1723608 on Build GRCh38
rs1020787438 Chr.4: 1723553-1723553 on Build GRCh38
rs768179397 Chr.4: 1723722-1723722 on Build GRCh38
rs200043853 Chr.4: 1723854-1723854 on Build GRCh38
rs1026228555 Chr.4: 1723644-1723644 on Build GRCh38
rs764314226 Chr.4: 1723481-1723481 on Build GRCh38
rs780872671 Chr.4: 1723836-1723836 on Build GRCh38
rs147088559 Chr.4: 1723843-1723843 on Build GRCh38
rs765617236 Chr.4: 1723862-1723862 on Build GRCh38
rs779163019 Chr.4: 1723704-1723704 on Build GRCh38
rs767813161 Chr.4: 1723672-1723672 on Build GRCh38
rs41286671 Chr.4: 1723763-1723763 on Build GRCh38
rs754886063 Chr.4: 1723885-1723885 on Build GRCh38
rs760807411 Chr.4: 1723678-1723678 on Build GRCh38
rs755768976 Chr.4: 1723841-1723841 on Build GRCh38
rs771949534 Chr.4: 1723615-1723615 on Build GRCh38
rs766595415 Chr.4: 1723482-1723482 on Build GRCh38
rs370071503 Chr.4: 1723839-1723839 on Build GRCh38
rs777648354 Chr.4: 1723580-1723580 on Build GRCh38
rs551856054 Chr.4: 1723638-1723638 on Build GRCh38
rs775999949 Chr.4: 1723635-1723635 on Build GRCh38
rs768358764 Chr.4: 1723627-1723627 on Build GRCh38
rs922607462 Chr.4: 1723793-1723793 on Build GRCh38
rs368314126 Chr.4: 1723705-1723705 on Build GRCh38
rs780013168 Chr.4: 1723617-1723617 on Build GRCh38
rs896432711 Chr.4: 1723913-1723913 on Build GRCh38
rs372508891 Chr.4: 1723720-1723720 on Build GRCh38
rs751594246 Chr.4: 1723903-1723903 on Build GRCh38
rs933226916 Chr.4: 1723798-1723798 on Build GRCh38
rs34708887 Chr.4: 1723762-1723762 on Build GRCh38
rs201394779 Chr.4: 1723724-1723724 on Build GRCh38
rs767901850 Chr.4: 1723545-1723545 on Build GRCh38
rs757131997 Chr.4: 1723600-1723600 on Build GRCh38
rs200472637 Chr.4: 1723573-1723573 on Build GRCh38
rs761960985 Chr.4: 1723773-1723773 on Build GRCh38
rs747504530 Chr.4: 1723858-1723858 on Build GRCh38
rs776214155 Chr.4: 1723533-1723533 on Build GRCh38
rs747578323 Chr.4: 1723733-1723733 on Build GRCh38
rs369375842 Chr.4: 1723908-1723908 on Build GRCh38
rs752868677 Chr.4: 1723551-1723551 on Build GRCh38
rs776875405 Chr.4: 1723765-1723765 on Build GRCh38
rs781449365 Chr.4: 1723495-1723495 on Build GRCh38
rs771318636 Chr.4: 1723667-1723667 on Build GRCh38
rs905523646 Chr.4: 1723519-1723519 on Build GRCh38
rs749122533 Chr.4: 1723512-1723512 on Build GRCh38
rs568321568 Chr.4: 1723788-1723788 on Build GRCh38
rs769300866 Chr.4: 1723742-1723742 on Build GRCh38
rs768335990 Chr.4: 1723880-1723881 on Build GRCh38

View Full Product Details