Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 73996334-73996840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL5
Entrez Gene ID
6374
Gene Name
C-X-C motif chemokine ligand 5
Gene Aliases
ENA-78, SCYB5
UniGene
Hs.89714
Gene Chromosome Location
Chr.4: 73995642-73998729 on Build GRCh38
Transcripts
NM_002994.4

SNP Detail

SNP ID Chromosome Location
rs910191869 Chr.4: 73996710-73996710 on Build GRCh38
rs79726157 Chr.4: 73996531-73996531 on Build GRCh38
rs958856126 Chr.4: 73996431-73996431 on Build GRCh38
rs536004740 Chr.4: 73996423-73996423 on Build GRCh38
rs200167638 Chr.4: 73996642-73996642 on Build GRCh38
rs994361430 Chr.4: 73996558-73996558 on Build GRCh38
rs200102061 Chr.4: 73996470-73996470 on Build GRCh38
rs1050140915 Chr.4: 73996493-73996493 on Build GRCh38
rs888737607 Chr.4: 73996575-73996575 on Build GRCh38
rs201135867 Chr.4: 73996592-73996592 on Build GRCh38
rs1013728659 Chr.4: 73996436-73996436 on Build GRCh38
rs1030589236 Chr.4: 73996424-73996424 on Build GRCh38
rs546610908 Chr.4: 73996369-73996369 on Build GRCh38
rs925894088 Chr.4: 73996481-73996481 on Build GRCh38
rs546005069 Chr.4: 73996795-73996795 on Build GRCh38
rs199505230 Chr.4: 73996722-73996722 on Build GRCh38
rs1025888698 Chr.4: 73996563-73996563 on Build GRCh38
rs201573085 Chr.4: 73996497-73996497 on Build GRCh38
rs920171004 Chr.4: 73996462-73996462 on Build GRCh38
rs202032683 Chr.4: 73996593-73996593 on Build GRCh38
rs1005961925 Chr.4: 73996584-73996584 on Build GRCh38
rs935210051 Chr.4: 73996581-73996581 on Build GRCh38
rs761587778 Chr.4: 73996594-73996594 on Build GRCh38
rs944156881 Chr.4: 73996499-73996499 on Build GRCh38
rs952672422 Chr.4: 73996379-73996379 on Build GRCh38
rs1052068532 Chr.4: 73996583-73996583 on Build GRCh38
rs199512183 Chr.4: 73996419-73996419 on Build GRCh38
rs986462277 Chr.4: 73996698-73996698 on Build GRCh38
rs575603094 Chr.4: 73996554-73996554 on Build GRCh38
rs931790466 Chr.4: 73996542-73996542 on Build GRCh38
rs960445906 Chr.4: 73996433-73996433 on Build GRCh38
rs200557338 Chr.4: 73996732-73996732 on Build GRCh38
rs1040164977 Chr.4: 73996503-73996503 on Build GRCh38
rs967163799 Chr.4: 73996637-73996637 on Build GRCh38
rs543053430 Chr.4: 73996560-73996560 on Build GRCh38
rs201695362 Chr.4: 73996459-73996459 on Build GRCh38
rs1040128856 Chr.4: 73996784-73996785 on Build GRCh38
rs768387482 Chr.4: 73996530-73996530 on Build GRCh38
rs188819105 Chr.4: 73996455-73996455 on Build GRCh38
rs200911081 Chr.4: 73996674-73996674 on Build GRCh38
rs572532692 Chr.4: 73996588-73996588 on Build GRCh38
rs200567202 Chr.4: 73996421-73996421 on Build GRCh38
rs200262009 Chr.4: 73996596-73996596 on Build GRCh38
rs201520116 Chr.4: 73996758-73996758 on Build GRCh38
rs993323893 Chr.4: 73996445-73996445 on Build GRCh38
rs139084547 Chr.4: 73996446-73996446 on Build GRCh38
rs996072954 Chr.4: 73996624-73996624 on Build GRCh38
rs202220641 Chr.4: 73996537-73996537 on Build GRCh38
rs903786347 Chr.4: 73996570-73996570 on Build GRCh38
rs200547811 Chr.4: 73996586-73996586 on Build GRCh38
rs352047 Chr.4: 73996373-73996373 on Build GRCh38
rs994393913 Chr.4: 73996376-73996376 on Build GRCh38
rs1043664906 Chr.4: 73996546-73996546 on Build GRCh38
rs201727036 Chr.4: 73996703-73996703 on Build GRCh38
rs201999508 Chr.4: 73996430-73996430 on Build GRCh38
rs199710276 Chr.4: 73996670-73996670 on Build GRCh38
rs951614929 Chr.4: 73996716-73996716 on Build GRCh38
rs935660348 Chr.4: 73996490-73996490 on Build GRCh38
rs200157678 Chr.4: 73996804-73996804 on Build GRCh38
rs201427693 Chr.4: 73996660-73996660 on Build GRCh38
rs199913779 Chr.4: 73996498-73996498 on Build GRCh38
rs896125095 Chr.4: 73996584-73996584 on Build GRCh38
rs200898994 Chr.4: 73996478-73996478 on Build GRCh38
rs201390855 Chr.4: 73996603-73996603 on Build GRCh38
rs763876705 Chr.4: 73996441-73996441 on Build GRCh38
rs146196659 Chr.4: 73996420-73996420 on Build GRCh38
rs957273790 Chr.4: 73996688-73996688 on Build GRCh38
rs183054356 Chr.4: 73996763-73996763 on Build GRCh38
rs1023785266 Chr.4: 73996611-73996611 on Build GRCh38
rs201283547 Chr.4: 73996380-73996380 on Build GRCh38
rs191166277 Chr.4: 73996621-73996621 on Build GRCh38

View Full Product Details