Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74312880-74313408 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPGN
Entrez Gene ID
255324
Gene Name
epithelial mitogen
Gene Aliases
ALGV3072, EPG, PRO9904
UniGene
Hs.401237
Gene Chromosome Location
Chr.4: 74307571-74317298 on Build GRCh38
Transcripts
NM_001270989.1 NM_001270990.1 NM_001270991.1 NM_001270992.1 NM_001270993.1 NR_073114.1 NR_073117.1 XM_005265666.4 XM_011531817.2

Gene Symbol
LOC105377276
Entrez Gene ID
105377276
Gene Name
uncharacterized LOC105377276
Gene Aliases
-
UniGene
Hs.662627
Gene Chromosome Location
Chr.4: 74278045-74330322 on Build GRCh38
Transcripts
NM_001270989.1 NM_001270990.1 NM_001270991.1 NM_001270992.1 NM_001270993.1 NR_073114.1 NR_073117.1 XM_005265666.4 XM_011531817.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6654870 c.387T>G p.F129L Chr.4: 74313150-74313150 on Build GRCh38
6654869 c.360T>G p.F120L Chr.4: 74313150-74313150 on Build GRCh38
6654872 c.273T>C p.T91T Chr.4: 74313036-74313036 on Build GRCh38
6999866 c.281G>A p.R94K Chr.4: 74313044-74313044 on Build GRCh38
6654871 c.246T>C p.T82T Chr.4: 74313036-74313036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372602605 Chr.4: 74313211-74313211 on Build GRCh38
rs199534769 Chr.4: 74313171-74313171 on Build GRCh38
rs770514022 Chr.4: 74313094-74313094 on Build GRCh38
rs754331060 Chr.4: 74313153-74313153 on Build GRCh38
rs762067449 Chr.4: 74313013-74313013 on Build GRCh38
rs774570877 Chr.4: 74313273-74313273 on Build GRCh38
rs771380797 Chr.4: 74313233-74313233 on Build GRCh38
rs200275702 Chr.4: 74313052-74313052 on Build GRCh38
rs770723786 Chr.4: 74312998-74312998 on Build GRCh38
rs972829704 Chr.4: 74313016-74313016 on Build GRCh38
rs781269217 Chr.4: 74313098-74313098 on Build GRCh38
rs988402550 Chr.4: 74313186-74313186 on Build GRCh38
rs757924190 Chr.4: 74313079-74313079 on Build GRCh38
rs749607941 Chr.4: 74312979-74312979 on Build GRCh38
rs747041721 Chr.4: 74313040-74313041 on Build GRCh38
rs368034390 Chr.4: 74313182-74313182 on Build GRCh38
rs371369261 Chr.4: 74312984-74312984 on Build GRCh38
rs1012744486 Chr.4: 74313368-74313368 on Build GRCh38
rs779309067 Chr.4: 74313072-74313072 on Build GRCh38
rs774697911 Chr.4: 74313012-74313012 on Build GRCh38
rs759372800 Chr.4: 74313002-74313002 on Build GRCh38
rs887002966 Chr.4: 74312981-74312981 on Build GRCh38
rs144389167 Chr.4: 74313160-74313160 on Build GRCh38
rs568497721 Chr.4: 74313007-74313007 on Build GRCh38
rs543681993 Chr.4: 74313232-74313232 on Build GRCh38
rs780786277 Chr.4: 74313180-74313180 on Build GRCh38
rs374840179 Chr.4: 74313085-74313085 on Build GRCh38
rs767306993 Chr.4: 74313240-74313240 on Build GRCh38
rs772140254 Chr.4: 74313125-74313125 on Build GRCh38
rs765708757 Chr.4: 74313224-74313224 on Build GRCh38
rs186759964 Chr.4: 74312980-74312980 on Build GRCh38
rs763240279 Chr.4: 74313212-74313212 on Build GRCh38
rs376864830 Chr.4: 74313230-74313230 on Build GRCh38
rs756863135 Chr.4: 74313238-74313238 on Build GRCh38
rs776830351 Chr.4: 74313170-74313170 on Build GRCh38
rs775356464 Chr.4: 74313104-74313104 on Build GRCh38
rs768879085 Chr.4: 74313143-74313143 on Build GRCh38
rs754386404 Chr.4: 74313054-74313054 on Build GRCh38
rs562291300 Chr.4: 74313265-74313265 on Build GRCh38
rs375409954 Chr.4: 74313174-74313174 on Build GRCh38
rs756508818 Chr.4: 74312969-74312969 on Build GRCh38
rs749900792 Chr.4: 74313243-74313243 on Build GRCh38
rs941091654 Chr.4: 74312929-74312929 on Build GRCh38
rs753076857 Chr.4: 74313076-74313076 on Build GRCh38
rs867720129 Chr.4: 74313328-74313328 on Build GRCh38
rs769812133 Chr.4: 74313188-74313188 on Build GRCh38
rs746534743 Chr.4: 74312994-74312994 on Build GRCh38
rs776627880 Chr.4: 74313215-74313215 on Build GRCh38
rs776757981 Chr.4: 74312999-74312999 on Build GRCh38
rs766515617 Chr.4: 74313140-74313140 on Build GRCh38
rs151189030 Chr.4: 74313162-74313162 on Build GRCh38
rs760644948 Chr.4: 74313132-74313132 on Build GRCh38
rs773306349 Chr.4: 74313209-74313209 on Build GRCh38
rs777502112 Chr.4: 74313087-74313087 on Build GRCh38
rs372816591 Chr.4: 74313107-74313107 on Build GRCh38
rs760833229 Chr.4: 74313035-74313035 on Build GRCh38
rs369806495 Chr.4: 74313092-74313092 on Build GRCh38
rs773534212 Chr.4: 74313129-74313129 on Build GRCh38
rs201853400 Chr.4: 74313065-74313065 on Build GRCh38
rs751880271 Chr.4: 74313234-74313234 on Build GRCh38
rs560687185 Chr.4: 74313347-74313347 on Build GRCh38
rs780212511 Chr.4: 74312999-74312999 on Build GRCh38
rs772540254 Chr.4: 74312989-74312989 on Build GRCh38
rs140226178 Chr.4: 74312950-74312950 on Build GRCh38
rs762835135 Chr.4: 74313110-74313110 on Build GRCh38
rs535933465 Chr.4: 74313018-74313018 on Build GRCh38
rs758845621 Chr.4: 74313172-74313172 on Build GRCh38
rs760906121 Chr.4: 74313138-74313138 on Build GRCh38
rs539372623 Chr.4: 74313083-74313083 on Build GRCh38
rs756704757 Chr.4: 74313175-74313175 on Build GRCh38
rs541076295 Chr.4: 74313346-74313346 on Build GRCh38
rs749017149 Chr.4: 74313204-74313204 on Build GRCh38
rs759979106 Chr.4: 74313154-74313154 on Build GRCh38
rs898452281 Chr.4: 74313360-74313360 on Build GRCh38
rs573701936 Chr.4: 74313320-74313320 on Build GRCh38
rs780166560 Chr.4: 74313189-74313189 on Build GRCh38
rs998108251 Chr.4: 74312938-74312938 on Build GRCh38
rs776103586 Chr.4: 74313227-74313227 on Build GRCh38
rs758446226 Chr.4: 74312998-74312999 on Build GRCh38
rs768540442 Chr.4: 74313205-74313205 on Build GRCh38
rs142205602 Chr.4: 74313101-74313101 on Build GRCh38
rs1034938814 Chr.4: 74313340-74313340 on Build GRCh38
rs759644172 Chr.4: 74313223-74313223 on Build GRCh38
rs780645867 Chr.4: 74312971-74312971 on Build GRCh38

View Full Product Details