Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 80286583-80287125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF5
Entrez Gene ID
2250
Gene Name
fibroblast growth factor 5
Gene Aliases
HBGF-5, Smag-82, TCMGLY
UniGene
Hs.37055
Gene Chromosome Location
Chr.4: 80266588-80291017 on Build GRCh38
Transcripts
NM_001291812.1 NM_004464.3 NM_033143.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1058062 c.763A>C p.N255H Chr.4: 80286628-80286628 on Build GRCh38
3606414 c.796C>T p.R266C Chr.4: 80286661-80286661 on Build GRCh38
1058063 c.785G>A p.R262K Chr.4: 80286650-80286650 on Build GRCh38
4758120 c.791A>C p.K264T Chr.4: 80286656-80286656 on Build GRCh38
5531251 c.803G>A p.G268E Chr.4: 80286668-80286668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766466566 Chr.4: 80286658-80286658 on Build GRCh38
rs890417080 Chr.4: 80286680-80286680 on Build GRCh38
rs773359249 Chr.4: 80286653-80286653 on Build GRCh38
rs570969510 Chr.4: 80287014-80287014 on Build GRCh38
rs758061656 Chr.4: 80286630-80286631 on Build GRCh38
rs181147947 Chr.4: 80286763-80286763 on Build GRCh38
rs35998213 Chr.4: 80286869-80286870 on Build GRCh38
rs771106077 Chr.4: 80287076-80287076 on Build GRCh38
rs1039349957 Chr.4: 80287082-80287082 on Build GRCh38
rs34334216 Chr.4: 80286706-80286706 on Build GRCh38
rs973995380 Chr.4: 80286912-80286912 on Build GRCh38
rs919724099 Chr.4: 80286973-80286973 on Build GRCh38
rs563860210 Chr.4: 80286759-80286759 on Build GRCh38
rs772667458 Chr.4: 80286711-80286711 on Build GRCh38
rs563131467 Chr.4: 80286796-80286796 on Build GRCh38
rs748527362 Chr.4: 80286702-80286702 on Build GRCh38
rs527709482 Chr.4: 80286860-80286860 on Build GRCh38
rs1031236697 Chr.4: 80286894-80286894 on Build GRCh38
rs3733336 Chr.4: 80286809-80286809 on Build GRCh38
rs960691203 Chr.4: 80286899-80286899 on Build GRCh38
rs769031234 Chr.4: 80286638-80286638 on Build GRCh38
rs745555063 Chr.4: 80286624-80286624 on Build GRCh38
rs777203075 Chr.4: 80286607-80286607 on Build GRCh38
rs1021434528 Chr.4: 80286778-80286778 on Build GRCh38
rs943946339 Chr.4: 80287065-80287065 on Build GRCh38
rs878967686 Chr.4: 80287081-80287081 on Build GRCh38
rs35279668 Chr.4: 80287081-80287082 on Build GRCh38
rs145215382 Chr.4: 80286832-80286832 on Build GRCh38
rs769867484 Chr.4: 80286921-80286921 on Build GRCh38
rs905786888 Chr.4: 80286799-80286799 on Build GRCh38
rs755889709 Chr.4: 80286688-80286688 on Build GRCh38
rs935457832 Chr.4: 80286713-80286713 on Build GRCh38
rs1001416696 Chr.4: 80286803-80286803 on Build GRCh38
rs974200806 Chr.4: 80286962-80286962 on Build GRCh38
rs565932196 Chr.4: 80286852-80286852 on Build GRCh38
rs963138937 Chr.4: 80286891-80286891 on Build GRCh38
rs893899582 Chr.4: 80286994-80286994 on Build GRCh38
rs752500500 Chr.4: 80286670-80286670 on Build GRCh38
rs33946030 Chr.4: 80287090-80287090 on Build GRCh38
rs199904159 Chr.4: 80286623-80286623 on Build GRCh38
rs905654120 Chr.4: 80287012-80287012 on Build GRCh38
rs768971081 Chr.4: 80286686-80286686 on Build GRCh38
rs773795381 Chr.4: 80286719-80286719 on Build GRCh38
rs895508517 Chr.4: 80287056-80287056 on Build GRCh38
rs1043288575 Chr.4: 80286999-80286999 on Build GRCh38
rs929853642 Chr.4: 80286975-80286975 on Build GRCh38
rs1022862417 Chr.4: 80286840-80286840 on Build GRCh38
rs371866408 Chr.4: 80286678-80286678 on Build GRCh38
rs968259220 Chr.4: 80286861-80286861 on Build GRCh38
rs762398537 Chr.4: 80286642-80286642 on Build GRCh38
rs375188495 Chr.4: 80286683-80286683 on Build GRCh38
rs1035356055 Chr.4: 80286807-80286807 on Build GRCh38
rs763074155 Chr.4: 80286995-80286995 on Build GRCh38
rs922091611 Chr.4: 80286914-80286914 on Build GRCh38
rs1052553104 Chr.4: 80286972-80286972 on Build GRCh38
rs942182569 Chr.4: 80286631-80286631 on Build GRCh38
rs34581362 Chr.4: 80286880-80286881 on Build GRCh38
rs891655241 Chr.4: 80286743-80286743 on Build GRCh38
rs765000741 Chr.4: 80286665-80286665 on Build GRCh38
rs932240570 Chr.4: 80286915-80286915 on Build GRCh38
rs999720997 Chr.4: 80286862-80286862 on Build GRCh38
rs770450045 Chr.4: 80286622-80286622 on Build GRCh38
rs746528773 Chr.4: 80286614-80286614 on Build GRCh38
rs542157486 Chr.4: 80286655-80286655 on Build GRCh38
rs764046663 Chr.4: 80286682-80286682 on Build GRCh38
rs551718154 Chr.4: 80286936-80286936 on Build GRCh38
rs774924885 Chr.4: 80286641-80286641 on Build GRCh38
rs564711050 Chr.4: 80286795-80286795 on Build GRCh38
rs760770821 Chr.4: 80286737-80286737 on Build GRCh38
rs781680571 Chr.4: 80286779-80286779 on Build GRCh38
rs759720974 Chr.4: 80286664-80286664 on Build GRCh38
rs932207982 Chr.4: 80286669-80286669 on Build GRCh38
rs756769098 Chr.4: 80286685-80286685 on Build GRCh38
rs773346447 Chr.4: 80286949-80286949 on Build GRCh38
rs983231020 Chr.4: 80286826-80286826 on Build GRCh38
rs368904140 Chr.4: 80286652-80286652 on Build GRCh38
rs115729960 Chr.4: 80286948-80286948 on Build GRCh38
rs137903755 Chr.4: 80286661-80286661 on Build GRCh38
rs1047012959 Chr.4: 80287016-80287016 on Build GRCh38
rs1035595279 Chr.4: 80287036-80287036 on Build GRCh38
rs1052463259 Chr.4: 80286740-80286740 on Build GRCh38
rs539936574 Chr.4: 80286784-80286784 on Build GRCh38
rs201516192 Chr.4: 80286690-80286690 on Build GRCh38
rs916593153 Chr.4: 80286942-80286942 on Build GRCh38

View Full Product Details