Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 81427358-81427866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGEF1B
Entrez Gene ID
153020
Gene Name
RasGEF domain family member 1B
Gene Aliases
GPIG4
UniGene
Hs.591696
Gene Chromosome Location
Chr.4: 81426393-81471928 on Build GRCh38
Transcripts
NM_001300735.1 NM_001300736.1 NM_152545.2 XM_017007812.1 XM_017007813.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2955423 c.1403G>A p.S468N Chr.4: 81427787-81427787 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772014603 Chr.4: 81427645-81427645 on Build GRCh38
rs760432663 Chr.4: 81427819-81427819 on Build GRCh38
rs566305305 Chr.4: 81427620-81427620 on Build GRCh38
rs113748036 Chr.4: 81427676-81427676 on Build GRCh38
rs780029233 Chr.4: 81427423-81427423 on Build GRCh38
rs948283668 Chr.4: 81427681-81427681 on Build GRCh38
rs1004621879 Chr.4: 81427783-81427783 on Build GRCh38
rs770891934 Chr.4: 81427826-81427826 on Build GRCh38
rs569310155 Chr.4: 81427543-81427543 on Build GRCh38
rs550359161 Chr.4: 81427477-81427477 on Build GRCh38
rs979138175 Chr.4: 81427688-81427688 on Build GRCh38
rs887816536 Chr.4: 81427595-81427595 on Build GRCh38
rs959055161 Chr.4: 81427693-81427693 on Build GRCh38
rs779218430 Chr.4: 81427801-81427801 on Build GRCh38
rs781637140 Chr.4: 81427782-81427782 on Build GRCh38
rs979511790 Chr.4: 81427455-81427455 on Build GRCh38
rs764867137 Chr.4: 81427760-81427760 on Build GRCh38
rs537455283 Chr.4: 81427706-81427706 on Build GRCh38
rs373018664 Chr.4: 81427426-81427426 on Build GRCh38
rs142042020 Chr.4: 81427682-81427682 on Build GRCh38
rs768167014 Chr.4: 81427745-81427746 on Build GRCh38
rs369254693 Chr.4: 81427751-81427751 on Build GRCh38
rs568931517 Chr.4: 81427576-81427579 on Build GRCh38
rs937914852 Chr.4: 81427491-81427491 on Build GRCh38
rs949271273 Chr.4: 81427512-81427512 on Build GRCh38
rs560989387 Chr.4: 81427534-81427534 on Build GRCh38
rs924908376 Chr.4: 81427435-81427435 on Build GRCh38
rs1007509638 Chr.4: 81427625-81427625 on Build GRCh38
rs775241866 Chr.4: 81427725-81427725 on Build GRCh38
rs748402557 Chr.4: 81427802-81427802 on Build GRCh38
rs771719655 Chr.4: 81427808-81427808 on Build GRCh38
rs1021512345 Chr.4: 81427655-81427655 on Build GRCh38
rs965925747 Chr.4: 81427381-81427382 on Build GRCh38
rs766263792 Chr.4: 81427731-81427731 on Build GRCh38
rs931489293 Chr.4: 81427708-81427708 on Build GRCh38
rs768858513 Chr.4: 81427796-81427796 on Build GRCh38
rs780475956 Chr.4: 81427791-81427791 on Build GRCh38
rs1055080453 Chr.4: 81427495-81427495 on Build GRCh38
rs1027613618 Chr.4: 81427575-81427575 on Build GRCh38
rs754544083 Chr.4: 81427759-81427759 on Build GRCh38
rs149035093 Chr.4: 81427628-81427628 on Build GRCh38
rs760542346 Chr.4: 81427594-81427594 on Build GRCh38
rs993506533 Chr.4: 81427550-81427550 on Build GRCh38
rs1017887168 Chr.4: 81427626-81427626 on Build GRCh38
rs757563649 Chr.4: 81427778-81427778 on Build GRCh38
rs375107913 Chr.4: 81427689-81427689 on Build GRCh38
rs530538409 Chr.4: 81427432-81427432 on Build GRCh38
rs1037247960 Chr.4: 81427529-81427529 on Build GRCh38
rs749372499 Chr.4: 81427795-81427795 on Build GRCh38
rs760695573 Chr.4: 81427724-81427724 on Build GRCh38
rs201095873 Chr.4: 81427426-81427427 on Build GRCh38
rs368896828 Chr.4: 81427511-81427511 on Build GRCh38
rs913678033 Chr.4: 81427420-81427420 on Build GRCh38
rs990466011 Chr.4: 81427696-81427696 on Build GRCh38
rs549582532 Chr.4: 81427560-81427560 on Build GRCh38
rs917859353 Chr.4: 81427714-81427714 on Build GRCh38
rs918375963 Chr.4: 81427789-81427789 on Build GRCh38
rs772970908 Chr.4: 81427720-81427720 on Build GRCh38
rs1028743722 Chr.4: 81427428-81427428 on Build GRCh38
rs752355463 Chr.4: 81427764-81427764 on Build GRCh38
rs949298763 Chr.4: 81427717-81427717 on Build GRCh38
rs746718881 Chr.4: 81427732-81427733 on Build GRCh38
rs750854940 Chr.4: 81427790-81427790 on Build GRCh38
rs773026523 Chr.4: 81427816-81427816 on Build GRCh38
rs779428932 Chr.4: 81427733-81427735 on Build GRCh38
rs529474201 Chr.4: 81427546-81427546 on Build GRCh38
rs761632206 Chr.4: 81427718-81427718 on Build GRCh38

View Full Product Details