Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 169684104-169684607 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOCK2
Entrez Gene ID
1794
Gene Name
dedicator of cytokinesis 2
Gene Aliases
IMD40
UniGene
Hs.586174
Gene Chromosome Location
Chr.5: 169637247-170083382 on Build GRCh38
Transcripts
NM_004946.2 XM_005265830.3 XM_011534448.2 XM_011534449.2 XM_011534450.2 XM_011534451.2 XM_017009189.1 XM_017009190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3919660 c.736G>A p.D246N Chr.5: 169684325-169684325 on Build GRCh38
336821 c.711T>A p.D237E Chr.5: 169684300-169684300 on Build GRCh38
6308813 c.621A>T p.P207P Chr.5: 169684210-169684210 on Build GRCh38
1672114 c.662A>G p.H221R Chr.5: 169684251-169684251 on Build GRCh38
6102960 c.653C>A p.S218Y Chr.5: 169684242-169684242 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014003740 Chr.5: 169684132-169684132 on Build GRCh38
rs77255427 Chr.5: 169684458-169684458 on Build GRCh38
rs777237537 Chr.5: 169684244-169684244 on Build GRCh38
rs140430355 Chr.5: 169684343-169684343 on Build GRCh38
rs1005183728 Chr.5: 169684549-169684549 on Build GRCh38
rs370094182 Chr.5: 169684256-169684256 on Build GRCh38
rs775347847 Chr.5: 169684270-169684270 on Build GRCh38
rs866182849 Chr.5: 169684242-169684242 on Build GRCh38
rs749352590 Chr.5: 169684347-169684347 on Build GRCh38
rs746960484 Chr.5: 169684311-169684311 on Build GRCh38
rs761811770 Chr.5: 169684145-169684145 on Build GRCh38
rs930717846 Chr.5: 169684438-169684438 on Build GRCh38
rs78243868 Chr.5: 169684341-169684341 on Build GRCh38
rs778499133 Chr.5: 169684252-169684252 on Build GRCh38
rs750077012 Chr.5: 169684386-169684386 on Build GRCh38
rs150969388 Chr.5: 169684229-169684229 on Build GRCh38
rs759880415 Chr.5: 169684334-169684334 on Build GRCh38
rs1020729517 Chr.5: 169684584-169684584 on Build GRCh38
rs572693616 Chr.5: 169684565-169684565 on Build GRCh38
rs139449745 Chr.5: 169684325-169684325 on Build GRCh38
rs569363874 Chr.5: 169684492-169684492 on Build GRCh38
rs771128747 Chr.5: 169684357-169684357 on Build GRCh38
rs756927816 Chr.5: 169684249-169684249 on Build GRCh38
rs900783634 Chr.5: 169684519-169684519 on Build GRCh38
rs535811475 Chr.5: 169684370-169684370 on Build GRCh38
rs151191554 Chr.5: 169684220-169684220 on Build GRCh38
rs760772001 Chr.5: 169684460-169684460 on Build GRCh38
rs1052973009 Chr.5: 169684511-169684511 on Build GRCh38
rs770251542 Chr.5: 169684319-169684319 on Build GRCh38
rs755691445 Chr.5: 169684239-169684239 on Build GRCh38
rs961641017 Chr.5: 169684566-169684566 on Build GRCh38
rs970140970 Chr.5: 169684533-169684533 on Build GRCh38
rs1014036549 Chr.5: 169684518-169684518 on Build GRCh38
rs765394561 Chr.5: 169684167-169684167 on Build GRCh38
rs1022689402 Chr.5: 169684533-169684534 on Build GRCh38
rs147838293 Chr.5: 169684134-169684145 on Build GRCh38
rs756691245 Chr.5: 169684376-169684376 on Build GRCh38
rs778444590 Chr.5: 169684377-169684377 on Build GRCh38
rs148418103 Chr.5: 169684214-169684214 on Build GRCh38
rs559096270 Chr.5: 169684562-169684562 on Build GRCh38
rs773634952 Chr.5: 169684168-169684168 on Build GRCh38
rs34039776 Chr.5: 169684246-169684246 on Build GRCh38
rs984797885 Chr.5: 169684230-169684230 on Build GRCh38
rs946712970 Chr.5: 169684436-169684436 on Build GRCh38
rs751888622 Chr.5: 169684204-169684204 on Build GRCh38
rs770032917 Chr.5: 169684321-169684321 on Build GRCh38
rs1033150545 Chr.5: 169684556-169684556 on Build GRCh38
rs774562096 Chr.5: 169684327-169684327 on Build GRCh38
rs1008929032 Chr.5: 169684479-169684479 on Build GRCh38
rs776890348 Chr.5: 169684530-169684530 on Build GRCh38
rs568572731 Chr.5: 169684237-169684237 on Build GRCh38
rs911234327 Chr.5: 169684360-169684360 on Build GRCh38
rs763235059 Chr.5: 169684181-169684181 on Build GRCh38
rs374550621 Chr.5: 169684138-169684149 on Build GRCh38
rs116358843 Chr.5: 169684577-169684577 on Build GRCh38
rs766896264 Chr.5: 169684203-169684203 on Build GRCh38
rs755325682 Chr.5: 169684213-169684213 on Build GRCh38
rs568199496 Chr.5: 169684405-169684405 on Build GRCh38
rs771842589 Chr.5: 169684268-169684268 on Build GRCh38
rs146746458 Chr.5: 169684342-169684342 on Build GRCh38
rs374307327 Chr.5: 169684324-169684324 on Build GRCh38
rs538867430 Chr.5: 169684517-169684517 on Build GRCh38

View Full Product Details