Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175442563-175443057 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD1
Entrez Gene ID
1812
Gene Name
dopamine receptor D1
Gene Aliases
DADR, DRD1A
UniGene
Hs.2624
Gene Chromosome Location
Chr.5: 175440672-175444160 on Build GRCh38
Transcripts
NM_000794.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5612824 c.126G>A p.T42T Chr.5: 175442974-175442974 on Build GRCh38
4472246 c.176C>T p.T59I Chr.5: 175442924-175442924 on Build GRCh38
4488234 c.329C>T p.S110F Chr.5: 175442771-175442771 on Build GRCh38
6850112 c.320C>T p.S107F Chr.5: 175442780-175442780 on Build GRCh38
1436095 c.79G>A p.A27T Chr.5: 175443021-175443021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761736618 Chr.5: 175442691-175442691 on Build GRCh38
rs771762936 Chr.5: 175442812-175442812 on Build GRCh38
rs752886523 Chr.5: 175442722-175442722 on Build GRCh38
rs765151364 Chr.5: 175442698-175442698 on Build GRCh38
rs372040450 Chr.5: 175442989-175442989 on Build GRCh38
rs369288437 Chr.5: 175442971-175442971 on Build GRCh38
rs767856135 Chr.5: 175442771-175442771 on Build GRCh38
rs572625837 Chr.5: 175443032-175443032 on Build GRCh38
rs764182655 Chr.5: 175442687-175442687 on Build GRCh38
rs779186397 Chr.5: 175442951-175442951 on Build GRCh38
rs905587833 Chr.5: 175442841-175442841 on Build GRCh38
rs775742033 Chr.5: 175443020-175443020 on Build GRCh38
rs191953032 Chr.5: 175442746-175442746 on Build GRCh38
rs774846844 Chr.5: 175442813-175442813 on Build GRCh38
rs745456137 Chr.5: 175442800-175442800 on Build GRCh38
rs555464688 Chr.5: 175442916-175442916 on Build GRCh38
rs776666364 Chr.5: 175442863-175442863 on Build GRCh38
rs755241449 Chr.5: 175442712-175442712 on Build GRCh38
rs753660657 Chr.5: 175442690-175442690 on Build GRCh38
rs746313806 Chr.5: 175442958-175442958 on Build GRCh38
rs760278176 Chr.5: 175442818-175442818 on Build GRCh38
rs771244904 Chr.5: 175442758-175442758 on Build GRCh38
rs777853990 Chr.5: 175442975-175442975 on Build GRCh38
rs756568052 Chr.5: 175442745-175442745 on Build GRCh38
rs149673312 Chr.5: 175442719-175442719 on Build GRCh38
rs141052935 Chr.5: 175442905-175442905 on Build GRCh38
rs749622213 Chr.5: 175442747-175442747 on Build GRCh38
rs116072750 Chr.5: 175442668-175442668 on Build GRCh38
rs867085762 Chr.5: 175442865-175442865 on Build GRCh38
rs5327 Chr.5: 175442991-175442991 on Build GRCh38
rs5329 Chr.5: 175442959-175442959 on Build GRCh38
rs759268810 Chr.5: 175442658-175442658 on Build GRCh38
rs752698175 Chr.5: 175442911-175442911 on Build GRCh38
rs768227443 Chr.5: 175442974-175442974 on Build GRCh38
rs768680344 Chr.5: 175442857-175442857 on Build GRCh38
rs756259088 Chr.5: 175442923-175442923 on Build GRCh38
rs757759969 Chr.5: 175443033-175443033 on Build GRCh38
rs751782298 Chr.5: 175442711-175442711 on Build GRCh38
rs767459799 Chr.5: 175443007-175443007 on Build GRCh38
rs867324709 Chr.5: 175442761-175442761 on Build GRCh38
rs759671921 Chr.5: 175442885-175442885 on Build GRCh38
rs374437918 Chr.5: 175442683-175442683 on Build GRCh38
rs769918422 Chr.5: 175442986-175442986 on Build GRCh38
rs372262906 Chr.5: 175442679-175442679 on Build GRCh38
rs764190400 Chr.5: 175442934-175442934 on Build GRCh38
rs549712601 Chr.5: 175442849-175442849 on Build GRCh38
rs1022706532 Chr.5: 175442913-175442913 on Build GRCh38
rs554208539 Chr.5: 175442973-175442973 on Build GRCh38
rs879844008 Chr.5: 175442990-175442990 on Build GRCh38
rs368319023 Chr.5: 175442661-175442661 on Build GRCh38
rs1042770 Chr.5: 175442878-175442878 on Build GRCh38
rs774032796 Chr.5: 175442644-175442644 on Build GRCh38
rs5330 Chr.5: 175442950-175442950 on Build GRCh38
rs1799914 Chr.5: 175442902-175442902 on Build GRCh38
rs1042768 Chr.5: 175443010-175443010 on Build GRCh38
rs767604497 Chr.5: 175443013-175443013 on Build GRCh38
rs779276408 Chr.5: 175442779-175442779 on Build GRCh38
rs748030620 Chr.5: 175442980-175442980 on Build GRCh38
rs367761486 Chr.5: 175442877-175442877 on Build GRCh38
rs754298471 Chr.5: 175442944-175442944 on Build GRCh38
rs749712205 Chr.5: 175442608-175442608 on Build GRCh38
rs775081512 Chr.5: 175442667-175442667 on Build GRCh38
rs770549660 Chr.5: 175442641-175442641 on Build GRCh38

View Full Product Details