Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 175521788-175522292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFXN1
Entrez Gene ID
94081
Gene Name
sideroflexin 1
Gene Aliases
TCC
UniGene
Hs.369440
Gene Chromosome Location
Chr.5: 175478498-175529742 on Build GRCh38
Transcripts
NM_001322977.1 NM_001322978.1 NM_001322980.1 NM_001322981.1 NM_001322982.1 NM_022754.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4585709 c.824+2T>C p.? Chr.5: 175521970-175521970 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs115077320 Chr.5: 175521832-175521832 on Build GRCh38
rs758813546 Chr.5: 175521991-175521991 on Build GRCh38
rs778079534 Chr.5: 175522212-175522212 on Build GRCh38
rs750576991 Chr.5: 175521984-175521984 on Build GRCh38
rs560263804 Chr.5: 175521935-175521935 on Build GRCh38
rs368480639 Chr.5: 175522024-175522024 on Build GRCh38
rs368210548 Chr.5: 175521927-175521927 on Build GRCh38
rs998424923 Chr.5: 175521839-175521839 on Build GRCh38
rs757560082 Chr.5: 175521952-175521952 on Build GRCh38
rs909638160 Chr.5: 175522070-175522070 on Build GRCh38
rs157475 Chr.5: 175522203-175522203 on Build GRCh38
rs767443925 Chr.5: 175521941-175521941 on Build GRCh38
rs2271210 Chr.5: 175522115-175522115 on Build GRCh38
rs549569666 Chr.5: 175522149-175522149 on Build GRCh38
rs764391057 Chr.5: 175521938-175521938 on Build GRCh38
rs1025599096 Chr.5: 175522265-175522265 on Build GRCh38
rs1034147996 Chr.5: 175521994-175521994 on Build GRCh38
rs1032764032 Chr.5: 175522102-175522102 on Build GRCh38
rs1022219376 Chr.5: 175522154-175522154 on Build GRCh38
rs957433449 Chr.5: 175521909-175521909 on Build GRCh38
rs754056882 Chr.5: 175521942-175521942 on Build GRCh38
rs1030974048 Chr.5: 175521861-175521861 on Build GRCh38
rs140616080 Chr.5: 175522093-175522093 on Build GRCh38
rs111866380 Chr.5: 175522075-175522075 on Build GRCh38
rs6892767 Chr.5: 175521813-175521813 on Build GRCh38
rs141630104 Chr.5: 175521953-175521953 on Build GRCh38
rs776214431 Chr.5: 175521924-175521924 on Build GRCh38
rs34907038 Chr.5: 175521940-175521940 on Build GRCh38
rs771788536 Chr.5: 175521921-175521922 on Build GRCh38
rs998521100 Chr.5: 175522101-175522101 on Build GRCh38
rs375878361 Chr.5: 175521864-175521864 on Build GRCh38
rs375020944 Chr.5: 175522001-175522001 on Build GRCh38
rs1052658263 Chr.5: 175522122-175522122 on Build GRCh38
rs755274839 Chr.5: 175522015-175522015 on Build GRCh38
rs762785906 Chr.5: 175521937-175521937 on Build GRCh38
rs1032772067 Chr.5: 175521891-175521891 on Build GRCh38
rs780273305 Chr.5: 175521878-175521878 on Build GRCh38
rs983779072 Chr.5: 175522051-175522051 on Build GRCh38
rs1018630869 Chr.5: 175521856-175521856 on Build GRCh38
rs902862287 Chr.5: 175522097-175522097 on Build GRCh38
rs545640961 Chr.5: 175522071-175522071 on Build GRCh38
rs1008498592 Chr.5: 175522092-175522092 on Build GRCh38
rs754926747 Chr.5: 175522234-175522234 on Build GRCh38
rs756503682 Chr.5: 175522215-175522215 on Build GRCh38
rs761464814 Chr.5: 175521926-175521926 on Build GRCh38
rs781539237 Chr.5: 175522017-175522017 on Build GRCh38
rs747490092 Chr.5: 175522009-175522009 on Build GRCh38
rs905278554 Chr.5: 175522161-175522161 on Build GRCh38
rs879064522 Chr.5: 175522033-175522033 on Build GRCh38
rs1010896813 Chr.5: 175522147-175522147 on Build GRCh38
rs992732656 Chr.5: 175522021-175522021 on Build GRCh38
rs113418476 Chr.5: 175522186-175522186 on Build GRCh38
rs1002272079 Chr.5: 175522194-175522194 on Build GRCh38
rs769036478 Chr.5: 175521903-175521903 on Build GRCh38
rs932817689 Chr.5: 175522237-175522237 on Build GRCh38
rs375679754 Chr.5: 175521877-175521877 on Build GRCh38
rs977151385 Chr.5: 175521871-175521871 on Build GRCh38
rs62388075 Chr.5: 175522264-175522264 on Build GRCh38
rs991763253 Chr.5: 175522125-175522125 on Build GRCh38
rs111494077 Chr.5: 175522138-175522138 on Build GRCh38
rs891394056 Chr.5: 175522042-175522042 on Build GRCh38
rs991420975 Chr.5: 175521930-175521930 on Build GRCh38
rs747460562 Chr.5: 175521901-175521901 on Build GRCh38
rs981370867 Chr.5: 175522160-175522160 on Build GRCh38
rs747968030 Chr.5: 175521906-175521906 on Build GRCh38
rs548371218 Chr.5: 175522023-175522023 on Build GRCh38

View Full Product Details