Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30952196-30952725 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPCR1
Entrez Gene ID
135656
Gene Name
diffuse panbronchiolitis critical region 1
Gene Aliases
PBLT
UniGene
Hs.631993
Gene Chromosome Location
Chr.6: 30941000-30954221 on Build GRCh38
Transcripts
NM_080870.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5113822 c.1256_1257insG p.S420fs*7 Chr.6: 30952348-30952349 on Build GRCh38
3624439 c.1333C>T p.P445S Chr.6: 30952425-30952425 on Build GRCh38
5569176 c.1218G>A p.T406T Chr.6: 30952310-30952310 on Build GRCh38
4003900 c.371C>T p.T124M Chr.6: 30952309-30952309 on Build GRCh38
1077507 c.1249G>T p.G417* Chr.6: 30952341-30952341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749215662 Chr.6: 30952234-30952234 on Build GRCh38
rs753273980 Chr.6: 30952407-30952407 on Build GRCh38
rs981608334 Chr.6: 30952335-30952335 on Build GRCh38
rs991233368 Chr.6: 30952647-30952647 on Build GRCh38
rs267600946 Chr.6: 30952406-30952406 on Build GRCh38
rs771583897 Chr.6: 30952383-30952383 on Build GRCh38
rs899900064 Chr.6: 30952489-30952489 on Build GRCh38
rs996874986 Chr.6: 30952496-30952496 on Build GRCh38
rs55710655 Chr.6: 30952270-30952270 on Build GRCh38
rs79792575 Chr.6: 30952309-30952309 on Build GRCh38
rs766673062 Chr.6: 30952271-30952271 on Build GRCh38
rs542065683 Chr.6: 30952619-30952619 on Build GRCh38
rs753642583 Chr.6: 30952437-30952437 on Build GRCh38
rs1033534027 Chr.6: 30952278-30952278 on Build GRCh38
rs1020788310 Chr.6: 30952697-30952697 on Build GRCh38
rs759999003 Chr.6: 30952289-30952289 on Build GRCh38
rs537037983 Chr.6: 30952393-30952393 on Build GRCh38
rs1038833770 Chr.6: 30952558-30952558 on Build GRCh38
rs746503909 Chr.6: 30952508-30952508 on Build GRCh38
rs760259062 Chr.6: 30952266-30952266 on Build GRCh38
rs112101060 Chr.6: 30952574-30952574 on Build GRCh38
rs777301242 Chr.6: 30952506-30952506 on Build GRCh38
rs769489613 Chr.6: 30952230-30952230 on Build GRCh38
rs1013876133 Chr.6: 30952669-30952669 on Build GRCh38
rs140721086 Chr.6: 30952395-30952395 on Build GRCh38
rs183461489 Chr.6: 30952403-30952403 on Build GRCh38
rs370719305 Chr.6: 30952521-30952521 on Build GRCh38
rs958465792 Chr.6: 30952641-30952641 on Build GRCh38
rs768530965 Chr.6: 30952352-30952352 on Build GRCh38
rs143551263 Chr.6: 30952346-30952346 on Build GRCh38
rs981266851 Chr.6: 30952607-30952607 on Build GRCh38
rs553252346 Chr.6: 30952531-30952531 on Build GRCh38
rs3132580 Chr.6: 30952347-30952347 on Build GRCh38
rs757472150 Chr.6: 30952430-30952430 on Build GRCh38
rs914273391 Chr.6: 30952661-30952661 on Build GRCh38
rs773780783 Chr.6: 30952244-30952244 on Build GRCh38
rs772856191 Chr.6: 30952260-30952260 on Build GRCh38
rs763505954 Chr.6: 30952408-30952408 on Build GRCh38
rs770558502 Chr.6: 30952516-30952516 on Build GRCh38
rs771778110 Chr.6: 30952256-30952256 on Build GRCh38
rs778816176 Chr.6: 30952359-30952359 on Build GRCh38
rs193206474 Chr.6: 30952250-30952250 on Build GRCh38
rs371574542 Chr.6: 30952432-30952432 on Build GRCh38
rs757462262 Chr.6: 30952336-30952336 on Build GRCh38
rs1002090854 Chr.6: 30952273-30952273 on Build GRCh38
rs200921598 Chr.6: 30952231-30952231 on Build GRCh38
rs765764467 Chr.6: 30952305-30952305 on Build GRCh38
rs962202096 Chr.6: 30952584-30952584 on Build GRCh38
rs770309840 Chr.6: 30952523-30952523 on Build GRCh38
rs188616708 Chr.6: 30952487-30952487 on Build GRCh38
rs937994658 Chr.6: 30952578-30952578 on Build GRCh38
rs571239037 Chr.6: 30952238-30952238 on Build GRCh38
rs865833655 Chr.6: 30952483-30952483 on Build GRCh38
rs947038811 Chr.6: 30952675-30952675 on Build GRCh38
rs751853704 Chr.6: 30952325-30952325 on Build GRCh38
rs763595601 Chr.6: 30952542-30952542 on Build GRCh38
rs770583650 Chr.6: 30952394-30952394 on Build GRCh38
rs1055495772 Chr.6: 30952391-30952391 on Build GRCh38
rs751659761 Chr.6: 30952425-30952425 on Build GRCh38
rs565896199 Chr.6: 30952324-30952324 on Build GRCh38
rs903357626 Chr.6: 30952420-30952420 on Build GRCh38
rs765457890 Chr.6: 30952405-30952405 on Build GRCh38
rs1006587896 Chr.6: 30952565-30952565 on Build GRCh38
rs367931222 Chr.6: 30952415-30952415 on Build GRCh38
rs748102841 Chr.6: 30952472-30952472 on Build GRCh38
rs141734710 Chr.6: 30952630-30952630 on Build GRCh38
rs921505615 Chr.6: 30952412-30952412 on Build GRCh38
rs772656235 Chr.6: 30952387-30952387 on Build GRCh38
rs570008856 Chr.6: 30952369-30952369 on Build GRCh38
rs778621895 Chr.6: 30952447-30952447 on Build GRCh38
rs149982150 Chr.6: 30952310-30952310 on Build GRCh38
rs774786589 Chr.6: 30952467-30952469 on Build GRCh38
rs754188367 Chr.6: 30952272-30952272 on Build GRCh38
rs773738458 Chr.6: 30952446-30952446 on Build GRCh38

View Full Product Details