Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31539055-31539552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1G2-DDX39B
Entrez Gene ID
100532737
Gene Name
ATP6V1G2-DDX39B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31546848 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Gene Symbol
DDX39B
Entrez Gene ID
7919
Gene Name
DEAD-box helicase 39B
Gene Aliases
BAT1, D6S81E, UAP56
UniGene
Hs.254042
Gene Chromosome Location
Chr.6: 31530219-31542475 on Build GRCh38
Transcripts
NR_037853.1 NM_004640.6 NM_080598.5 NR_037852.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4676819 c.257T>C p.L86P Chr.6: 31539229-31539229 on Build GRCh38
7196027 c.349_350insG p.T119fs*39 Chr.6: 31539205-31539206 on Build GRCh38
4857250 c.239T>C p.I80T Chr.6: 31539247-31539247 on Build GRCh38
4676821 c.257T>C p.L86P Chr.6: 31539229-31539229 on Build GRCh38
7196024 c.280_281insG p.T96fs*39 Chr.6: 31539205-31539206 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768125874 Chr.6: 31539284-31539284 on Build GRCh38
rs756521014 Chr.6: 31539264-31539264 on Build GRCh38
rs775195232 Chr.6: 31539108-31539108 on Build GRCh38
rs1032432098 Chr.6: 31539219-31539219 on Build GRCh38
rs146591430 Chr.6: 31539383-31539383 on Build GRCh38
rs2071595 Chr.6: 31539285-31539285 on Build GRCh38
rs770351691 Chr.6: 31539424-31539424 on Build GRCh38
rs746801385 Chr.6: 31539487-31539487 on Build GRCh38
rs539689423 Chr.6: 31539410-31539410 on Build GRCh38
rs754377451 Chr.6: 31539321-31539321 on Build GRCh38
rs148931659 Chr.6: 31539278-31539278 on Build GRCh38
rs766929998 Chr.6: 31539158-31539158 on Build GRCh38
rs2523511 Chr.6: 31539077-31539077 on Build GRCh38
rs369901505 Chr.6: 31539491-31539491 on Build GRCh38
rs779310325 Chr.6: 31539083-31539083 on Build GRCh38
rs568633747 Chr.6: 31539342-31539342 on Build GRCh38
rs745792250 Chr.6: 31539100-31539100 on Build GRCh38
rs1054019951 Chr.6: 31539521-31539521 on Build GRCh38
rs774041679 Chr.6: 31539292-31539292 on Build GRCh38
rs563488082 Chr.6: 31539079-31539079 on Build GRCh38
rs1030226900 Chr.6: 31539369-31539369 on Build GRCh38
rs763684827 Chr.6: 31539140-31539140 on Build GRCh38
rs150991114 Chr.6: 31539153-31539153 on Build GRCh38
rs765696501 Chr.6: 31539326-31539326 on Build GRCh38
rs748191884 Chr.6: 31539091-31539091 on Build GRCh38
rs1054478244 Chr.6: 31539328-31539328 on Build GRCh38
rs755185820 Chr.6: 31539080-31539080 on Build GRCh38
rs879229025 Chr.6: 31539119-31539119 on Build GRCh38
rs931312118 Chr.6: 31539367-31539367 on Build GRCh38
rs748820273 Chr.6: 31539452-31539452 on Build GRCh38
rs770533129 Chr.6: 31539514-31539514 on Build GRCh38
rs758717148 Chr.6: 31539286-31539286 on Build GRCh38
rs766945253 Chr.6: 31539320-31539320 on Build GRCh38
rs961774317 Chr.6: 31539438-31539439 on Build GRCh38
rs900053535 Chr.6: 31539352-31539352 on Build GRCh38
rs905442565 Chr.6: 31539096-31539096 on Build GRCh38
rs371761523 Chr.6: 31539287-31539287 on Build GRCh38
rs3219187 Chr.6: 31539098-31539098 on Build GRCh38
rs751909909 Chr.6: 31539297-31539297 on Build GRCh38
rs772529969 Chr.6: 31539301-31539301 on Build GRCh38
rs576904415 Chr.6: 31539522-31539522 on Build GRCh38
rs762764612 Chr.6: 31539112-31539112 on Build GRCh38
rs777243628 Chr.6: 31539439-31539439 on Build GRCh38
rs115959271 Chr.6: 31539361-31539361 on Build GRCh38
rs746273190 Chr.6: 31539283-31539284 on Build GRCh38
rs754216255 Chr.6: 31539283-31539283 on Build GRCh38
rs920470241 Chr.6: 31539213-31539213 on Build GRCh38
rs773967863 Chr.6: 31539142-31539142 on Build GRCh38
rs569114346 Chr.6: 31539387-31539387 on Build GRCh38
rs62395764 Chr.6: 31539280-31539280 on Build GRCh38
rs767211584 Chr.6: 31539298-31539298 on Build GRCh38
rs769831448 Chr.6: 31539103-31539103 on Build GRCh38
rs768406725 Chr.6: 31539292-31539292 on Build GRCh38
rs781487959 Chr.6: 31539282-31539283 on Build GRCh38
rs183581681 Chr.6: 31539290-31539290 on Build GRCh38
rs187497360 Chr.6: 31539294-31539294 on Build GRCh38
rs905328050 Chr.6: 31539460-31539460 on Build GRCh38
rs775426504 Chr.6: 31539288-31539288 on Build GRCh38
rs760264014 Chr.6: 31539309-31539309 on Build GRCh38
rs750567223 Chr.6: 31539099-31539099 on Build GRCh38
rs760164023 Chr.6: 31539322-31539322 on Build GRCh38
rs750007290 Chr.6: 31539167-31539167 on Build GRCh38

View Full Product Details