Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 984305-984798 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Gene Symbol
CYP2W1
Entrez Gene ID
54905
Gene Name
cytochrome P450 family 2 subfamily W member 1
Gene Aliases
-
UniGene
Hs.272795
Gene Chromosome Location
Chr.7: 983157-989640 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4631157 c.329G>A p.R110Q Chr.7: 984566-984566 on Build GRCh38
3394432 c.307G>A p.A103T Chr.7: 984544-984544 on Build GRCh38
6400832 c.308C>A p.A103D Chr.7: 984545-984545 on Build GRCh38
6193415 c.237G>A p.L79L Chr.7: 984474-984474 on Build GRCh38
1453297 c.194C>T p.P65L Chr.7: 984431-984431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370387134 Chr.7: 984602-984602 on Build GRCh38
rs535558848 Chr.7: 984442-984442 on Build GRCh38
rs899112171 Chr.7: 984407-984407 on Build GRCh38
rs1050821611 Chr.7: 984611-984611 on Build GRCh38
rs539780943 Chr.7: 984637-984637 on Build GRCh38
rs781449323 Chr.7: 984382-984382 on Build GRCh38
rs372907345 Chr.7: 984502-984502 on Build GRCh38
rs1006741893 Chr.7: 984516-984516 on Build GRCh38
rs944417002 Chr.7: 984630-984630 on Build GRCh38
rs760828905 Chr.7: 984532-984532 on Build GRCh38
rs752488924 Chr.7: 984487-984487 on Build GRCh38
rs1010573007 Chr.7: 984658-984658 on Build GRCh38
rs1049327933 Chr.7: 984590-984590 on Build GRCh38
rs34743661 Chr.7: 984359-984360 on Build GRCh38
rs890133451 Chr.7: 984431-984431 on Build GRCh38
rs1020083301 Chr.7: 984671-984671 on Build GRCh38
rs1001700850 Chr.7: 984343-984343 on Build GRCh38
rs770347969 Chr.7: 984400-984400 on Build GRCh38
rs554143165 Chr.7: 984399-984399 on Build GRCh38
rs746351743 Chr.7: 984432-984432 on Build GRCh38
rs57806492 Chr.7: 984543-984543 on Build GRCh38
rs760602189 Chr.7: 984605-984605 on Build GRCh38
rs981505541 Chr.7: 984705-984705 on Build GRCh38
rs751441462 Chr.7: 984553-984553 on Build GRCh38
rs370828705 Chr.7: 984460-984460 on Build GRCh38
rs773142415 Chr.7: 984529-984529 on Build GRCh38
rs200427519 Chr.7: 984544-984544 on Build GRCh38
rs758296066 Chr.7: 984488-984488 on Build GRCh38
rs566772597 Chr.7: 984767-984767 on Build GRCh38
rs887958175 Chr.7: 984365-984365 on Build GRCh38
rs957350023 Chr.7: 984654-984654 on Build GRCh38
rs111457738 Chr.7: 984507-984507 on Build GRCh38
rs772134346 Chr.7: 984597-984597 on Build GRCh38
rs73259906 Chr.7: 984483-984483 on Build GRCh38
rs770182157 Chr.7: 984506-984506 on Build GRCh38
rs147129162 Chr.7: 984477-984477 on Build GRCh38
rs989285134 Chr.7: 984441-984441 on Build GRCh38
rs3832539 Chr.7: 984367-984368 on Build GRCh38
rs758099630 Chr.7: 984386-984386 on Build GRCh38
rs774498436 Chr.7: 984514-984514 on Build GRCh38
rs1006542969 Chr.7: 984635-984635 on Build GRCh38
rs749854305 Chr.7: 984574-984574 on Build GRCh38
rs869207906 Chr.7: 984368-984369 on Build GRCh38
rs1027786286 Chr.7: 984359-984359 on Build GRCh38
rs3735689 Chr.7: 984366-984366 on Build GRCh38
rs755681884 Chr.7: 984581-984581 on Build GRCh38
rs971651150 Chr.7: 984679-984679 on Build GRCh38
rs369965540 Chr.7: 984427-984427 on Build GRCh38
rs927970435 Chr.7: 984357-984357 on Build GRCh38
rs111817088 Chr.7: 984333-984333 on Build GRCh38
rs982777649 Chr.7: 984564-984564 on Build GRCh38
rs769014090 Chr.7: 984513-984513 on Build GRCh38
rs571796456 Chr.7: 984384-984384 on Build GRCh38
rs571019656 Chr.7: 984596-984596 on Build GRCh38
rs534442982 Chr.7: 984397-984397 on Build GRCh38
rs767481652 Chr.7: 984476-984476 on Build GRCh38
rs1015366213 Chr.7: 984374-984374 on Build GRCh38
rs374086630 Chr.7: 984501-984501 on Build GRCh38
rs996479309 Chr.7: 984417-984417 on Build GRCh38
rs866636484 Chr.7: 984557-984557 on Build GRCh38
rs746474810 Chr.7: 984398-984398 on Build GRCh38
rs377507861 Chr.7: 984528-984528 on Build GRCh38
rs199628199 Chr.7: 984425-984425 on Build GRCh38
rs74859998 Chr.7: 984558-984558 on Build GRCh38
rs397708660 Chr.7: 984365-984366 on Build GRCh38
rs754839024 Chr.7: 984479-984479 on Build GRCh38
rs765223121 Chr.7: 984533-984533 on Build GRCh38
rs927389167 Chr.7: 984724-984724 on Build GRCh38
rs1002337463 Chr.7: 984662-984662 on Build GRCh38
rs763972171 Chr.7: 984549-984549 on Build GRCh38
rs1010628248 Chr.7: 984653-984653 on Build GRCh38
rs957420485 Chr.7: 984526-984526 on Build GRCh38
rs756831513 Chr.7: 984563-984563 on Build GRCh38
rs774588175 Chr.7: 984470-984470 on Build GRCh38
rs771144248 Chr.7: 984615-984615 on Build GRCh38
rs779217184 Chr.7: 984592-984592 on Build GRCh38
rs545987026 Chr.7: 984500-984500 on Build GRCh38
rs1015586204 Chr.7: 984768-984768 on Build GRCh38
rs960939392 Chr.7: 984740-984740 on Build GRCh38
rs377382764 Chr.7: 984566-984566 on Build GRCh38
rs887927083 Chr.7: 984484-984484 on Build GRCh38
rs201787342 Chr.7: 984366-984367 on Build GRCh38
rs59398243 Chr.7: 984588-984588 on Build GRCh38
rs939335949 Chr.7: 984360-984361 on Build GRCh38
rs376088753 Chr.7: 984591-984591 on Build GRCh38
rs764253603 Chr.7: 984469-984469 on Build GRCh38
rs757483321 Chr.7: 984373-984373 on Build GRCh38
rs546771861 Chr.7: 984684-984684 on Build GRCh38
rs373912411 Chr.7: 984445-984445 on Build GRCh38
rs1050340285 Chr.7: 984421-984421 on Build GRCh38
rs961319877 Chr.7: 984379-984379 on Build GRCh38
rs749614470 Chr.7: 984512-984512 on Build GRCh38
rs769549569 Chr.7: 984612-984613 on Build GRCh38
rs889433219 Chr.7: 984632-984632 on Build GRCh38
rs373124110 Chr.7: 984589-984589 on Build GRCh38
rs761936350 Chr.7: 984519-984519 on Build GRCh38
rs143434274 Chr.7: 984465-984465 on Build GRCh38
rs976477685 Chr.7: 984381-984381 on Build GRCh38
rs937543360 Chr.7: 984480-984480 on Build GRCh38
rs777498080 Chr.7: 984497-984497 on Build GRCh38
rs950086705 Chr.7: 984447-984447 on Build GRCh38
rs1005079033 Chr.7: 984489-984489 on Build GRCh38

View Full Product Details