Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5320425-5320921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNRC18
Entrez Gene ID
84629
Gene Name
trinucleotide repeat containing 18
Gene Aliases
CAGL79, TNRC18A
UniGene
Hs.520638
Gene Chromosome Location
Chr.7: 5306790-5426043 on Build GRCh38
Transcripts
NM_001080495.2 XM_017012728.1 XM_017012729.1 XM_017012730.1 XM_017012731.1 XM_017012732.1 XM_017012733.1 XM_017012734.1 XM_017012735.1 XM_017012736.1 XM_017012737.1 XM_017012738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6110450 c.6619C>T p.Q2207* Chr.7: 5320549-5320549 on Build GRCh38
6805601 c.6595C>T p.R2199W Chr.7: 5320573-5320573 on Build GRCh38
5188330 c.6636+8G>A p.? Chr.7: 5320524-5320524 on Build GRCh38
5622023 c.6562T>C p.Y2188H Chr.7: 5320606-5320606 on Build GRCh38
6805602 c.91C>T p.R31W Chr.7: 5320573-5320573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780322598 Chr.7: 5320656-5320656 on Build GRCh38
rs942984348 Chr.7: 5320494-5320494 on Build GRCh38
rs572163346 Chr.7: 5320834-5320834 on Build GRCh38
rs767308782 Chr.7: 5320507-5320507 on Build GRCh38
rs928596260 Chr.7: 5320603-5320603 on Build GRCh38
rs182028524 Chr.7: 5320867-5320867 on Build GRCh38
rs73673115 Chr.7: 5320820-5320820 on Build GRCh38
rs868104425 Chr.7: 5320702-5320702 on Build GRCh38
rs371289178 Chr.7: 5320508-5320508 on Build GRCh38
rs192715194 Chr.7: 5320601-5320601 on Build GRCh38
rs976927036 Chr.7: 5320868-5320868 on Build GRCh38
rs766269259 Chr.7: 5320511-5320511 on Build GRCh38
rs1036997468 Chr.7: 5320885-5320885 on Build GRCh38
rs1003557518 Chr.7: 5320896-5320896 on Build GRCh38
rs527240456 Chr.7: 5320468-5320468 on Build GRCh38
rs568232040 Chr.7: 5320626-5320626 on Build GRCh38
rs940057340 Chr.7: 5320884-5320884 on Build GRCh38
rs888468480 Chr.7: 5320739-5320739 on Build GRCh38
rs749807425 Chr.7: 5320611-5320611 on Build GRCh38
rs958626424 Chr.7: 5320826-5320826 on Build GRCh38
rs138203393 Chr.7: 5320630-5320630 on Build GRCh38
rs746241160 Chr.7: 5320517-5320517 on Build GRCh38
rs753782370 Chr.7: 5320514-5320514 on Build GRCh38
rs773957907 Chr.7: 5320615-5320615 on Build GRCh38
rs760397778 Chr.7: 5320510-5320510 on Build GRCh38
rs771506227 Chr.7: 5320597-5320597 on Build GRCh38
rs1004205136 Chr.7: 5320788-5320788 on Build GRCh38
rs766141247 Chr.7: 5320631-5320631 on Build GRCh38
rs779810021 Chr.7: 5320643-5320643 on Build GRCh38
rs932817895 Chr.7: 5320879-5320879 on Build GRCh38
rs372197291 Chr.7: 5320627-5320627 on Build GRCh38
rs748766772 Chr.7: 5320495-5320495 on Build GRCh38
rs184246206 Chr.7: 5320474-5320474 on Build GRCh38
rs768249584 Chr.7: 5320613-5320613 on Build GRCh38
rs765192983 Chr.7: 5320640-5320640 on Build GRCh38
rs374605459 Chr.7: 5320515-5320515 on Build GRCh38
rs545970616 Chr.7: 5320847-5320847 on Build GRCh38
rs140885422 Chr.7: 5320799-5320799 on Build GRCh38
rs977106279 Chr.7: 5320858-5320858 on Build GRCh38
rs564051981 Chr.7: 5320857-5320857 on Build GRCh38
rs781467500 Chr.7: 5320459-5320460 on Build GRCh38
rs761597523 Chr.7: 5320503-5320503 on Build GRCh38
rs539405768 Chr.7: 5320644-5320644 on Build GRCh38
rs184327185 Chr.7: 5320728-5320728 on Build GRCh38
rs888698103 Chr.7: 5320883-5320883 on Build GRCh38
rs758579564 Chr.7: 5320875-5320875 on Build GRCh38
rs761498940 Chr.7: 5320850-5320850 on Build GRCh38
rs991789205 Chr.7: 5320829-5320829 on Build GRCh38
rs746251891 Chr.7: 5320487-5320487 on Build GRCh38
rs190790304 Chr.7: 5320745-5320745 on Build GRCh38
rs561854121 Chr.7: 5320869-5320869 on Build GRCh38
rs537727429 Chr.7: 5320766-5320766 on Build GRCh38
rs781294400 Chr.7: 5320529-5320529 on Build GRCh38
rs111839264 Chr.7: 5320800-5320800 on Build GRCh38
rs560013192 Chr.7: 5320464-5320464 on Build GRCh38
rs746187256 Chr.7: 5320546-5320546 on Build GRCh38
rs775877176 Chr.7: 5320492-5320492 on Build GRCh38
rs770289085 Chr.7: 5320491-5320491 on Build GRCh38
rs138391418 Chr.7: 5320881-5320881 on Build GRCh38
rs932854807 Chr.7: 5320678-5320678 on Build GRCh38
rs747680661 Chr.7: 5320621-5320621 on Build GRCh38
rs547280577 Chr.7: 5320890-5320890 on Build GRCh38
rs770269475 Chr.7: 5320520-5320521 on Build GRCh38
rs1021522910 Chr.7: 5320837-5320837 on Build GRCh38
rs747293123 Chr.7: 5320459-5320459 on Build GRCh38
rs565837758 Chr.7: 5320632-5320632 on Build GRCh38
rs367846701 Chr.7: 5320498-5320498 on Build GRCh38
rs757432559 Chr.7: 5320525-5320525 on Build GRCh38
rs900087348 Chr.7: 5320651-5320651 on Build GRCh38
rs756439690 Chr.7: 5320555-5320555 on Build GRCh38
rs756488095 Chr.7: 5320649-5320649 on Build GRCh38
rs751753956 Chr.7: 5320521-5320521 on Build GRCh38
rs531370165 Chr.7: 5320859-5320859 on Build GRCh38
rs73673114 Chr.7: 5320811-5320811 on Build GRCh38
rs750678999 Chr.7: 5320648-5320648 on Build GRCh38
rs903324922 Chr.7: 5320484-5320484 on Build GRCh38
rs758130042 Chr.7: 5320516-5320516 on Build GRCh38
rs187002220 Chr.7: 5320499-5320499 on Build GRCh38
rs764212617 Chr.7: 5320517-5320517 on Build GRCh38
rs777963203 Chr.7: 5320452-5320452 on Build GRCh38

View Full Product Details