Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 5987197-5987687 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PMS2
Entrez Gene ID
5395
Gene Name
PMS1 homolog 2, mismatch repair system component
Gene Aliases
HNPCC4, MLH4, PMS2CL, PMSL2
UniGene
Hs.632637
Gene Chromosome Location
Chr.7: 5970925-6009106 on Build GRCh38
Transcripts
NM_000535.6 NM_001322003.1 NM_001322004.1 NM_001322005.1 NM_001322006.1 NM_001322007.1 NM_001322008.1 NM_001322009.1 NM_001322010.1 NM_001322011.1 NM_001322012.1 NM_001322013.1 NM_001322014.1 NM_001322015.1 NR_136154.1 XM_006715744.3 XM_017012342.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447492 c.1559C>T p.A520V Chr.7: 5987206-5987206 on Build GRCh38
3881947 c.1481C>T p.S494L Chr.7: 5987284-5987284 on Build GRCh38
4855770 c.1321G>A p.E441K Chr.7: 5987444-5987444 on Build GRCh38
179345 c.1261C>T p.R421* Chr.7: 5987504-5987504 on Build GRCh38
3412220 c.1351A>G p.R451G Chr.7: 5987414-5987414 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757730609 Chr.7: 5987568-5987568 on Build GRCh38
rs63750106 Chr.7: 5987458-5987459 on Build GRCh38
rs200726484 Chr.7: 5987456-5987456 on Build GRCh38
rs774325053 Chr.7: 5987529-5987529 on Build GRCh38
rs756335943 Chr.7: 5987382-5987382 on Build GRCh38
rs779306532 Chr.7: 5987393-5987393 on Build GRCh38
rs587778615 Chr.7: 5987404-5987405 on Build GRCh38
rs587782500 Chr.7: 5987285-5987285 on Build GRCh38
rs876658440 Chr.7: 5987606-5987606 on Build GRCh38
rs144389038 Chr.7: 5987333-5987333 on Build GRCh38
rs148642064 Chr.7: 5987288-5987288 on Build GRCh38
rs876661243 Chr.7: 5987620-5987620 on Build GRCh38
rs786202274 Chr.7: 5987216-5987216 on Build GRCh38
rs786202871 Chr.7: 5987533-5987534 on Build GRCh38
rs138387687 Chr.7: 5987522-5987522 on Build GRCh38
rs786202946 Chr.7: 5987409-5987409 on Build GRCh38
rs63751028 Chr.7: 5987246-5987246 on Build GRCh38
rs786204104 Chr.7: 5987580-5987580 on Build GRCh38
rs530105089 Chr.7: 5987292-5987292 on Build GRCh38
rs772659239 Chr.7: 5987404-5987404 on Build GRCh38
rs758048239 Chr.7: 5987525-5987526 on Build GRCh38
rs587780039 Chr.7: 5987596-5987596 on Build GRCh38
rs587782832 Chr.7: 5987533-5987533 on Build GRCh38
rs370701800 Chr.7: 5987520-5987520 on Build GRCh38
rs763954903 Chr.7: 5987460-5987460 on Build GRCh38
rs786203076 Chr.7: 5987295-5987295 on Build GRCh38
rs751973268 Chr.7: 5987633-5987651 on Build GRCh38
rs878854034 Chr.7: 5987439-5987439 on Build GRCh38
rs863224496 Chr.7: 5987468-5987468 on Build GRCh38
rs63750221 Chr.7: 5987227-5987228 on Build GRCh38
rs864622579 Chr.7: 5987342-5987342 on Build GRCh38
rs558565527 Chr.7: 5987631-5987631 on Build GRCh38
rs267608163 Chr.7: 5987257-5987258 on Build GRCh38
rs755348833 Chr.7: 5987392-5987392 on Build GRCh38
rs587779328 Chr.7: 5987302-5987302 on Build GRCh38
rs759151952 Chr.7: 5987265-5987265 on Build GRCh38
rs370752614 Chr.7: 5987525-5987525 on Build GRCh38
rs886062401 Chr.7: 5987406-5987406 on Build GRCh38
rs200116037 Chr.7: 5987381-5987381 on Build GRCh38
rs759692592 Chr.7: 5987548-5987548 on Build GRCh38
rs569947936 Chr.7: 5987411-5987411 on Build GRCh38
rs876658862 Chr.7: 5987567-5987567 on Build GRCh38
rs138697590 Chr.7: 5987445-5987445 on Build GRCh38
rs786203033 Chr.7: 5987427-5987427 on Build GRCh38
rs587780041 Chr.7: 5987526-5987527 on Build GRCh38
rs864622595 Chr.7: 5987574-5987574 on Build GRCh38
rs750448069 Chr.7: 5987379-5987379 on Build GRCh38
rs773048076 Chr.7: 5987286-5987286 on Build GRCh38
rs764854869 Chr.7: 5987292-5987294 on Build GRCh38
rs777920014 Chr.7: 5987257-5987257 on Build GRCh38
rs199739859 Chr.7: 5987275-5987275 on Build GRCh38
rs368516768 Chr.7: 5987255-5987255 on Build GRCh38
rs587780724 Chr.7: 5987389-5987389 on Build GRCh38
rs765384654 Chr.7: 5987651-5987651 on Build GRCh38
rs63750272 Chr.7: 5987243-5987243 on Build GRCh38
rs752287874 Chr.7: 5987319-5987321 on Build GRCh38
rs763694602 Chr.7: 5987223-5987223 on Build GRCh38
rs766400994 Chr.7: 5987545-5987545 on Build GRCh38
rs876661256 Chr.7: 5987420-5987420 on Build GRCh38
rs375905814 Chr.7: 5987266-5987266 on Build GRCh38
rs148956636 Chr.7: 5987462-5987462 on Build GRCh38
rs786203427 Chr.7: 5987348-5987348 on Build GRCh38
rs753856135 Chr.7: 5987250-5987250 on Build GRCh38
rs1805323 Chr.7: 5987311-5987311 on Build GRCh38
rs373917897 Chr.7: 5987326-5987326 on Build GRCh38
rs587781827 Chr.7: 5987341-5987341 on Build GRCh38
rs63750685 Chr.7: 5987328-5987328 on Build GRCh38
rs376344586 Chr.7: 5987330-5987330 on Build GRCh38
rs878854036 Chr.7: 5987225-5987225 on Build GRCh38
rs745306528 Chr.7: 5987582-5987582 on Build GRCh38
rs587780040 Chr.7: 5987532-5987532 on Build GRCh38
rs143865305 Chr.7: 5987280-5987280 on Build GRCh38
rs267608164 Chr.7: 5987232-5987233 on Build GRCh38
rs776194799 Chr.7: 5987412-5987412 on Build GRCh38
rs863224675 Chr.7: 5987579-5987579 on Build GRCh38
rs760727820 Chr.7: 5987466-5987466 on Build GRCh38
rs876660834 Chr.7: 5987413-5987413 on Build GRCh38
rs63751228 Chr.7: 5987354-5987354 on Build GRCh38
rs770421413 Chr.7: 5987350-5987350 on Build GRCh38
rs876660060 Chr.7: 5987300-5987300 on Build GRCh38
rs587778617 Chr.7: 5987504-5987504 on Build GRCh38
rs373611083 Chr.7: 5987366-5987366 on Build GRCh38
rs542522853 Chr.7: 5987298-5987298 on Build GRCh38
rs587781382 Chr.7: 5987477-5987477 on Build GRCh38
rs1004189664 Chr.7: 5987557-5987557 on Build GRCh38
rs876660459 Chr.7: 5987263-5987273 on Build GRCh38
rs776725795 Chr.7: 5987546-5987546 on Build GRCh38
rs116094787 Chr.7: 5987265-5987265 on Build GRCh38
rs747495075 Chr.7: 5987506-5987506 on Build GRCh38
rs192720342 Chr.7: 5987436-5987436 on Build GRCh38
rs74902811 Chr.7: 5987233-5987233 on Build GRCh38
rs878854035 Chr.7: 5987264-5987265 on Build GRCh38
rs786203757 Chr.7: 5987423-5987423 on Build GRCh38
rs533551639 Chr.7: 5987630-5987630 on Build GRCh38
rs587778619 Chr.7: 5987498-5987498 on Build GRCh38
rs2228007 Chr.7: 5987234-5987234 on Build GRCh38
rs63750503 Chr.7: 5987239-5987239 on Build GRCh38
rs587780723 Chr.7: 5987397-5987397 on Build GRCh38
rs786202704 Chr.7: 5987578-5987578 on Build GRCh38
rs587782640 Chr.7: 5987512-5987512 on Build GRCh38
rs762894051 Chr.7: 5987447-5987447 on Build GRCh38
rs754945468 Chr.7: 5987301-5987301 on Build GRCh38
rs141084758 Chr.7: 5987371-5987371 on Build GRCh38
rs749826312 Chr.7: 5987276-5987276 on Build GRCh38
rs778482303 Chr.7: 5987503-5987503 on Build GRCh38
rs752122569 Chr.7: 5987315-5987315 on Build GRCh38
rs587776715 Chr.7: 5987544-5987544 on Build GRCh38
rs754082085 Chr.7: 5987390-5987390 on Build GRCh38
rs540287433 Chr.7: 5987264-5987264 on Build GRCh38
rs587782602 Chr.7: 5987284-5987284 on Build GRCh38
rs786202950 Chr.7: 5987226-5987226 on Build GRCh38
rs573123885 Chr.7: 5987576-5987576 on Build GRCh38
rs587781110 Chr.7: 5987637-5987637 on Build GRCh38
rs755578413 Chr.7: 5987595-5987595 on Build GRCh38
rs752665758 Chr.7: 5987310-5987310 on Build GRCh38
rs63750826 Chr.7: 5987235-5987235 on Build GRCh38
rs587780722 Chr.7: 5987408-5987408 on Build GRCh38
rs587778616 Chr.7: 5987531-5987531 on Build GRCh38
rs267608159 Chr.7: 5987526-5987528 on Build GRCh38
rs748920792 Chr.7: 5987598-5987598 on Build GRCh38
rs373114291 Chr.7: 5987345-5987345 on Build GRCh38
rs876661291 Chr.7: 5987335-5987335 on Build GRCh38
rs587782175 Chr.7: 5987501-5987501 on Build GRCh38
rs150201462 Chr.7: 5987386-5987386 on Build GRCh38
rs576825560 Chr.7: 5987636-5987636 on Build GRCh38
rs864622398 Chr.7: 5987283-5987283 on Build GRCh38
rs146848345 Chr.7: 5987327-5987327 on Build GRCh38
rs142839559 Chr.7: 5987523-5987523 on Build GRCh38
rs756883400 Chr.7: 5987497-5987497 on Build GRCh38
rs542520309 Chr.7: 5987232-5987232 on Build GRCh38
rs63750354 Chr.7: 5987259-5987259 on Build GRCh38
rs921253631 Chr.7: 5987496-5987496 on Build GRCh38
rs376042544 Chr.7: 5987486-5987486 on Build GRCh38
rs1805320 Chr.7: 5987277-5987277 on Build GRCh38
rs780709321 Chr.7: 5987517-5987517 on Build GRCh38
rs147399413 Chr.7: 5987556-5987556 on Build GRCh38
rs766256390 Chr.7: 5987245-5987245 on Build GRCh38
rs864622553 Chr.7: 5987540-5987540 on Build GRCh38
rs1805321 Chr.7: 5987357-5987357 on Build GRCh38
rs878854033 Chr.7: 5987453-5987453 on Build GRCh38
rs748643694 Chr.7: 5987332-5987332 on Build GRCh38
rs876661186 Chr.7: 5987494-5987494 on Build GRCh38
rs570853668 Chr.7: 5987484-5987484 on Build GRCh38
rs112902065 Chr.7: 5987485-5987485 on Build GRCh38
rs876658441 Chr.7: 5987458-5987458 on Build GRCh38
rs768480216 Chr.7: 5987601-5987601 on Build GRCh38
rs138049175 Chr.7: 5987499-5987499 on Build GRCh38
rs746064480 Chr.7: 5987269-5987269 on Build GRCh38
rs748698776 Chr.7: 5987401-5987401 on Build GRCh38
rs775894396 Chr.7: 5987656-5987656 on Build GRCh38
rs899935666 Chr.7: 5987308-5987308 on Build GRCh38
rs587779327 Chr.7: 5987595-5987596 on Build GRCh38
rs752666485 Chr.7: 5987367-5987367 on Build GRCh38
rs759306219 Chr.7: 5987363-5987363 on Build GRCh38
rs267608157 Chr.7: 5987625-5987625 on Build GRCh38
rs766309532 Chr.7: 5987289-5987289 on Build GRCh38
rs148069478 Chr.7: 5987566-5987566 on Build GRCh38
rs587782789 Chr.7: 5987561-5987561 on Build GRCh38
rs864622497 Chr.7: 5987258-5987258 on Build GRCh38
rs770181766 Chr.7: 5987271-5987271 on Build GRCh38
rs759192470 Chr.7: 5987421-5987421 on Build GRCh38
rs786203510 Chr.7: 5987321-5987321 on Build GRCh38
rs536111818 Chr.7: 5987554-5987554 on Build GRCh38

View Full Product Details