Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6140037-6140491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP42
Entrez Gene ID
84132
Gene Name
ubiquitin specific peptidase 42
Gene Aliases
-
UniGene
Hs.31856
Gene Chromosome Location
Chr.7: 6078743-6161564 on Build GRCh38
Transcripts
NM_032172.2 XM_005249883.4 XM_006715790.1 XM_006715791.2 XM_011515573.1 XM_011515574.1 XM_011515577.1 XM_011515578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6027359 c.657-9G>A p.? Chr.7: 6140119-6140119 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748615868 Chr.7: 6140090-6140090 on Build GRCh38
rs73335428 Chr.7: 6140074-6140074 on Build GRCh38
rs1004047495 Chr.7: 6140292-6140292 on Build GRCh38
rs34811274 Chr.7: 6140124-6140124 on Build GRCh38
rs763514616 Chr.7: 6140096-6140096 on Build GRCh38
rs751598676 Chr.7: 6140226-6140226 on Build GRCh38
rs772588747 Chr.7: 6140224-6140225 on Build GRCh38
rs746090816 Chr.7: 6140242-6140242 on Build GRCh38
rs774175231 Chr.7: 6140090-6140090 on Build GRCh38
rs781567916 Chr.7: 6140084-6140084 on Build GRCh38
rs777797383 Chr.7: 6140072-6140072 on Build GRCh38
rs911846113 Chr.7: 6140240-6140240 on Build GRCh38
rs922217572 Chr.7: 6140268-6140268 on Build GRCh38
rs10046499 Chr.7: 6140433-6140433 on Build GRCh38
rs746152213 Chr.7: 6140085-6140085 on Build GRCh38
rs955821607 Chr.7: 6140427-6140427 on Build GRCh38
rs1049261011 Chr.7: 6140319-6140319 on Build GRCh38
rs955033474 Chr.7: 6140388-6140388 on Build GRCh38
rs531681225 Chr.7: 6140278-6140278 on Build GRCh38
rs1028670920 Chr.7: 6140382-6140382 on Build GRCh38
rs763820 Chr.7: 6140404-6140404 on Build GRCh38
rs759332445 Chr.7: 6140099-6140099 on Build GRCh38
rs760442443 Chr.7: 6140208-6140208 on Build GRCh38
rs1039232820 Chr.7: 6140266-6140266 on Build GRCh38
rs771091597 Chr.7: 6140078-6140078 on Build GRCh38
rs886184193 Chr.7: 6140331-6140331 on Build GRCh38
rs990009768 Chr.7: 6140139-6140139 on Build GRCh38
rs568082295 Chr.7: 6140429-6140429 on Build GRCh38
rs189763464 Chr.7: 6140257-6140257 on Build GRCh38
rs907179391 Chr.7: 6140183-6140183 on Build GRCh38
rs757475270 Chr.7: 6140230-6140230 on Build GRCh38
rs369163067 Chr.7: 6140110-6140110 on Build GRCh38
rs753912389 Chr.7: 6140425-6140425 on Build GRCh38
rs547487969 Chr.7: 6140442-6140442 on Build GRCh38
rs750242421 Chr.7: 6140146-6140146 on Build GRCh38
rs770348434 Chr.7: 6140094-6140094 on Build GRCh38
rs767243418 Chr.7: 6140130-6140130 on Build GRCh38
rs777852271 Chr.7: 6140223-6140223 on Build GRCh38
rs1045489639 Chr.7: 6140432-6140432 on Build GRCh38
rs556277606 Chr.7: 6140092-6140092 on Build GRCh38
rs752587116 Chr.7: 6140270-6140270 on Build GRCh38
rs767183385 Chr.7: 6140236-6140236 on Build GRCh38
rs754885701 Chr.7: 6140218-6140218 on Build GRCh38
rs747150685 Chr.7: 6140241-6140242 on Build GRCh38
rs1047776177 Chr.7: 6140259-6140259 on Build GRCh38
rs962608475 Chr.7: 6140352-6140352 on Build GRCh38
rs1001219857 Chr.7: 6140451-6140451 on Build GRCh38
rs779436653 Chr.7: 6140462-6140462 on Build GRCh38
rs760965341 Chr.7: 6140322-6140322 on Build GRCh38
rs1003300101 Chr.7: 6140238-6140238 on Build GRCh38
rs775707566 Chr.7: 6140331-6140331 on Build GRCh38
rs781214320 Chr.7: 6140234-6140234 on Build GRCh38
rs996790754 Chr.7: 6140354-6140354 on Build GRCh38
rs777693276 Chr.7: 6140234-6140235 on Build GRCh38
rs761583358 Chr.7: 6140118-6140118 on Build GRCh38
rs758537358 Chr.7: 6140069-6140069 on Build GRCh38
rs372642571 Chr.7: 6140210-6140210 on Build GRCh38

View Full Product Details